About this item
These QSP™ low retention pipette tips offer a solution with a hydrophobic inner surface that reduces liquid retention.
- Minimise the formation of films on inside of tip
- Reduce sample retention
- Improve accuracy and precision
- Improve sample recovery
- High clarity pipette tips
- Free of RNase/DNase, DNA, pyrogen
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- LABSTLR102RS-Q
- LABSTLR112NXLRS-Q
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- 环境认证:
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)