ContactUsLinkComponent
Noise Protection Boxes, Elma Mutebox
Noise Protection Boxes, Elma Mutebox
  ELMA1111901-001
undefined
Noise Protection Boxes, Elma Mutebox
  ELMA1111901-001
  • 说明:
    Elma Mutebox ML
  • 顺序:
    01
  • 宽度x深度x高度:
    Ext.: 545×530×525 mm; Int.: 430×380×445 mm
  • 重量:
    18,9 kg
  • 包装规格:
    1
  • 用于:
    Unit size: 10, 20, 30, 40, 60, 100, 120 or 180
Elma Mutebox facilitates optimum cleaning, every time. It is hygienic, low-noise and environmentally friendly.
  • Noise of ultrasonic cleaning unit operated in the Mutebox is reduced by up to 85%
  • Vapors are exhausted via a silenced drain, so no heat or moisture builds up inside the box
  • Laterally arranged halogen lamps
  • Stainless steel jacket
Ingeniously innovative, Elma Mutebox guarantees low-noise cleaning. Its quiet fan and optimal insulation make working with ultrasonic equipment a pleasant, health-friendly and quiet experience, whether for cleaning or sample preparation. In addition, the fan extracts condensation water and vapours to the rear. Noise levels are reduced by up to 85% with the Elma Mutebox (ie. by a further 30% compared to the previous model).
The interior lighting and glass door allow the user to keep an eye on the cleaning process at all times. Two independent access points ensure that the items being cleaned and the ultrasonic unit are always easily accessible. The Mutebox, is made of high-quality steel and is easy to clean, thus also ensuring hygienic operation.
 : Ultrasonic cleaning device must be ordered separately.
 : The Elma Mutebox ML is supplied with 1× bottom tray with gland plate, 1× power supply unit and 1× manual. The Elma Mutebox XL includes the same as the ML unit and in addition has another bottom tray with gland plate and 1× splash plate, 1×power supply unit and 1× manual.