[EN]MASTERSENSE AUX/BARE WIRE ADAPTER 1 * 1 件
目录编号 MFLA07526-99
规格
- 最大压力:100 psi
- 说明:Masterflex® universal pressure sensor adaptor cable, bare wire- MasterSense™ pump systems, 6' cable
- 包装规格:1
- Included accessories:6' cable
规格
关于此项目
Adapts a variety of sensors to the MasterSense™ Sensor AUX input.
- Designed for use with 0-20/4-20 mA or 0-5,0-10 V sensors
- Includes 24VDC power out pins for loop powered circuits up to 50 mA
Connection cable allows for integration of variety of sensors into Masterflex® MasterSense™ pump systems. Reference MasterSense™ instruction manual for wiring diagrams and detail.
- 最大压力:100 psi
- 说明:Masterflex® universal pressure sensor adaptor cable, bare wire- MasterSense™ pump systems, 6' cable
- Environmentally Preferable:
- 包装规格:1
- Included accessories:6' cable