Orders LinkContactUsLinkComponent
Bucket, Rectangular
Bucket, Rectangular
11, 25 l桶, 灰色, 22 L
  VILE131662
Bucket, Rectangular
  VILE131662
Bucket, Rectangular
Bucket, Rectangular
11, 25 l桶, 灰色, 22 L
1 * 1 件
Polypropylene bucket with stainless steel handle. For use on the CE Bucket Trolley or the CE pre-prepared trolley.
  • Liquid measurement lines molded inside of bucket
  • Color coding clips included to allow differentiation in colours of buckets (red, blue, green, yellow)
  • Autoclavable
Highest performance for the best contamination control.
  • 容量:
    22 L
  • 颜色:
    灰色
  • 说明:
    25 l桶
  • 顺序:
    11