384-well high content screening microplates with glass bottom
CORN4581
- 孔数量:384
- 灭菌:+
- 颜色:黑色/透明
- 说明:384-well microplate, black with flat, optical glass bottom, with lid, individually wrapped
- 顺序:04
- Environmentally Preferable:Low Manufacturing Impact
- 环境认证:RECs, ISO 14001 - Kennebunk
- 包装规格:10
High optical quality, glass bottom black microplates are ideal for performing high-content cell-based assays using imaging systems. The glass bottom provides a flat and optically clear surface that reduces autofocus time, increases throughput, and is ideal for cell growth.
- High optical quality and scratch resistant glass
- Glass bottom thickness of 200 µm is well suited for imaging microscopy
- Bottom flatness <50 µm to ensure planarity for imaging devices
- Low background fluorescence and minimal crosstalk provide the highest possible optical quality for cell-based assays