Chromatography columns, pre-packed
供应商: GE HEALTHCARE
证书
关于此项目
Prepacked validation columns are suitable for scale-down studies such as viral clearance.
- High-quality packing
- Excellent dimensions for scale-down studies
- Prepacked with the most commonly used BioProcess resins
- High consistency between columns in terms of efficiency and asymmetry
Results of Analysis certificate available for each individual column - includes column performance testing such as asymmetry and efficiency. Developed together with biopharmaceutical experts to ensure optimal function for scale-down studies such as viral clearance.
They feature a high-quality packed bed, enabling robust and reproducible process development or validation. Columns available for custom packing include Tricorn, XK, and HiScale.
规格
- Catalog No:
- GEHE29321910
- GEHE29315195
- GEHE29321909
- pH 范围:
- 2 - 14
- 2 - 14
- 2 - 14
- 存储:
- 2...8 °C, 20% ethanol
- 2...8 °C, 20% ethanol
- 2...8 °C, 20% ethanol