ContactUsLinkComponent
Glass snap top vials, 2 ml, level 3 high performance applications, SureSTART™
Glass snap top vials, 2 ml, level 3 high performance applications, SureSTART™
目录编号 HYPE6PRV11-1P
undefined
Glass snap top vials, 2 ml, level 3 high performance applications, SureSTART™
目录编号 HYPE6PRV11-1P
规格
  • Cap material:
    PP
  • 盖尺寸:
    11 mm
  • 直径×高度:
    12×32 mm
  • 包装规格:
    100
  • 容量:
    2,00 ml
  • 颜色:
    透明
  • 说明:
    Snap vial (wide opening) with write-on patch
  • 材质:
    玻璃
  • 顺序:
    01
  • Cap type:
    Snap
  • 盖颜色:
    Colour-21390420|Colour-21390392
规格
关于此项目
Choose Thermo Scientific™ SureSTART™ 2 ml glass snap top vials, performance level 3, for high performance applications, when sensitivity is a must, and when you are running many samples.
Our level 3 vials are our highest performance vials, designed to protect your most valuable samples and deliver consistent, reproducible results for your most demanding analyses. These 2 ml clear glass snap vials are specification certified to ensure their physical dimensions and autosampler compatibility. Use 11 mm snap caps or 11 mm crimp caps to seal this vial. Our snap caps provide a very tight seal due to their soft polyethylene (PE) material and double snap function. These vials meet all requirements of US, EU, JPN Pharmacopeia. Buy in bulk for greater savings.