Masterflex® Versatile Carboy Cap Adapter Inserts, Avantor®
Versatile Cap Adapter Insert, 83B, Autoclavable, 3 x 1/4" ID Quick Connect Ports, PP, 白色, 83B
目录编号 MFLZ06058-54
供应商: MASTERFLEX
Jump to:
Product Details & Documents
Interchangeable ports for flexibility.
- Choose from a variety of sizes and configurations
- Change the top connections of each carboy to match your application needs
- Autoclavable
Versatile carboy cap adapter inserts are made of autoclavable polypropylene or clear polycarbonate. Quick-connect hosebarb fittings are also available. Vent ports are 1/4" in diameter.
规格
- Cap material:PP
- 盖尺寸:83B
- 说明:Versatile Cap Adapter Insert, 83B, Autoclavable, 3 x 1/4" ID Quick Connect Ports
- 盖颜色:白色