关于此项目
Built-in compartment for storage of PCR rotor and adapters.
- Eight position microtube rotor
- Storage compartment for PCR rotor
- Near silent operation
- Starts and stops in seconds
- Conserves valuable bench space
This personal-sized microcentrifuge allows for a quick spin down of microtubes and PCR tubes. Rotor changeover is quick and easy—no need for set screws and tools. Features transparent blue lid.
- Environmentally Preferable:
Some Products May Appear Restricted