Masterflex® Ismatec® Pump Tubing, 2-Stop Color-Coded Autoanalysis/Microbore, Platinum-Cured Silicone, Avantor®
Supplier: MASTERFLEX PROPRIETARY
Certificates
About this item
Designed for optimal operation in your Ismatec pumps.
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimise downtime
- Regulatory approvals to match your unique application
- No leachable additives or plasticizers
Appearance: Opaque
Temperature: -70 to 52 °C
Life at 100 rpm: 1000 hrs
The overall length of platinum-cured silicone 95602-series stop tubing is 16" (406 mm). The legacy Ismatec® stop tubing was 400 mm long.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLA95602-44
- MFLA95602-40
- MFLA95602-34
- MFLA95602-22
- MFLA95602-32
- MFLA95602-30
- MFLA95602-48
- MFLA95602-42
- MFLA95602-26
- MFLA95602-38
- MFLA95602-28
- MFLA95602-24
- MFLA95602-36
- 型号:
- 95602-44
- 95602-40
- 95602-22
- 95602-32
- 95602-30
- Fixed RPM:
- 15 rpm
- 15 rpm
- 15 rpm
- 15 rpm
- 15 rpm
- 15 rpm
- 15 rpm
- 15 rpm
- 15 rpm
- 15 rpm
- 15 rpm
- 15 rpm
- 15 rpm
- Max. flow:
- 240 ml/min
- 240 ml/min
- 240 ml/min
- 240 ml/min
- 240 ml/min
- 240 ml/min
- 240 ml/min
- 240 ml/min
- 240 ml/min
- 240 ml/min
- 240 ml/min
- 240 ml/min
- 240 ml/min
- Min. flow:
- 0,11 ml/min
- 0,11 ml/min
- 0,11 ml/min
- 0,11 ml/min
- 0,11 ml/min
- 0,11 ml/min
- 0,11 ml/min
- 0,11 ml/min
- 0,11 ml/min
- 0,11 ml/min
- 0,11 ml/min
- 0,11 ml/min
- 0,11 ml/min
- Max. RPM:
- 0,62 rpm
- 0,43 rpm
- 0,026 rpm
- 0,006 rpm
- 0,022 rpm
- 0,017 rpm
- 0,88 rpm
- 0,52 rpm
- 0,011 rpm
- 0,035 rpm
- 0,014 rpm
- 0,009 rpm
- 0,03 rpm
- 系列:
- Similar to SC0632
- Similar to SC0630
- Similar to SC0627
- Similar to SC0621
- Similar to SC0626
- Similar to SC0625
- Similar to SC0634
- Similar to SC0631
- Similar to SC0623
- Similar to SC0629
- Similar to SC0624
- Similar to SC0622
- Similar to SC0628
- Tubes per pack:
- 6
- 6
- 6
- 6
- 6
- 6
- 6
- 6
- 6
- 6
- 6
- 6
- 6