用于气体和水的 Masterflex® 丙烯酸流量计,Avantor®
关于此项目
为使用多种气体的实验室和工厂工艺而设计。
- 英制和公制刻度
这款多功能丙烯酸流量计是专为空气、氧气、氮气、二氧化碳、氩气、氦气或水而设计的。这种流量计分为带或不带内置低滞后针阀等两种款式。不带针阀的流量计可选择面板安装,也可采用部分或完全管线内安装配置。这些流量计可轻松拆卸进行清洗。流量计配备一个稳定的、易于读取的浮标。
警告:本产品未获准、不适用于且不得用于医疗、临床、手术或其他面向患者的应用领域。
27 Options Available Below
Product Details & Documents
为使用多种气体的实验室和工厂工艺而设计。
- 英制和公制刻度
这款多功能丙烯酸流量计是专为空气、氧气、氮气、二氧化碳、氩气、氦气或水而设计的。这种流量计分为带或不带内置低滞后针阀等两种款式。不带针阀的流量计可选择面板安装,也可采用部分或完全管线内安装配置。这些流量计可轻松拆卸进行清洗。流量计配备一个稳定的、易于读取的浮标。
警告:本产品未获准、不适用于且不得用于医疗、临床、手术或其他面向患者的应用领域。
Specifications for Product Family
规格
- Catalog No:
- MFLA68560-00
- MFLA68560-01
- MFLA68560-02
- MFLB68560-03
- MFLA68560-04
- MFLA68560-06
- MFLA68560-07
- MFLA68560-08
- MFLA68560-09
- MFLA68560-10
- MFLA68560-11
- MFLB68560-12
- MFLA68560-13
- MFLA68560-14
- MFLA68560-15
- MFLA68560-16
- MFLA68560-17
- MFLA68560-18
- MFLA68560-19
- MFLA68560-20
- MFLA68560-21
- MFLA68560-22
- MFLA68560-23
- MFLA68560-24
- MFLA68560-25
- MFLA68560-26
- MFLA68560-27
- 型号:
- 68560-00
- 68560-01
- 68560-02
- 68560-03
- 68560-04
- 68560-06
- 68560-07
- 68560-08
- 68560-09
- 68560-10
- 68560-11
- 68560-12
- 68560-13
- 68560-14
- 68560-15
- 68560-16
- 68560-17
- 68560-18
- 68560-19
- 68560-20
- 68560-21
- 68560-22
- 68560-23
- 68560-24
- 68560-25
- 68560-26
- 68560-27
- 准确度:
- ±5% full-scale
- ±5% full-scale
- ±5% full-scale
- ±5% full-scale
- ±5% full-scale
- ±5% full-scale
- ±5% full-scale
- ±5% full-scale
- ±5% full-scale
- ±5% full-scale
- ±5% full-scale
- ±5% full-scale
- ±5% full-scale
- ±5% full-scale
- ±5% full-scale
- ±5% full-scale
- ±5% full-scale
- ±5% full-scale
- ±5% full-scale
- ±5% full-scale
- ±5% full-scale
- ±5% full-scale
- ±5% full-scale
- ±5% full-scale
- ±5% full-scale
- ±5% full-scale
- ±5% full-scale
- 最高运行温度:
- 65 °C (150 °F)
- 65 °C (150 °F)
- 65 °C (150 °F)
- 65 °C (150 °F)
- 65 °C (150 °F)
- 65 °C (150 °F)
- 65 °C (150 °F)
- 65 °C (150 °F)
- 65 °C (150 °F)
- 65 °C (150 °F)
- 65 °C (150 °F)
- 65 °C (150 °F)
- 65 °C (150 °F)
- 65 °C (150 °F)
- 65 °C (150 °F)
- 65 °C (150 °F)
- 65 °C (150 °F)
- 65 °C (150 °F)
- 65 °C (150 °F)
- 65 °C (150 °F)
- 65 °C (150 °F)
- 65 °C (150 °F)
- 65 °C (150 °F)
- 65 °C (150 °F)
- 65 °C (150 °F)
- 65 °C (150 °F)
- 65 °C (150 °F)
- 最大压力:
- 100 psi
- 100 psi
- 100 psi
- 100 psi
- 100 psi
- 100 psi
- 100 psi
- 100 psi
- 100 psi
- 100 psi
- 100 psi
- 100 psi
- 100 psi
- 100 psi
- 100 psi
- 100 psi
- 100 psi
- 100 psi
- 100 psi
- 100 psi
- 100 psi
- 100 psi
- 100 psi
- 100 psi
- 100 psi
- 100 psi
- 100 psi
- O-Ring material:
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- 浸湿材料:
- Brass
- Brass
- Brass
- Brass
- Brass
- 316 SS
- 316 SS
- 316 SS
- 316 SS
- 316 SS
- 316 SS
- 316 SS
- 316 SS
- Brass
- Brass
- Brass
- Brass
- Brass
- Brass
- Brass
- 316 SS
- 316 SS
- 316 SS
- 316 SS
- 316 SS
- 316 SS
- 316 SS
- 质保:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
规格
Product Family Options
产品信息
- 顺序说明Maximum Flow RateMedia typeValveFitting materialHousing material可用库存Price
- 01Acrylic Flowmeter w/o Valve for Air, Brass, 1,4 L/min1,4 L/minAirNoBrassAcrylicLog in 查看库存1 * 1 件规格:
More
Specifications
Cat. No.MFLA68560-00型号68560-00准确度±5% full-scale最高运行温度65 °C (150 °F)最大压力100 psiO-Ring materialViton®浸湿材料Brass质保1 year - 02Acrylic Flowmeter w/o Valve for Air, Brass, 2,75 L/min2,75 L/minAirNoBrassAcrylicLog in 查看库存1 * 1 件规格:
More
Specifications
Cat. No.MFLA68560-01型号68560-01准确度±5% full-scale最高运行温度65 °C (150 °F)最大压力100 psiO-Ring materialViton®浸湿材料Brass质保1 year - 03Acrylic Flowmeter w/o Valve for Air, Brass, 3,5 L/min3,5 L/minAirNoBrassAcrylicLog in 查看库存1 * 1 件规格:
More
Specifications
Cat. No.MFLA68560-02型号68560-02准确度±5% full-scale最高运行温度65 °C (150 °F)最大压力100 psiO-Ring materialViton®浸湿材料Brass质保1 year - 04Acrylic Flowmeter w/o Valve for Air, Brass, 8,5 L/min8,5 L/minAirNoBrassAcrylicLog in 查看库存1 * 1 件规格:
More
Specifications
Cat. No.MFLB68560-03型号68560-03准确度±5% full-scale最高运行温度65 °C (150 °F)最大压力100 psiO-Ring materialViton®浸湿材料Brass质保1 year - 规格:
More
Specifications
Cat. No.MFLA68560-04型号68560-04准确度±5% full-scale最高运行温度65 °C (150 °F)最大压力100 psiO-Ring materialViton®浸湿材料Brass质保1 year - 规格:
More
Specifications
Cat. No.MFLA68560-06型号68560-06准确度±5% full-scale最高运行温度65 °C (150 °F)最大压力100 psiO-Ring materialViton®浸湿材料316 SS质保1 year - 07Acrylic Flowmeter w/o Valve for Air, SS, 1,4 L/min1,4 L/minAirNo316 stainless steelAcrylicLog in 查看库存1 * 1 件规格:
More
Specifications
Cat. No.MFLA68560-07型号68560-07准确度±5% full-scale最高运行温度65 °C (150 °F)最大压力100 psiO-Ring materialViton®浸湿材料316 SS质保1 year - 08Acrylic Flowmeter w/o Valve for Air, SS, 2,75 L/min2,75 L/minAirNo316 stainless steelAcrylicLog in 查看库存1 * 1 件规格:
More
Specifications
Cat. No.MFLA68560-08型号68560-08准确度±5% full-scale最高运行温度65 °C (150 °F)最大压力100 psiO-Ring materialViton®浸湿材料316 SS质保1 year - 09Acrylic Flowmeter w/o Valve for Air, SS, 3,5 L/min3,5 L/minAirNo316 stainless steelAcrylicLog in 查看库存1 * 1 件规格:
More
Specifications
Cat. No.MFLA68560-09型号68560-09准确度±5% full-scale最高运行温度65 °C (150 °F)最大压力100 psiO-Ring materialViton®浸湿材料316 SS质保1 year - 10Acrylic Flowmeter w/o Valve for Air, SS, 8,5 L/min8,5 L/minAirNo316 stainless steelAcrylicLog in 查看库存1 * 1 件规格:
More
Specifications
Cat. No.MFLA68560-10型号68560-10准确度±5% full-scale最高运行温度65 °C (150 °F)最大压力100 psiO-Ring materialViton®浸湿材料316 SS质保1 year - 11Acrylic Flowmeter w/o Valve for Air, SS, 16 L/min16 L/minAirNo316 stainless steelAcrylicLog in 查看库存1 * 1 件规格:
More
Specifications
Cat. No.MFLA68560-11型号68560-11准确度±5% full-scale最高运行温度65 °C (150 °F)最大压力100 psiO-Ring materialViton®浸湿材料316 SS质保1 year - 12Acrylic Flowmeter w/o Valve for Air, SS, 22 L/min22 L/minAirNo316 stainless steelAcrylicLog in 查看库存1 * 1 件规格:
More
Specifications
Cat. No.MFLB68560-12型号68560-12准确度±5% full-scale最高运行温度65 °C (150 °F)最大压力100 psiO-Ring materialViton®浸湿材料316 SS质保1 year - 13Acrylic Flowmeter w/o Valve for Air, SS, 50 L/min50 L/minAirNo316 stainless steelAcrylicLog in 查看库存1 * 1 件规格:
More
Specifications
Cat. No.MFLA68560-13型号68560-13准确度±5% full-scale最高运行温度65 °C (150 °F)最大压力100 psiO-Ring materialViton®浸湿材料316 SS质保1 year - 规格:
More
Specifications
Cat. No.MFLA68560-14型号68560-14准确度±5% full-scale最高运行温度65 °C (150 °F)最大压力100 psiO-Ring materialViton®浸湿材料Brass质保1 year - 15Acrylic Flowmeter w/ Valve for Air, Brass, 2,75 L/min2,75 L/minAirYesBrassAcrylicLog in 查看库存1 * 1 件规格:
More
Specifications
Cat. No.MFLA68560-15型号68560-15准确度±5% full-scale最高运行温度65 °C (150 °F)最大压力100 psiO-Ring materialViton®浸湿材料Brass质保1 year - 16Acrylic Flowmeter w/ Valve for Air, Brass, 3,5 L/min3,5 L/minAirYesBrassAcrylicLog in 查看库存1 * 1 件规格:
More
Specifications
Cat. No.MFLA68560-16型号68560-16准确度±5% full-scale最高运行温度65 °C (150 °F)最大压力100 psiO-Ring materialViton®浸湿材料Brass质保1 year - 17Acrylic Flowmeter w/ Valve for Air, Brass, 8,5 L/min8,5 L/minAirYesBrassAcrylicLog in 查看库存1 * 1 件规格:
More
Specifications
Cat. No.MFLA68560-17型号68560-17准确度±5% full-scale最高运行温度65 °C (150 °F)最大压力100 psiO-Ring materialViton®浸湿材料Brass质保1 year - 规格:
More
Specifications
Cat. No.MFLA68560-18型号68560-18准确度±5% full-scale最高运行温度65 °C (150 °F)最大压力100 psiO-Ring materialViton®浸湿材料Brass质保1 year - 规格:
More
Specifications
Cat. No.MFLA68560-19型号68560-19准确度±5% full-scale最高运行温度65 °C (150 °F)最大压力100 psiO-Ring materialViton®浸湿材料Brass质保1 year - 规格:
More
Specifications
Cat. No.MFLA68560-20型号68560-20准确度±5% full-scale最高运行温度65 °C (150 °F)最大压力100 psiO-Ring materialViton®浸湿材料Brass质保1 year - 21Acrylic Flowmeter w/ Valve for Air, SS, 1,4 L/min1,4 L/minAirYes316 stainless steelAcrylicLog in 查看库存1 * 1 件规格:
More
Specifications
Cat. No.MFLA68560-21型号68560-21准确度±5% full-scale最高运行温度65 °C (150 °F)最大压力100 psiO-Ring materialViton®浸湿材料316 SS质保1 year - 22Acrylic Flowmeter w/ Valve for Air, SS, 2,75 L/min2,75 L/minAirYes316 stainless steelAcrylicLog in 查看库存1 * 1 件规格:
More
Specifications
Cat. No.MFLA68560-22型号68560-22准确度±5% full-scale最高运行温度65 °C (150 °F)最大压力100 psiO-Ring materialViton®浸湿材料316 SS质保1 year - 23Acrylic Flowmeter w/ Valve for Air, SS, 3,5 L/min3,5 L/minAirYes316 stainless steelAcrylicLog in 查看库存1 * 1 件规格:
More
Specifications
Cat. No.MFLA68560-23型号68560-23准确度±5% full-scale最高运行温度65 °C (150 °F)最大压力100 psiO-Ring materialViton®浸湿材料316 SS质保1 year - 24Acrylic Flowmeter w/ Valve for Air, SS, 8,5 L/min8,5 L/minAirYes316 stainless steelAcrylicLog in 查看库存1 * 1 件规格:
More
Specifications
Cat. No.MFLA68560-24型号68560-24准确度±5% full-scale最高运行温度65 °C (150 °F)最大压力100 psiO-Ring materialViton®浸湿材料316 SS质保1 year - 25Acrylic Flowmeter With Valve, For Air, 16 L/min, 1/8" NPT(F) SS Connections16000 ml/minAirYes316 stainless steelAcrylicLog in 查看库存1 * 1 件规格:
More
Specifications
Cat. No.MFLA68560-25型号68560-25准确度±5% full-scale最高运行温度65 °C (150 °F)最大压力100 psiO-Ring materialViton®浸湿材料316 SS质保1 year - 26Acrylic Flowmeter With Valve, For Air, 22 L/min, 1/8" NPT(F) SS Connections22000 ml/minAirYes316 stainless steelAcrylicLog in 查看库存1 * 1 件规格:
More
Specifications
Cat. No.MFLA68560-26型号68560-26准确度±5% full-scale最高运行温度65 °C (150 °F)最大压力100 psiO-Ring materialViton®浸湿材料316 SS质保1 year - 27Acrylic Flowmeter With Valve, For Air, 50 L/min, 1/8" NPT(F) SS Connections50000 ml/minAirYes316 stainless steelAcrylicLog in 查看库存1 * 1 件规格:
More
Specifications
Cat. No.MFLA68560-27型号68560-27准确度±5% full-scale最高运行温度65 °C (150 °F)最大压力100 psiO-Ring materialViton®浸湿材料316 SS质保1 year