Orders LinkContactUsLinkComponent
Masterflex® L/S®计算机兼容的数字驱动器,Avantor®
Masterflex® L/S®计算机兼容的数字驱动器,Avantor®
Masterflex® L/S®计算机兼容的数字驱动器,Avantor®
Masterflex® L/S®计算机兼容的数字驱动器,Avantor®
Masterflex® L/S®计算机兼容的数字驱动器,Avantor®
将这款数字驱动器接入电脑,即可用于自动化实验室和自动化的工艺应用;它也可用作独立的分配泵。
  • 精密数字驱动器具有免维护高精度的电机和全新的图形界面,使设置和操作前所未有地轻松
  • 使用L/S泵管,流速范围为0.001 - 3400毫升/分。流速取决于驱动器转速和泵管尺寸。
  • 图形LCD显示四种操作模式,分别是持续运行、定时分配、重复分配和按量分配。可持续查看泵性能。
  • 菜单驱动式界面便于编程和操作,并提供七种语言供您选择
  • 按量分配——以毫升为单位,每次可分配0.001 - 9999毫升;重复分配——可设置1 - 9999个分配周期;或定时分配——每次分配周期的时间间隔(1秒 - 99小时、59分钟、59秒)
  • 无刷免维护电机具有±0.1%的速度控制精度,调节比为6000:1
  • 可通过RS-232或USB接口与电脑连接
  • 通过驱动器上的DB25母接口进行远程控制:
  • --速度控制输入0 - 20 mA、4 - 20 mA,或0 - 10V;
  • --泵送方向要求开集电极或触点闭合;
  • --开启/停止/充液要求开集电极或触点闭合;
  • --流速计输出0 - 20 mA、4 - 20 mA,或0 - 10V,开集电极
这些精密驱动器可进行模拟远程控制,并有简单的编程——是自动化工艺应用的理想选择。驱动器可设置分配间隔,即分配周期间的延迟时间,从而更方便且自动化地进行分配。可编程设置的校准确保了分配和显示的精确度——驱动器为每种泵管尺寸存储有一个校准值。
通过操作简单的覆膜键盘设置驱动器;键盘锁定功能可保护设置。电机可逆,便于在泵送之前或之后冲洗泵管,亦可往两个方向泵送流体。
Ordering information: Each model includes a country specific plug/cord set; choose the catalog number with desired country plug type. All units include an IEC 320 socket with a detachable standard US three prong power cable. See country plug description https://uk.vwr.com/cms/tech-article-ac-power-plugs-sockets-universal-pattern-guide.
Delivery information: 长6英尺(1.8米)的电线,带IEC 320/CEE22插座。这些型号会随附指定国家的插头/电线套装;敬请在订购时注明最终目的国。
  • 型号:
  • 07551-20
  • 07551-30
  • 07551-42
  • Cloud connected:
  • No
  • No
  • Yes
  • 控制类型:
  • Digital
  • Digital
  • Digital Variable Speed
  • Digital dispensing:
  • Yes
  • Yes
  • Yes
  • Drive height:
  • 216 mm (8,5")
  • 216 mm (8,5")
  • 216 mm (8,5")
  • Drive IP rating:
  • IP 33
  • IP 33
  • IP 33
  • Drive length:
  • 254 mm (10")
  • 254 mm (10")
  • 254 mm (10")
  • Drive width:
  • 216 mm (8,5")
  • 216 mm (8,5")
  • 216 mm (8,5")
  • Min. RPM:
  • 0,1 rpm
  • 0,02 rpm
  • 0,1 rpm
  • 电机:
  • 75 W
  • 75 W
  • 75 W
  • Motor hp:
  • 1/10
  • 1/10
  • 1/10
  • Open-head sensor:
  • No
  • No
  • Yes
  • Pump series:
  • L/S
  • L/S
  • L/S
  • Remote I/O:
  • RS-232, USB, 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
  • RS-232, USB, 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
  • Profibus, Bluetooth LE, USB, 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
  • Reversible motor:
  • Yes
  • Yes
  • Yes
  • 速度:
  • Variable
  • Variable
  • Variable
  • Speed control:
  • ±0,1% (0,1 rpm at 600 rpm)
  • ±0,1% (0,01 rpm at 100 rpm)
  • ±0,1% (0,01 rpm at 100 rpm)
  • Type of pump heads accepted:
  • Standard, Easy-Load, Easy-Load II, Easy-Load 3, High Performance, Cartridge, Multichannel, PTFE Tubing, PTFE Diaphragm, Cytoflow
  • Standard, Easy-Load, Easy-Load II, Easy-Load 3, High Performance, Cartridge, Multichannel, PTFE Tubing, PTFE Diaphragm, Cytoflow
  • Standard, Easy-Load, Easy-Load OHS, Easy-Load II, Easy-Load II OHS, High Performance, High Performance OHS, PTFE Tubing, PTFE Diaphragm, Cytoflow
  • Washdown capable:
  • No
  • No
  • No
  • 电流:
  • 2,2 A
  • 2,2 A
  • 2,2 A
  • 频率:
  • 50/60 Hz
  • 50/60 Hz
  • 50/60 Hz
  • 电压:
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 90 - 260 VAC
  • 质保:
  • 2 years
  • 2 years
  • 2 years
Product Family Options
  • 顺序
    说明
    Electrical plug type
    No. of pump heads accepted
    Maximum Flow Rate
    Max. RPM
    Housing material
    Power
  • 11
    L/S® Digital Drive, Computer-Compatible, 0,1 to 600 rpm; 115/230 VAC
    USS
    2
    3400 ml/min
    600 rpm
    ABS plastic and aluminium
    90 - 260 VAC; 50/60 Hz; 2,2 A
    1 * 1 件
    1 * 1 件
    2 year warranty
    CE Compliant
    Specifications
    Cat. No.
    MFLA07551-20-USS
    型号
    07551-20
    Cloud connected
    No
    控制类型
    Digital
    Digital dispensing
    Yes
    Drive height
    216 mm (8,5")
    Drive IP rating
    IP 33
    Drive length
    254 mm (10")
    Drive width
    216 mm (8,5")
    Min. RPM
    0,1 rpm
    电机
    75 W
    Motor hp
    1/10
    Open-head sensor
    No
    Pump series
    L/S
    Remote I/O
    RS-232, USB, 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
    Reversible motor
    Yes
    速度
    Variable
    Speed control
    ±0,1% (0,1 rpm at 600 rpm)
    Type of pump heads accepted
    Standard, Easy-Load, Easy-Load II, Easy-Load 3, High Performance, Cartridge, Multichannel, PTFE Tubing, PTFE Diaphragm, Cytoflow
    Washdown capable
    No
    电流
    2,2 A
    频率
    50/60 Hz
    电压
    90 - 260 VAC
    质保
    2 years
    2 year warranty
    CE Compliant
  • 22
    L/S® Digital Drive, Computer-Compatible, 0,02 to 100 rpm; 115/230 VAC
    USS
    4
    566 ml/min
    100 rpm
    ABS plastic and aluminium
    90 - 260 VAC; 50/60 Hz; 2,2 A
    1 * 1 件
    1 * 1 件
    2 year warranty
    CE Compliant
    Specifications
    Cat. No.
    MFLA07551-30-USS
    型号
    07551-30
    Cloud connected
    No
    控制类型
    Digital
    Digital dispensing
    Yes
    Drive height
    216 mm (8,5")
    Drive IP rating
    IP 33
    Drive length
    254 mm (10")
    Drive width
    216 mm (8,5")
    Min. RPM
    0,02 rpm
    电机
    75 W
    Motor hp
    1/10
    Open-head sensor
    No
    Pump series
    L/S
    Remote I/O
    RS-232, USB, 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
    Reversible motor
    Yes
    速度
    Variable
    Speed control
    ±0,1% (0,01 rpm at 100 rpm)
    Type of pump heads accepted
    Standard, Easy-Load, Easy-Load II, Easy-Load 3, High Performance, Cartridge, Multichannel, PTFE Tubing, PTFE Diaphragm, Cytoflow
    Washdown capable
    No
    电流
    2,2 A
    频率
    50/60 Hz
    电压
    90 - 260 VAC
    质保
    2 years
    2 year warranty
    CE Compliant
  • 44
    L/S Profibus Network-Compatible Digital Drive, 0,1 to 600 rpm, 115/230 VAC
    USS
    2
    3400 ml/min
    600 rpm
    ABS plastic and aluminium
    90 - 260 VAC; 50/60 Hz; 2,2 A
    1 * 1 件
    1 * 1 件
    2 year warranty
    ETL Canada listed
    ETL listed
    Specifications
    Cat. No.
    MFLA07551-42-USS
    型号
    07551-42
    Cloud connected
    Yes
    控制类型
    Digital Variable Speed
    Digital dispensing
    Yes
    Drive height
    216 mm (8,5")
    Drive IP rating
    IP 33
    Drive length
    254 mm (10")
    Drive width
    216 mm (8,5")
    Min. RPM
    0,1 rpm
    电机
    75 W
    Motor hp
    1/10
    Open-head sensor
    Yes
    Pump series
    L/S
    Remote I/O
    Profibus, Bluetooth LE, USB, 0 to 20 mA, 4 to 20 mA, 0 to 10 V, start/stop, direction, and prime via contact closure
    Reversible motor
    Yes
    速度
    Variable
    Speed control
    ±0,1% (0,01 rpm at 100 rpm)
    Type of pump heads accepted
    Standard, Easy-Load, Easy-Load OHS, Easy-Load II, Easy-Load II OHS, High Performance, High Performance OHS, PTFE Tubing, PTFE Diaphragm, Cytoflow
    Washdown capable
    No
    电流
    2,2 A
    频率
    50/60 Hz
    电压
    90 - 260 VAC
    质保
    2 years
    2 year warranty
    ETL Canada listed
    ETL listed
Accessories
  •  
    说明
  • MFLA07523-97
    Handheld Remote Controller; DB25(M) Connector; 6 ft (1,8 m) Cable
    1 * 1 件
  •  
    说明
    颜色
    长度
  • MFLA22050-60
    USB cable; type A male to type B male
    米色
    10 ft.
    1 * 10 Feet
  •  
    说明
  • MFLA22050-54
    Interface cable with DB9 male/DB9 female connectors
    1 * 1 件
  •  
    说明
  • MFLA07523-92
    Footswitch, momentary start/stop, DB25, 6' cable
    1 * 1 件
  •  
    说明
  • MFLA07523-94
    DB25 Male connector; for creating your own cable
    1 * 1 件
  •  
    顺序
    说明
    兼容
  • MFLA77097-12
    3
    Leak Detector Sensor with Plastic Catch Basin
    Masterflex peristaltic pump with Open Head Sensor (OHS) input
    1 * 1 件
  •  
    顺序
    说明
    兼容
  • MFLA77097-10
    1
    Leak Detector Sensor
    Masterflex peristaltic pump with Open Head Sensor (OHS) input
    1 * 1 件