John Guest Polypropylene Push-To-Connect Elbow Adapter Fittings
Supplier: MASTERFLEX PREMIUM
Certificates
About this item
Quick, leakproof connection.
- Polypropylene and EPDM materials meet NSF 51, and NSF 61 requirements for food-grade applications, and is FDA compliant
Push-to-connect fittings offer secure connections and quick disconnection. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth—tubing slides right out.
Fittings work with semi-rigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing.
Adapters let you permanently modify basic fittings for customized installations.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLB34007-60
- MFLB34007-58
- Collet material:
- Stainless Steel
- Stainless Steel
- 最大运行压力:
- 150 psi at 70 °F, 60 psi at 140 °F (for water)
- 150 psi at 70 °F, 60 psi at 140 °F (for water)
- 最高温度:
- 60 °C (140 °F)
- 60 °C (140 °F)
- 最小温度:
- 1 °C (33 °F)
- 1 °C (33 °F)
- O-Ring material:
- EPDM
- EPDM
- 灭菌:
- Autoclave only
- Autoclave only
- 质保:
- 20 years
- 20 years