Masterflex® 聚碳酸酯Luer-接-Luer 接头和Luer插头,Avantor®
Supplier: MASTERFLEX PROPRIETARY
Certificates
About this item
Easily transition to a luer fitting.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
这些优质接头可以轻松完备您的luer接头系统。内含一些聚碳酸酯材质的特殊配置。所有的母端口可以与滑动连接或luer锁连接配套使用。还有插头和帽。
- Environmentally Preferable:
Specifications
- 型号:
- 45501-28
- 45501-22
- 45504-56
- 45501-84
- 45501-82
- 45501-56
- 30526-10
- Connection type (port 1):
- Female luer
- Female luer
- Male luer lock
- Female luer
- Female luer
- Female luer
- Female luer
- Connection type (port 2):
- Cap
- Female luer
- Plug
- Male luer slip
- Male luer slip
- Female luer
- Male luer lock
- Connection type (port 3):
- Female luer
- No. of ports:
- 1
- 2
- 1
- 2
- 2
- 3
- 2
- 质保:
- 3 Months
- 3 Months
- 3 Months
- 3 Months
- 3 Months
- 3 Months
- Out of box