Masterflex® 倒刺Y形接头,Avantor®
Supplier: MASTERFLEX PROPRIETARY
Certificates
About this item
No tools needed for assembly.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Designed for quick and easy assembly with flexible tubing
最适合用于药物、实验室、工业/制造、水样采集、食品加工应用。
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLA30614-67
- MFLA40726-43
- MFLA30614-71
- MFLA40726-44
- MFLA30614-59
- MFLA40726-41
- MFLA30614-63
- MFLA40726-42
- MFLA40726-47
- MFLA40726-48
- MFLA40726-45
- MFLA40727-47
- MFLA40726-46
- MFLB40617-87
- MFLB40617-80
- MFLA06295-40
- MFLB40617-76
- MFLB40617-79
- MFLA40617-77
- MFLA30614-51
- MFLA30614-43
- MFLA40727-44
- MFLA40727-45
- MFLA40727-42
- MFLA40727-43
- MFLA40727-46
- MFLA40726-50
- MFLA41526-28
- MFLA40727-41
- MFLA41526-39
- MFLA40726-49
- 型号:
- 30614-67
- 40726-43
- 30614-71
- 40726-44
- 30614-59
- 40726-41
- 30614-63
- 40726-42
- 40726-47
- 40726-48
- 40726-45
- 40727-47
- 40726-46
- 06295-40
- 30614-51
- 30614-43
- 40727-44
- 40727-45
- 40727-42
- 40727-43
- 40727-46
- 40726-50
- 41526-28
- 40727-41
- 41526-39
- 40726-49
- Orifice dimensions:
- 6 mm
- 灭菌:
- Gamma Irradiation, Autoclave, and EtO
- Gamma Irradiation, Autoclave, and EtO
- Gamma Irradiation, Autoclave, and EtO
- Gamma Irradiation, Autoclave, and EtO
- Gamma Irradiation, Autoclave, and EtO
- Gamma Irradiation, Autoclave, and EtO
- Gamma Irradiation, Autoclave, and EtO
- Gamma Irradiation, Autoclave, and EtO
- Gamma Irradiation, Autoclave, and EtO
- Gamma Irradiation, Autoclave, and EtO
- Gamma Irradiation, Autoclave, and EtO
- Gamma Irradiation, Autoclave, and EtO
- Gamma Irradiation, Autoclave, and EtO
- Autoclave only
- Gamma Irradiation, Autoclave, and EtO
- Gamma Irradiation, Autoclave, and EtO
- Gamma Irradiation, Autoclave, and EtO
- Gamma Irradiation, Autoclave, and EtO
- Gamma Irradiation, Autoclave, and EtO
- Gamma Irradiation, Autoclave, and EtO
- Gamma Irradiation, Autoclave, and EtO
- Gamma Irradiation, Autoclave, and EtO
- Gamma Irradiation, Autoclave, and EtO
- Gamma Irradiation, Autoclave, and EtO
- Gamma Irradiation, Autoclave, and EtO
- Gamma Irradiation, Autoclave, and EtO
- 质保:
- 3 Months
- 3 Months
- 3 Months
- 3 Months
- 3 Months
- 3 Months
- 3 Months
- 3 Months
- 3 Months
- Shelf life
- Shelf life
- 1 year
- Shelf life
- Shelf life
- Shelf life
- 1 year
- 3 Months
- 3 Months
- 3 Months
- 3 Months
- 3 Months
- 3 Months