Masterflex® PVDF 卫生级接头夹具,Avantor®
PVDF Sanitary Clamp, 1" and 1 1/2" —40 to 280 °F, 聚偏二氟乙烯, Sanitary
MFLA31811-01
3A Sanitary approval
Maintain sanitary conditions in your processes.
- 满足药品使用的USP Class VI 要求和食品接触的 FDA, USDA, 和3A 标准
- 所有接头与三头管夹都相容
- 优秀的耐化学性,低滤取特性
These sanitary clamps are compatible with Tri-Clamp® fittings. The PVDF material has excellent chemical resistance with low leachable characteristics.
- 说明:PVDF Sanitary Clamp, 1" and 1 1/2" —40 to 280 °F
- 材质:聚偏二氟乙烯
- Clamp type:Sanitary
- 最高温度:138 °C (280 °F)
- 最大运行压力:150 psi
- Cat. No.:MFLA31811-01
- 灭菌:Autoclave or ethylene oxide (ETO)
- Chemical resistance:Very good
- 质保:1 year
- 最小温度:–40 °C (–40 °F)
- 型号:31811-01
- Sanitary clamp size (port 1):1-1/2"
- 连接类型:Sanitary