Masterflex® USB to DB9 (M) RS-232 Serial Adapter Cable
Catalog # MFLA22050-58
Supplier: MASTERFLEX PROPRIETARY
Specifications
- 说明:USB to DB9 (M) RS-232 serial adapter cable
- 型号:22050-58
- 质保:1 year
- 包装规格:1
Specifications
About this item
Optimize performance of your Masterflex® pump.
- Connect your Masterflex® drive via DB9 (M) RS-232 connection to USB
使用此电缆通过 USB 端口连接 Masterflex 计算机兼容驱动器 07550 和 07551 系列与 PC。将来还可以连接其他配有 RS-232 串行接口的外围设备,包括平衡仪、打印机、扫描仪和 PDA。随附 2 ft 线缆和驱动程序软件。支持高达 12 Mbps 的数据传输速度;兼容 Windows® 2000 / XP / Vista / 7。
- 说明:USB to DB9 (M) RS-232 serial adapter cable
- Environmentally Preferable:
- 型号:22050-58
- 质保:1 year
- 包装规格:1