Masterflex® L/S® Standard Long-Shaft Pump Heads, Avantor®
Supplier: MASTERFLEX PROPRIETARY
Certificates
About this item
Ideal for fluid transfer applications requiring high accuracy and repeatability.
- Standard heads are precision molded to accommodate a single size of tubing
- Pump head is compact and simple in design with same side tubing entry/exit—perfect for space-limited applications
- Standard heads are compact and easy to integrate into your OEM applications
This Standard pump head accepts a single tubing size to ensure optimal performance from your Masterflex pump. Use with Masterflex L/S pump drives requiring the long-shaft style pump head. These pump heads are not compatible with standard shaft drives.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLA07024-10
- MFLA07024-00
- MFLA07015-50
- MFLA07016-10
- MFLA07015-10
- MFLA07015-00
- MFLA07016-00
- MFLA07013-10
- 型号:
- 07024-10
- 07024-00
- 07015-50
- 07016-10
- 07015-10
- 07015-00
- 07016-00
- 07013-10
- Gear ratio:
- 1:1
- 1:1
- 1:1
- 1:1
- 1:1
- 1:1
- 最小流速:
- 2,8 ml/min
- 2,8 ml/min
- 1,7 ml/min
- 0,8 ml/min
- 1,7 ml/min
- 1,7 ml/min
- 0,8 ml/min
- 0,06 ml/min
- 通道数量:
- 1
- 1
- 1
- 1
- 1
- 1
- Occlusion:
- Fixed
- Fixed
- Fixed
- Fixed
- Fixed
- Fixed
- Optimum RPM range:
- 0,02 - 600 rpm
- 0,02 - 600 rpm
- 0,02 - 600 rpm
- 0,02 - 600 rpm
- 0,02 - 600 rpm
- 0,02 - 600 rpm
- Pump series:
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- Tubing retention:
- Manual
- Manual
- Manual
- Manual
- Manual
- Manual
- Tubing sizes accepted:
- L/S® 24
- L/S® 24
- L/S® 15
- L/S® 16
- L/S® 15
- L/S® 15
- L/S® 16
- L/S® 13
- Volume per revolution with L/S 13 tubing:
- 0,06 ml
- Volume per revolution with L/S 15 tubing:
- 1,7 ml
- 1,7 ml
- 1,7 ml
- Volume per revolution with L/S 16 tubing:
- 0,8 ml
- 0,8 ml
- Volume per revolution with L/S 24 tubing:
- 2,8 ml
- 2,8 ml
- 质保:
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years
- 2 years