GF Signet Digital Flow Rate Monitor/Totalizer; Battery Powered
Catalog # MFLB05629-22
Supplier: MASTERFLEX PREMIUM
CE Compliant
CSA Certified
IP65 washdown
UL Listed
ISO9001
Specifications
- 准确度:±0,5% of reading
- 显示器类型:4-digit LCD (upper). Lower: 8-digit LCD
- 说明:3-8150-1P Digital Flow Rate Monitor/ Totalizer Battery Powered
- 电池使用寿命:4 years nominal
- 最高运行温度:55 °C (131 °F)
- 质保:2 years
- 包装规格:1
Specifications
About this item
Use in areas where AC power is not readily available.
- Use in areas where wired power is power is not readily available
Independently calibrate flow and totalizer readings. Monitor includes both fixed and resettable totalizers. Features a four-digit flow LCD, and eight-digit totalizer LCD, automatic contrast adjustment, and low-battery indication. Monitor includes unit/multiplier decals for field calibration.
Operates on two internal lithium batteries (included). Use monitor only with the Roto-X™ and Metalex™ flow sensors sold separately. Note: not for use with the Roto-X™ Low-Flow sensors.
- 准确度:±0,5% of reading
- 显示器类型:4-digit LCD (upper). Lower: 8-digit LCD
- 说明:3-8150-1P Digital Flow Rate Monitor/ Totalizer Battery Powered
- Environmentally Preferable:
- 电池使用寿命:4 years nominal
- 最高运行温度:55 °C (131 °F)
- 质保:2 years
- 包装规格:1