Orders LinkContactUsLinkComponent
Masterflex® 2通常闭电磁挤压阀,Avantor®
Masterflex® 2通常闭电磁挤压阀,Avantor®
Supplier:   MASTERFLEX PREMIUM
Masterflex® 2通常闭电磁挤压阀,Avantor®
Supplier:  MASTERFLEX PREMIUM
Certificates
Masterflex® 2通常闭电磁挤压阀,Avantor®
Masterflex® 2通常闭电磁挤压阀,Avantor®
18 Options Available Below
Product Details & Documents
No contamination of valve.
  • Control flow in your tubing without direct contact between the fluid and the valve
  • Fast response time
  • Low-power consumption and high-cycle life
  • Excellent for sanitary and pharmaceutical applications
Choose from two-way NC, two-way NO, or three-way pinch valves. These are ideal where contamination is an issue, as the fluid only comes in contact with the tubing, not the valve. Tubing replacement is quick and easy, making these suitable for pharmaceutical or sanitary applications. Mounting clip or mounting flange available separately for increased versatility.
Delivery information: 12" 长硅胶管。其他管线单独出售。所有阀门都带有 15" 26 规格的 PTFE 涂层导线
Specifications for Product Family
Specifications
  • 型号:
  • 98302-00
  • 98302-02
  • 98302-06
  • 98302-10
  • 98302-12
  • 98302-16
  • 98302-20
  • 98302-22
  • 98302-26
  • 98302-30
  • 98302-32
  • 98302-36
  • 98302-40
  • 98302-42
  • 98302-46
  • 98302-48
  • 98302-50
  • 98302-54
  • 内管直径:
  • 0,0312"
  • 0,0625"
  • 0,125"
  • 0,0312"
  • 0,0625"
  • 0,125"
  • 0,0625"
  • 0,0312"
  • 0,0625"
  • 0,125"
  • 0,0625"
  • 0,125"
  • 最大压力:
  • 15 psi
  • 15 psi
  • 20 psi
  • 15 psi
  • 15 psi
  • 20 psi
  • 15 psi
  • 15 psi
  • 15 psi
  • 20 psi
  • 15 psi
  • 15 psi
  • 20 psi
  • 20 psi
  • Pinch valve tube size:
  • 1/32" ID x 3/32" OD
  • 1/16" ID x 1/8" OD
  • 1/8" ID x 1/4" OD
  • 1/32" ID x 3/32" OD
  • 1/16" ID x 1/8" OD
  • 1/8" ID x 1/4" OD
  • 1/32" ID x 3/32" OD
  • 1/16" ID x 1/8" OD
  • 1/8" ID x 1/4" OD
  • 1/32" ID x 3/32" OD
  • 1/16" ID x 1/8" OD
  • 1/8" ID x 1/4" OD
  • 1/32" ID x 3/32" OD
  • 1/16" ID x 1/8" OD
  • 1/8" ID x 1/4" OD
  • 1/32" ID x 3/32" OD
  • 1/16" ID x 1/8" OD
  • 1/8" ID x 1/4" OD
  • 功率:
  • 12 VDC, 2,6 W
  • 12 VDC, 2,6 W
  • 12 VDC, 4 W
  • 24 VDC, 2,6 W
  • 24 VDC, 2,6 W
  • 24 VDC, 4 W
  • 12 VDC, 2,6 W
  • 12 VDC, 2,6 W
  • 12 VDC, 4 W
  • 24 VDC, 2,6 W
  • 24 VDC, 2,6 W
  • 24 VDC, 4 W
  • 12 VDC, 2,6 W
  • 12 VDC, 2,6 W
  • 12 VDC, 4 W
  • 24 VDC, 2,6 W
  • 24 VDC, 2,6 W
  • 24 VDC, 4 W
  • 响应时间:
  • 25 ms
  • 25 ms
  • 35 ms
  • 25 ms
  • 25 ms
  • 35 ms
  • 25 ms
  • 25 ms
  • 35 ms
  • 25 ms
  • 25 ms
  • 35 ms
  • 25 ms
  • 25 ms
  • 35 ms
  • 25 ms
  • 25 ms
  • 35 ms
  • 质保:
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
Specifications
Product Family Options
Product Information
  • 顺序
    说明
    Connection Type
    Flow pattern
    Availability
    Price
  • MFLB98302-00
    01
    Two-way normally closed solenoid pinch valve; 12 VDC, 1/32" ID x 3/32" OD tubing
    Pinch Valve for Tubing
    Two-Way, Normally Closed
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
    Specifications:
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLB98302-00
    型号
    98302-00
    内管直径
    0,0312"
    最大压力
    15 psi
    Pinch valve tube size
    1/32" ID x 3/32" OD
    功率
    12 VDC, 2,6 W
    响应时间
    25 ms
    质保
    2 years
    CE Compliant
  • MFLB98302-02
    02
    Two-way normally closed solenoid pinch valve; 12 VDC, 1/16" ID x 1/8" OD tubing
    Pinch Valve for Tubing
    Two-Way, Normally Closed
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
    Specifications:
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLB98302-02
    型号
    98302-02
    内管直径
    0,0625"
    最大压力
    15 psi
    Pinch valve tube size
    1/16" ID x 1/8" OD
    功率
    12 VDC, 2,6 W
    响应时间
    25 ms
    质保
    2 years
    CE Compliant
  • MFLB98302-06
    03
    Two-way normally closed solenoid pinch valve; 12 VDC, 1/8" ID x 1/4" OD tubing
    Pinch Valve for Tubing
    Two-Way, Normally Closed
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
    Specifications:
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLB98302-06
    型号
    98302-06
    内管直径
    0,125"
    最大压力
    20 psi
    Pinch valve tube size
    1/8" ID x 1/4" OD
    功率
    12 VDC, 4 W
    响应时间
    35 ms
    质保
    2 years
    CE Compliant
  • MFLB98302-10
    04
    Two-way normally closed solenoid pinch valve; 24 VDC, 1/32" ID x 3/32" OD tubing
    Pinch Valve for Tubing
    Two-Way, Normally Closed
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
    Specifications:
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLB98302-10
    型号
    98302-10
    内管直径
    0,0312"
    最大压力
    15 psi
    Pinch valve tube size
    1/32" ID x 3/32" OD
    功率
    24 VDC, 2,6 W
    响应时间
    25 ms
    质保
    2 years
    CE Compliant
  • MFLB98302-12
    05
    Two-way normally closed solenoid pinch valve; 24 VDC, 1/16" ID x 1/8" OD tubing
    Pinch Valve for Tubing
    Two-Way, Normally Closed
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
    Specifications:
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLB98302-12
    型号
    98302-12
    内管直径
    0,0625"
    最大压力
    15 psi
    Pinch valve tube size
    1/16" ID x 1/8" OD
    功率
    24 VDC, 2,6 W
    响应时间
    25 ms
    质保
    2 years
    CE Compliant
  • MFLB98302-16
    06
    Two-way normally closed solenoid pinch valve; 24 VDC, 1/8" ID x 1/4" OD tubing
    Pinch Valve for Tubing
    Two-Way, Normally Closed
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
    Specifications:
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLB98302-16
    型号
    98302-16
    内管直径
    0,125"
    最大压力
    20 psi
    Pinch valve tube size
    1/8" ID x 1/4" OD
    功率
    24 VDC, 4 W
    响应时间
    35 ms
    质保
    2 years
    CE Compliant
  • MFLB98302-20
    07
    Two-way normally open solenoid pinch valve; 12 VDC, 1/32" ID x 3/32" OD tubing
    Pinch Valve for Tubing
    Two-Way, Normally Open
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
    Specifications:
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLB98302-20
    型号
    98302-20
    内管直径
    最大压力
    Pinch valve tube size
    1/32" ID x 3/32" OD
    功率
    12 VDC, 2,6 W
    响应时间
    25 ms
    质保
    2 years
    CE Compliant
  • MFLB98302-22
    08
    Two-way normally open solenoid pinch valve; 12 VDC, 1/16" ID x 1/8" OD tubing
    Pinch Valve for Tubing
    Two-Way, Normally Open
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
    Specifications:
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLB98302-22
    型号
    98302-22
    内管直径
    0,0625"
    最大压力
    15 psi
    Pinch valve tube size
    1/16" ID x 1/8" OD
    功率
    12 VDC, 2,6 W
    响应时间
    25 ms
    质保
    2 years
    CE Compliant
  • MFLB98302-26
    09
    Two-way normally open solenoid pinch valve; 12 VDC, 1/8" ID x 1/4" OD tubing
    Pinch Valve for Tubing
    Two-Way, Normally Closed
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
    Specifications:
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLB98302-26
    型号
    98302-26
    内管直径
    最大压力
    Pinch valve tube size
    1/8" ID x 1/4" OD
    功率
    12 VDC, 4 W
    响应时间
    35 ms
    质保
    2 years
    CE Compliant
  • MFLB98302-30
    10
    Two-way normally open solenoid pinch valve; 24 VDC, 1/32" ID x 3/32" OD tubing
    Pinch Valve for Tubing
    Two-Way, Normally Open
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
    Specifications:
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLB98302-30
    型号
    98302-30
    内管直径
    0,0312"
    最大压力
    15 psi
    Pinch valve tube size
    1/32" ID x 3/32" OD
    功率
    24 VDC, 2,6 W
    响应时间
    25 ms
    质保
    2 years
    CE Compliant
  • MFLB98302-32
    11
    Two-way normally open solenoid pinch valve; 24 VDC, 1/16" ID x 1/8" OD tubing
    Pinch Valve for Tubing
    Two-Way, Normally Open
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
    Specifications:
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLB98302-32
    型号
    98302-32
    内管直径
    最大压力
    15 psi
    Pinch valve tube size
    1/16" ID x 1/8" OD
    功率
    24 VDC, 2,6 W
    响应时间
    25 ms
    质保
    2 years
    CE Compliant
  • MFLB98302-36
    12
    Two-way normally open solenoid pinch valve; 24 VDC, 1/8" ID x 1/4" OD tubing
    Pinch Valve for Tubing
    Two-Way, Normally Closed
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
    Specifications:
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLB98302-36
    型号
    98302-36
    内管直径
    最大压力
    20 psi
    Pinch valve tube size
    1/8" ID x 1/4" OD
    功率
    24 VDC, 4 W
    响应时间
    35 ms
    质保
    2 years
    CE Compliant
  • MFLB98302-40
    13
    Three-way solenoid pinch valve; 12 VDC, 1/32" ID x 3/32" OD tubing
    Pinch Valve for Tubing
    Three-Way, One Normally Open, One Normally Closed
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
    Specifications:
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLB98302-40
    型号
    98302-40
    内管直径
    最大压力
    15 psi
    Pinch valve tube size
    1/32" ID x 3/32" OD
    功率
    12 VDC, 2,6 W
    响应时间
    25 ms
    质保
    2 years
    CE Compliant
  • MFLB98302-42
    14
    Three-way solenoid pinch valve; 12 VDC, 1/16" ID x 1/8" OD tubing
    Pinch Valve for Tubing
    Three-Way, One Normally Open, One Normally Closed
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
    Specifications:
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLB98302-42
    型号
    98302-42
    内管直径
    0,0625"
    最大压力
    15 psi
    Pinch valve tube size
    1/16" ID x 1/8" OD
    功率
    12 VDC, 2,6 W
    响应时间
    25 ms
    质保
    2 years
    CE Compliant
  • MFLB98302-46
    15
    Three-way solenoid pinch valve; 12 VDC, 1/8" ID x 1/4" OD tubing
    Pinch Valve for Tubing
    Three-Way, One Normally Open, One Normally Closed
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
    Specifications:
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLB98302-46
    型号
    98302-46
    内管直径
    0,125"
    最大压力
    20 psi
    Pinch valve tube size
    1/8" ID x 1/4" OD
    功率
    12 VDC, 4 W
    响应时间
    35 ms
    质保
    2 years
    CE Compliant
  • MFLB98302-48
    16
    Three-way solenoid pinch valve; 24 VDC, 1/32" ID x 3/32" OD tubing
    Pinch Valve for Tubing
    Three-Way, One Normally Open, One Normally Closed
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
    Specifications:
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLB98302-48
    型号
    98302-48
    内管直径
    最大压力
    Pinch valve tube size
    1/32" ID x 3/32" OD
    功率
    24 VDC, 2,6 W
    响应时间
    25 ms
    质保
    2 years
    CE Compliant
  • MFLB98302-50
    17
    Three-way solenoid pinch valve; 24 VDC, 1/16" ID x 1/8" OD tubing
    Pinch Valve for Tubing
    Three-Way, One Normally Open, One Normally Closed
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
    Specifications:
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLB98302-50
    型号
    98302-50
    内管直径
    0,0625"
    最大压力
    Pinch valve tube size
    1/16" ID x 1/8" OD
    功率
    24 VDC, 2,6 W
    响应时间
    25 ms
    质保
    2 years
    CE Compliant
  • MFLB98302-54
    18
    Three-way solenoid pinch valve; 24 VDC, 1/8" ID x 1/4" OD tubing
    Pinch Valve for Tubing
    Three-Way, One Normally Open, One Normally Closed
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
    Specifications:
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLB98302-54
    型号
    98302-54
    内管直径
    0,125"
    最大压力
    20 psi
    Pinch valve tube size
    1/8" ID x 1/4" OD
    功率
    24 VDC, 4 W
    响应时间
    35 ms
    质保