Masterflex® L/S® High-Performance Pump Head Tubing Retainer Kit
Catalog # MFLA77250-66
Supplier: MASTERFLEX PROPRIETARY
Specifications
- Pump series:L/S
- 说明:Tubing retainer kit for L/S high-performance pump head
- 质保:2 years
- 包装规格:1
Specifications
About this item
Extend the life of your Masterflex pump head with this service kit.
- Tubing retainer prevents tubing from moving within the pump head
Compatible with the Masterflex® L/S® High-Performance Pump Heads 77250-62.
- Pump series:L/S
- 说明:Tubing retainer kit for L/S high-performance pump head
- Environmentally Preferable:
- 质保:2 years
- 包装规格:1