Masterflex® I/P® High-Performance Pump Head Replacement Rotor and Body Assembly
Catalog # MFLA77600-69
Supplier: MASTERFLEX PROPRIETARY
Specifications
- Pump series:I/P
- 说明:KIT REPL ROTOR & BODY ASSEMBLY (Rotor/body assembly with tang boot)
- 包装规格:1
- Housing material:Stainless steel, thermoset polyester
- Rotor material:Stainless Steel
Specifications
About this item
Get the most our of your Masterflex® I/P® High-Performance Pump Head with these replacement parts.
- Stainless steel (SS) rotor / roller for greater durability and chemical resistance
Replacement parts to keep your pump head operating even longer.
Delivery: Includes rotor, body, and tang boot.
- Pump series:I/P
- 说明:KIT REPL ROTOR & BODY ASSEMBLY (Rotor/body assembly with tang boot)
- Environmentally Preferable:
- 包装规格:1
- Housing material:Stainless steel, thermoset polyester
- Rotor material:Stainless Steel