PTFE 分流管
供应商: MASTERFLEX PREMIUM
关于此项目
Chemically-inert PTFE handles some of your most aggressive chemical solutions.
- Constructed of durable PTFE
Come in a variety of configurations—choose the number of ports and connections to suit your needs!
11 Options Available Below
Product Details & Documents
Chemically-inert PTFE handles some of your most aggressive chemical solutions.
- Constructed of durable PTFE
Come in a variety of configurations—choose the number of ports and connections to suit your needs!
Specifications for Product Family
规格
- Catalog No:
- MFLB31521-01
- MFLB31521-07
- MFLB31521-09
- MFLB31521-11
- MFLB31521-13
- MFLB31521-15
- MFLB31521-17
- MFLB31521-19
- MFLB31521-21
- MFLB31521-25
- MFLB31521-27
- Inlet NPT(F):
- 0,25"
- 0,25"
- 0,375"
- 0,5"
- 0,25"
- 0,375"
- 0,5"
- 0,25"
- 0,375"
- 0,25"
- 0,375"
- 最大压力:
- 120 psi
- 120 psi
- 120 psi
- 120 psi
- 120 psi
- 120 psi
- 120 psi
- 120 psi
- 120 psi
- 120 psi
- 120 psi
- 最高温度:
- 150 °C (300 °F)
- 150 °C (300 °F)
- 150 °C (300 °F)
- 150 °C (300 °F)
- 150 °C (300 °F)
- 150 °C (300 °F)
- 150 °C (300 °F)
- 150 °C (300 °F)
- 150 °C (300 °F)
- 150 °C (300 °F)
- 150 °C (300 °F)
- 最小温度:
- –129 °C (–200 °F)
- –129 °C (–200 °F)
- –129 °C (–200 °F)
- –129 °C (–200 °F)
- –129 °C (–200 °F)
- –129 °C (–200 °F)
- –129 °C (–200 °F)
- –129 °C (–200 °F)
- –129 °C (–200 °F)
- –129 °C (–200 °F)
- –129 °C (–200 °F)
- Mounting hole:
- 0,17"
- 0,17"
- 0,2"
- 0,2"
- 0,17"
- 0,2"
- 0,2"
- 0,17"
- 0,2"
- 0,17"
- 0,2"
- 出口数量:
- 2
- 4
- 4
- 4
- 6
- 6
- 6
- 8
- 8
- 10
- 10
- Outlet NPT(F):
- 0,125"
- 0,125"
- 0,25"
- 0,375"
- 0,125"
- 0,25"
- 0,375"
- 0,125"
- 0,25"
- 0,125"
- 0,25"
- 质保:
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
规格
Product Family Options
产品信息
- 说明宽度长度高度材质可用库存Price
- 规格:
More
Specifications
Cat. No.MFLB31521-01Inlet NPT(F)0,25"最大压力120 psi最高温度150 °C (300 °F)最小温度–129 °C (–200 °F)Mounting hole0,17"出口数量2Outlet NPT(F)0,125"质保Out of box - 规格:
More
Specifications
Cat. No.MFLB31521-07Inlet NPT(F)0,25"最大压力120 psi最高温度150 °C (300 °F)最小温度–129 °C (–200 °F)Mounting hole0,17"出口数量4Outlet NPT(F)0,125"质保Out of box - 规格:
More
Specifications
Cat. No.MFLB31521-09Inlet NPT(F)0,375"最大压力120 psi最高温度150 °C (300 °F)最小温度–129 °C (–200 °F)Mounting hole0,2"出口数量4Outlet NPT(F)0,25"质保Out of box - 规格:
More
Specifications
Cat. No.MFLB31521-11Inlet NPT(F)0,5"最大压力120 psi最高温度150 °C (300 °F)最小温度–129 °C (–200 °F)Mounting hole0,2"出口数量4Outlet NPT(F)0,375"质保Out of box - 规格:
More
Specifications
Cat. No.MFLB31521-13Inlet NPT(F)0,25"最大压力120 psi最高温度150 °C (300 °F)最小温度–129 °C (–200 °F)Mounting hole0,17"出口数量6Outlet NPT(F)0,125"质保Out of box - 规格:
More
Specifications
Cat. No.MFLB31521-15Inlet NPT(F)0,375"最大压力120 psi最高温度150 °C (300 °F)最小温度–129 °C (–200 °F)Mounting hole0,2"出口数量6Outlet NPT(F)0,25"质保Out of box - 规格:
More
Specifications
Cat. No.MFLB31521-17Inlet NPT(F)0,5"最大压力120 psi最高温度150 °C (300 °F)最小温度–129 °C (–200 °F)Mounting hole0,2"出口数量6Outlet NPT(F)0,375"质保Out of box - 规格:
More
Specifications
Cat. No.MFLB31521-19Inlet NPT(F)0,25"最大压力120 psi最高温度150 °C (300 °F)最小温度–129 °C (–200 °F)Mounting hole0,17"出口数量8Outlet NPT(F)0,125"质保Out of box - 规格:
More
Specifications
Cat. No.MFLB31521-21Inlet NPT(F)0,375"最大压力120 psi最高温度150 °C (300 °F)最小温度–129 °C (–200 °F)Mounting hole0,2"出口数量8Outlet NPT(F)0,25"质保Out of box - 规格:
More
Specifications
Cat. No.MFLB31521-25Inlet NPT(F)0,25"最大压力120 psi最高温度150 °C (300 °F)最小温度–129 °C (–200 °F)Mounting hole0,17"出口数量10Outlet NPT(F)0,125"质保Out of box - 规格:
More
Specifications
Cat. No.MFLB31521-27Inlet NPT(F)0,375"最大压力120 psi最高温度150 °C (300 °F)最小温度–129 °C (–200 °F)Mounting hole0,2"出口数量10Outlet NPT(F)0,25"质保Out of box