Masterflex® Transfer Tubing, Terminator®, Avantor®
Superior abrasion resistance at high pressure.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Global market leader in range of formulations for most extensive chemical compatibility
- Outstanding resistance to heat and ozone
Masterflex® Terminator® transfer tubing is ideal for use with severe service applications due to its superior abrasion resistance. The Buna-N nitrile tube, modified Buna-N nitrile tube cover, along with the high tensile textile core tube reinforcement makes this tubing suitable for high pressure applications. The tube and tube cover meet RMA Class A high oil resistance and nonconductive at 1000 VDC certification.
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.
- Environmentally Preferable:
- 型号:
- 95954-11
- 95954-07
- 95954-05
- 95954-03
- 95954-01
- Bend radius:
- 355,6 mm (14")
- 203,2 mm (8")
- 152,4 mm (6")
- 127 mm (5")
- 76,2 mm (3")
- 透明度:
- Opaque
- Opaque
- Opaque
- Opaque
- Opaque
- Length per pack:
- 7,6 m (25 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 30,5 m (100 ft.)
- 30,5 m (100 ft.)
- 最大压力:
- 34,6 bar (501 psi)
- 34,6 bar (501 psi)
- 34,6 bar (501 psi)
- 34,6 bar (501 psi)
- 34,6 bar (501 psi)
- 最高温度:
- 100 °C
- 100 °C
- 100 °C
- 100 °C
- 100 °C
- 最小温度:
- –40 °C (–40 °F)
- –40 °C (–40 °F)
- –40 °C (–40 °F)
- –40 °C (–40 °F)
- –40 °C (–40 °F)
- Sterilisation method autoclave:
- No
- No
- No
- No
- No
- Sterilisation method EtO:
- No
- No
- No
- No
- No
- Sterilisation method gamma:
- No
- No
- No
- No
- No
- 工作压力:
- 501 psi
- 501 psi
- 501 psi
- 501 psi
- 501 psi
- Weight per ft.:
- 0,54 kg (1,2 lbs.)
- 0,26 kg (0,58 lbs.)
- 0,18 kg (0,4 lbs.)
- 0,11 kg (0,25 lbs.)
- 0,05 kg (0,12 lbs.)