Masterflex® Transfer Tubing, Premo Flex®, Avantor®
Resists weather and ozone.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Global market leader in range of formulations for most extensive chemical compatibility
- Outstanding resistance to heat and ozone
Masterflex® Premp Flex® transfer tubing is ideal for use with fuels such as petroleum oils, gasoline, kerosene, and lubricating oils, while maintaining tube integrity. The Buna-N tube, modified Buna-N nitrile tube cover, along with the high tensile textile core tube reinforcement makes this tubing suitable for high pressure applications. The tube meets RMA Class A high oil resistance and nonconductive at 1000 VDC certification. The tube cover meets Class B medium oil resistance and nonconductive at 1000 VDC certification.
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.
- Environmentally Preferable:
- 型号:
- 95953-07
- 95953-05
- 95953-03
- 95953-01
- Bend radius:
- 203,2 mm (8")
- 127 mm (5")
- 101,6 mm (4")
- 50,8 mm (2")
- 透明度:
- Opaque
- Opaque
- Opaque
- Opaque
- Length per pack:
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 30,5 m (100 ft.)
- 30,5 m (100 ft.)
- 最大压力:
- 17,2 bar (250 psi)
- 17,2 bar (250 psi)
- 17,2 bar (250 psi)
- 17,2 bar (250 psi)
- 最高温度:
- 100 °C
- 100 °C
- 100 °C
- 100 °C
- 最小温度:
- –28 °C (–20 °F)
- –28 °C (–20 °F)
- –28 °C (–20 °F)
- –28 °C (–20 °F)
- Sterilisation method autoclave:
- No
- No
- No
- No
- Sterilisation method EtO:
- No
- No
- No
- No
- Sterilisation method gamma:
- No
- No
- No
- No
- 工作压力:
- 17,2 bar (250 psi)
- 17,2 bar (250 psi)
- 17,2 bar (250 psi)
- 17,2 bar (250 psi)
- Weight per ft.:
- 0,24 kg (0,54 lbs.)
- 0,16 kg (0,36 lbs.)
- 0,1 kg (0,22 lbs.)
- 0,04 kg (0,09 lbs.)