Orders LinkContactUsLinkComponent
Masterflex® 两通挤压阀;常闭,Avantor®
Masterflex® 两通挤压阀;常闭,Avantor®
Masterflex® 两通挤压阀;常闭,Avantor®
Masterflex® 两通挤压阀;常闭,Avantor®
Masterflex® 两通挤压阀;常闭,Avantor®
Excellent choice for viscous fluids.
  • Specifically designed for metric sized tubing
These solenoid-operated 2- and 3-Way pinch valves are ideal for use with fluids that are difficult for traditional solenoid valves to handle, such as viscous fluids. They are ideal for on/off flow control applications and where frequent changes of wetted parts are necessary. All models are compatible with silicone, C-FLEX®, and PharMed® BPT tubing. The two-way valves are available in both normally closed and normally open configurations. The three-way valves accept two tubes, the bottom tube is normally open while the top is normally closed.
  • 型号:
  • 98305-00
  • 98305-01
  • 98305-02
  • 98305-03
  • 98305-05
  • 98305-06
  • 98305-07
  • 98305-10
  • 98305-11
  • 98305-12
  • 98305-13
  • 98305-14
  • 98305-15
  • Pinch valve tube size:
  • 2,4 mm OD
  • 2,4 mm OD
  • 10 mm OD
  • 10 mm OD
  • 13 mm OD
  • 19 mm OD
  • 19 mm OD
  • 10 mm OD
  • 10 mm OD
  • 13 mm OD
  • 13 mm OD
  • 19 mm OD
  • 19 mm OD
  • 功率:
  • 12 VDC, 4,4 W
  • 24 VDC, 4,4 W
  • 12 VDC, 13 W
  • 24 VDC, 13 W
  • 24 VDC, 60 W
  • 12 VDC, 60 W
  • 24 VDC, 60 W
  • 12 VDC, 13 W
  • 24 VDC, 13 W
  • 12 VDC, 60 W
  • 24 VDC, 60 W
  • 12 VDC, 60 W
  • 24 VDC, 60 W
  • 质保:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
Product Family Options
  • 顺序
    说明
    Connection Type
    Flow pattern
  • 01
    Solenoid-Operated Three-Way Pinch Valve, 12 VDC
    Pinch Valve for Tubing
    Three-Way, One Normally Closed, One Normally Open
    1 * 1 件
    1 * 1 件
    Specifications
    Cat. No.
    MFLB98305-00
    型号
    98305-00
    Pinch valve tube size
    2,4 mm OD
    功率
    12 VDC, 4,4 W
    质保
    1 year
  • 02
    Solenoid-Operated Three-Way Pinch Valve, 24 VDC
    Pinch Valve for Tubing
    Three-Way, One Normally Closed, One Normally Open
    1 * 1 件
    1 * 1 件
    Specifications
    Cat. No.
    MFLB98305-01
    型号
    98305-01
    Pinch valve tube size
    2,4 mm OD
    功率
    24 VDC, 4,4 W
    质保
    1 year
  • 03
    Solenoid-Operated Two-Way Pinch Valve; Normally-Open, 10 mm Tube OD, 12 VDC
    Pinch Valve for Tubing
    Two-Way, Normally Open
    1 * 1 件
    1 * 1 件
    Specifications
    Cat. No.
    MFLB98305-02
    型号
    98305-02
    Pinch valve tube size
    10 mm OD
    功率
    12 VDC, 13 W
    质保
    1 year
  • 04
    Solenoid-Operated Two-Way Pinch Valve; Normally-Open, 10 mm Tube OD, 24 VDC
    Pinch Valve for Tubing
    Two-Way, Normally Open
    1 * 1 件
    1 * 1 件
    Specifications
    Cat. No.
    MFLB98305-03
    型号
    98305-03
    Pinch valve tube size
    10 mm OD
    功率
    24 VDC, 13 W
    质保
    1 year
  • 05
    Solenoid-Operated Two-Way Pinch Valve; Normally-Open, 13 mm Tube OD, 24 VDC
    Pinch Valve for Tubing
    Two-Way, Normally Open
    1 * 1 件
    1 * 1 件
    Specifications
    Cat. No.
    MFLB98305-05
    型号
    98305-05
    Pinch valve tube size
    13 mm OD
    功率
    24 VDC, 60 W
    质保
    1 year
  • 06
    Solenoid-Operated Two-Way Pinch Valve; Normally-Open, 19 mm Tube OD, 12 VDC
    Pinch Valve for Tubing
    Two-Way, Normally Open
    1 * 1 件
    1 * 1 件
    Specifications
    Cat. No.
    MFLB98305-06
    型号
    98305-06
    Pinch valve tube size
    19 mm OD
    功率
    12 VDC, 60 W
    质保
    1 year
  • 07
    Solenoid-Operated Two-Way Pinch Valve; Normally-Open, 19 mm Tube OD, 24 VDC
    Pinch Valve for Tubing
    Two-Way, Normally Open
    1 * 1 件
    1 * 1 件
    Specifications
    Cat. No.
    MFLB98305-07
    型号
    98305-07
    Pinch valve tube size
    19 mm OD
    功率
    24 VDC, 60 W
    质保
    1 year
  • 08
    Solenoid-Operated Two-Way Pinch Valve; Normally-Closed, 10 mm Tube OD, 12 VDC
    Pinch Valve for Tubing
    Two-Way, Normally Closed
    1 * 1 件
    1 * 1 件
    Specifications
    Cat. No.
    MFLB98305-10
    型号
    98305-10
    Pinch valve tube size
    10 mm OD
    功率
    12 VDC, 13 W
    质保
    1 year
  • 09
    Solenoid-Operated Two-Way Pinch Valve; Normally-Closed, 10 mm Tube OD, 24 VDC
    Pinch Valve for Tubing
    Two-Way, Normally Closed
    1 * 1 件
    1 * 1 件
    Specifications
    Cat. No.
    MFLB98305-11
    型号
    98305-11
    Pinch valve tube size
    10 mm OD
    功率
    24 VDC, 13 W
    质保
    1 year
  • 10
    Solenoid-Operated Two-Way Pinch Valve; Normally-Closed, 13 mm Tube OD, 12 VDC
    Pinch Valve for Tubing
    Two-Way, Normally Closed
    1 * 1 件
    1 * 1 件
    Specifications
    Cat. No.
    MFLB98305-12
    型号
    98305-12
    Pinch valve tube size
    13 mm OD
    功率
    12 VDC, 60 W
    质保
    1 year
  • 11
    Solenoid-Operated Two-Way Pinch Valve; Normally-Closed, 13 mm Tube OD, 24 VDC
    Pinch Valve for Tubing
    Two-Way, Normally Closed
    1 * 1 件
    1 * 1 件
    Specifications
    Cat. No.
    MFLB98305-13
    型号
    98305-13
    Pinch valve tube size
    13 mm OD
    功率
    24 VDC, 60 W
    质保
    1 year
  • 12
    Solenoid-Operated Two-Way Pinch Valve; Normally-Closed, 19 mm Tube OD, 12 VDC
    Pinch Valve for Tubing
    Two-Way, Normally Closed
    1 * 1 件
    1 * 1 件
    Specifications
    Cat. No.
    MFLB98305-14
    型号
    98305-14
    Pinch valve tube size
    19 mm OD
    功率
    12 VDC, 60 W
    质保
    1 year
  • 13
    Solenoid-Operated Two-Way Pinch Valve; Normally-Closed, 19 mm Tube OD, 24 VDC
    Pinch Valve for Tubing
    Two-Way, Normally Closed
    1 * 1 件
    1 * 1 件
    Specifications
    Cat. No.
    MFLB98305-15
    型号
    98305-15
    Pinch valve tube size
    19 mm OD
    功率
    24 VDC, 60 W
    质保
    1 year