Masterflex® 聚碳酸酯Luer旋塞阀,Avantor®
Supplier: MASTERFLEX PROPRIETARY
Certificates
About this item
Regulate flow without any leaking.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
旋塞阀有带luer阳锁和带luer阳插销两种可选。单向旋塞阀旋杆可旋转90度,三通旋塞阀旋杆可旋转180度,四通旋塞阀旋杆可旋转360度。所有阀体均为PC材质,旋杆为HDPE材质。
- Environmentally Preferable:
Specifications
- 浸湿材料:
- Polycarbonate
- Polycarbonate
- Polycarbonate
- Polycarbonate
- 质保:
- Out of box
- Out of box
- Out of box
- Out of box