流量控制夹- 棘轮泵管夹
Supplier: MASTERFLEX PROPRIETARY
About this item
Won't rust, corrode, or distort.
- Constructed of durable Celcon plastic
耐用的CELCON®塑料夹具可高温高压灭菌;不生锈,耐腐蚀,不会变形。12-位棘轮控制可从完全流通调节至完全闭合。接受挠性管。
2 Options Available Below
Product Details & Documents
Won't rust, corrode, or distort.
- Constructed of durable Celcon plastic
耐用的CELCON®塑料夹具可高温高压灭菌;不生锈,耐腐蚀,不会变形。12-位棘轮控制可从完全流通调节至完全闭合。接受挠性管。
Specifications for Product Family
Specifications
- Catalog No:
- MFLA06833-00
- MFLA06833-01
- 外壳材质:
- Polyoxymethylene (POM) Acetal
- Polyoxymethylene (POM) Acetal
- Pinch valve tube size:
- 1/8" to 1/2" OD
- 1/16" to 1/4" OD
- 质保:
- 3 Months
Specifications
Product Family Options
Product Information
- 说明AvailabilityPrice
- Specifications:
More
Specifications
Cat. No.MFLA06833-00外壳材质Polyoxymethylene (POM) AcetalPinch valve tube size1/8" to 1/2" OD质保3 Months - Specifications:
More
Specifications
Cat. No.MFLA06833-01外壳材质Polyoxymethylene (POM) AcetalPinch valve tube size1/16" to 1/4" OD质保