Masterflex® 聚酰亚胺管,Avantor®
Supplier: MASTERFLEX PREMIUM
About this item
Provides excellent temperature stability.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimise downtime
- Regulatory approvals to match your unique application
应用范围:用作电和热绝缘体,用于输送气体和液体,或用作坚韧耐用的玻璃纤维护套。
特点:优异的耐化学性、耐热性、耐辐射性和耐磨性;绝缘强度高。透明琥珀色。
认证:树脂符合 USP VI 类要求和 NEMA 要求。
温度范围:–418 至 896°F(–250 至 480°C)
灭菌:高压灭菌、环氧乙烷或 γ 射线照射灭菌。
14 Options Available Below
Product Details & Documents
Provides excellent temperature stability.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimise downtime
- Regulatory approvals to match your unique application
应用范围:用作电和热绝缘体,用于输送气体和液体,或用作坚韧耐用的玻璃纤维护套。
特点:优异的耐化学性、耐热性、耐辐射性和耐磨性;绝缘强度高。透明琥珀色。
认证:树脂符合 USP VI 类要求和 NEMA 要求。
温度范围:–418 至 896°F(–250 至 480°C)
灭菌:高压灭菌、环氧乙烷或 γ 射线照射灭菌。
Documents
Certificates
Search by Lot NumberSpecifications for Product Family
Specifications
- Catalog No:
- MFLB95820-00
- MFLB95820-01
- MFLB95820-02
- MFLB95820-03
- MFLB95820-04
- MFLB95820-05
- MFLB95820-06
- MFLB95820-07
- MFLB95820-08
- MFLB95820-09
- MFLB95820-10
- MFLB95820-11
- MFLB95820-12
- MFLB95820-13
- 型号:
- 95820-00
- 95820-01
- 95820-02
- 95820-03
- 95820-04
- 95820-05
- 95820-06
- 95820-07
- 95820-08
- 95820-09
- 95820-10
- 95820-11
- 95820-12
- 95820-13
- 透明度:
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Length per pack:
- 1,5 m (5 ft.)
- 1,5 m (5 ft.)
- 1,5 m (5 ft.)
- 1,5 m (5 ft.)
- 1,5 m (5 ft.)
- 1,5 m (5 ft.)
- 1,5 m (5 ft.)
- 1,5 m (5 ft.)
- 1,5 m (5 ft.)
- 1,5 m (5 ft.)
- 1,5 m (5 ft.)
- 1,5 m (5 ft.)
- 1,5 m (5 ft.)
- 1,5 m (5 ft.)
- 最大压力:
- 48,3 bar (700 psi)
- 48,3 bar (700 psi)
- 48,3 bar (700 psi)
- 48,3 bar (700 psi)
- 34,5 bar (500 psi)
- 27,6 bar (400 psi)
- 20,7 bar (300 psi)
- 37,9 bar (550 psi)
- 37,9 bar (550 psi)
- 34,5 bar (500 psi)
- 31 bar (450 psi)
- 27,6 bar (400 psi)
- 24,1 bar (350 psi)
- 19 bar (275 psi)
- 最高温度:
- 480 °C (896 °F)
- 480 °C (896 °F)
- 480 °C (896 °F)
- 480 °C (896 °F)
- 480 °C (896 °F)
- 480 °C (896 °F)
- 480 °C (896 °F)
- 480 °C (896 °F)
- 480 °C (896 °F)
- 480 °C (896 °F)
- 480 °C (896 °F)
- 480 °C (896 °F)
- 480 °C (896 °F)
- 480 °C (896 °F)
- 最小温度:
- –250 °C (–418 °F)
- –250 °C (–418 °F)
- –250 °C (–418 °F)
- –250 °C (–418 °F)
- –250 °C (–418 °F)
- –250 °C (–418 °F)
- –250 °C (–418 °F)
- –250 °C (–418 °F)
- –250 °C (–418 °F)
- –250 °C (–418 °F)
- –250 °C (–418 °F)
- –250 °C (–418 °F)
- –250 °C (–418 °F)
- –250 °C (–418 °F)
- Regulatory approvals:
- USP
- USP
- USP
- USP
- USP
- USP
- USP
- USP
- USP
- USP
- USP
- USP
- USP
- USP
- Sterilisation method autoclave:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Sterilisation method EtO:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Sterilisation method gamma:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- 工作压力:
- 48,3 bar (700 psi)
- 48,3 bar (700 psi)
- 48,3 bar (700 psi)
- 48,3 bar (700 psi)
- 34,5 bar (500 psi)
- 27,6 bar (400 psi)
- 20,7 bar (300 psi)
- 37,9 bar (550 psi)
- 37,9 bar (550 psi)
- 34,5 bar (500 psi)
- 31 bar (450 psi)
- 27,6 bar (400 psi)
- 24,1 bar (350 psi)
- 19 bar (275 psi)
- 质保:
- 3 Months
- 3 Months
- 3 Months
- 3 Months
- 3 Months
- 3 Months
- 3 Months
- 3 Months
- 3 Months
- 3 Months
- 3 Months
- 3 Months
- 3 Months
- 3 Months
Specifications
Product Family Options
Product Information
- 顺序说明Tubing int.Ø壁厚Tubing ext.ØTubing material长度颜色AvailabilityPrice
- 01Transfer Tubing, Polyimide, 0,0049" ID x 0,0064" OD, 12"L; 5/PK0,124 mm (0,0049")0,019 mm0,163 mm (0,0064")Polymide0,3 m琥珀色Log in for availability1 * 5 件Specifications:CE certificate
More
Specifications
Cat. No.MFLB95820-00型号95820-00透明度TranslucentLength per pack1,5 m (5 ft.)最大压力48,3 bar (700 psi)最高温度480 °C (896 °F)最小温度–250 °C (–418 °F)Regulatory approvalsUSPSterilisation method autoclaveYesSterilisation method EtOYesSterilisation method gammaYes工作压力48,3 bar (700 psi)质保3 MonthsCE certificate - 02Transfer Tubing, Polyimide, 0,0078" ID x 0,0093" OD, 12"L; 5/PK0,198 mm (0,0078")0,019 mm0,236 mm (0,0093")Polymide0,3 m琥珀色Log in for availability1 * 5 件Specifications:CE certificate
More
Specifications
Cat. No.MFLB95820-01型号95820-01透明度TranslucentLength per pack1,5 m (5 ft.)最大压力48,3 bar (700 psi)最高温度480 °C (896 °F)最小温度–250 °C (–418 °F)Regulatory approvalsUSPSterilisation method autoclaveYesSterilisation method EtOYesSterilisation method gammaYes工作压力48,3 bar (700 psi)质保3 MonthsCE certificate - 03Transfer Tubing, Polyimide, 0,0120" ID x 0,0140" OD, 12"L; 5/PK0,305 mm (0,012")0,025 mm0,356 mm (0,014")Polymide0,3 m琥珀色Log in for availability1 * 5 件Specifications:CE certificate
More
Specifications
Cat. No.MFLB95820-02型号95820-02透明度TranslucentLength per pack1,5 m (5 ft.)最大压力48,3 bar (700 psi)最高温度480 °C (896 °F)最小温度–250 °C (–418 °F)Regulatory approvalsUSPSterilisation method autoclaveYesSterilisation method EtOYesSterilisation method gammaYes工作压力48,3 bar (700 psi)质保3 MonthsCE certificate - 04Transfer Tubing, Polyimide, 0,0135" ID x 0,0155" OD, 12"L; 5/PK0,343 mm (0,0135")0,025 mm0,394 mm (0,0155")Polymide0,3 m琥珀色Log in for availability1 * 5 件Specifications:CE certificate
More
Specifications
Cat. No.MFLB95820-03型号95820-03透明度TranslucentLength per pack1,5 m (5 ft.)最大压力48,3 bar (700 psi)最高温度480 °C (896 °F)最小温度–250 °C (–418 °F)Regulatory approvalsUSPSterilisation method autoclaveYesSterilisation method EtOYesSterilisation method gammaYes工作压力48,3 bar (700 psi)质保3 MonthsCE certificate - 05Transfer Tubing, Polyimide, 0,0195" ID x 0,0215" OD, 12"L; 5/PK0,495 mm (0,0195")0,025 mm0,546 mm (0,0215")Polymide0,3 m琥珀色Log in for availability1 * 5 件Specifications:CE certificate
More
Specifications
Cat. No.MFLB95820-04型号95820-04透明度TranslucentLength per pack1,5 m (5 ft.)最大压力34,5 bar (500 psi)最高温度480 °C (896 °F)最小温度–250 °C (–418 °F)Regulatory approvalsUSPSterilisation method autoclaveYesSterilisation method EtOYesSterilisation method gammaYes工作压力34,5 bar (500 psi)质保3 MonthsCE certificate - 06Transfer Tubing, Polyimide, 0,0250" ID x 0,0270" OD, 12"L; 5/PK0,635 mm (0,025")0,025 mm0,686 mm (0,027")Polymide0,3 m琥珀色Log in for availability1 * 5 件Specifications:CE certificate
More
Specifications
Cat. No.MFLB95820-05型号95820-05透明度TranslucentLength per pack1,5 m (5 ft.)最大压力27,6 bar (400 psi)最高温度480 °C (896 °F)最小温度–250 °C (–418 °F)Regulatory approvalsUSPSterilisation method autoclaveYesSterilisation method EtOYesSterilisation method gammaYes工作压力27,6 bar (400 psi)质保3 MonthsCE certificate - 07Transfer Tubing, Polyimide, 0,0320" ID x 0,0340" OD, 12"L; 5/PK0,813 mm (0,032")0,025 mm0,864 mm (0,034")Polymide0,3 m琥珀色Log in for availability1 * 5 件Specifications:CE certificate
More
Specifications
Cat. No.MFLB95820-06型号95820-06透明度TranslucentLength per pack1,5 m (5 ft.)最大压力20,7 bar (300 psi)最高温度480 °C (896 °F)最小温度–250 °C (–418 °F)Regulatory approvalsUSPSterilisation method autoclaveYesSterilisation method EtOYesSterilisation method gammaYes工作压力20,7 bar (300 psi)质保3 MonthsCE certificate - 08Transfer Tubing, Polyimide, 0,0340" ID x 0,0380" OD, 12"L; 5/PK0,864 mm (0,034")0,051 mm0,965 mm (0,038")Polymide0,3 m琥珀色Log in for availability1 * 5 件Specifications:CE certificate
More
Specifications
Cat. No.MFLB95820-07型号95820-07透明度TranslucentLength per pack1,5 m (5 ft.)最大压力37,9 bar (550 psi)最高温度480 °C (896 °F)最小温度–250 °C (–418 °F)Regulatory approvalsUSPSterilisation method autoclaveYesSterilisation method EtOYesSterilisation method gammaYes工作压力37,9 bar (550 psi)质保3 MonthsCE certificate - 09Transfer Tubing, Polyimide, 0,0360" ID x 0,0400" OD, 12"L; 5/PK0,914 mm (0,036")0,051 mm1,016 mm (0,04")Polymide0,3 m琥珀色Log in for availability1 * 5 件Specifications:CE certificate
More
Specifications
Cat. No.MFLB95820-08型号95820-08透明度TranslucentLength per pack1,5 m (5 ft.)最大压力37,9 bar (550 psi)最高温度480 °C (896 °F)最小温度–250 °C (–418 °F)Regulatory approvalsUSPSterilisation method autoclaveYesSterilisation method EtOYesSterilisation method gammaYes工作压力37,9 bar (550 psi)质保3 MonthsCE certificate - 10Transfer Tubing, Polyimide, 0,0395" ID x 0,0435" OD, 12"L; 5/PK1,03 mm (0,0395")0,051 mm1,105 mm (0,0435")Polymide0,3 m琥珀色Log in for availability1 * 5 件Specifications:CE certificate
More
Specifications
Cat. No.MFLB95820-09型号95820-09透明度TranslucentLength per pack1,5 m (5 ft.)最大压力34,5 bar (500 psi)最高温度480 °C (896 °F)最小温度–250 °C (–418 °F)Regulatory approvalsUSPSterilisation method autoclaveYesSterilisation method EtOYesSterilisation method gammaYes工作压力34,5 bar (500 psi)质保3 MonthsCE certificate - 11Transfer Tubing, Polyimide, 0,0445" ID x 0,0485" OD, 12"L; 5/PK1,13 mm (0,0445")0,051 mm1,232 mm (0,0485")Polymide0,3 m琥珀色Log in for availability1 * 5 件Specifications:CE certificate
More
Specifications
Cat. No.MFLB95820-10型号95820-10透明度TranslucentLength per pack1,5 m (5 ft.)最大压力31 bar (450 psi)最高温度480 °C (896 °F)最小温度–250 °C (–418 °F)Regulatory approvalsUSPSterilisation method autoclaveYesSterilisation method EtOYesSterilisation method gammaYes工作压力31 bar (450 psi)质保3 MonthsCE certificate - 12Transfer Tubing, Polyimide, 0,0500" ID x 0,0540" OD, 12"L; 5/PK1,27 mm (0,05")0,051 mm1,372 mm (0,054")Polymide0,3 m琥珀色Log in for availability1 * 5 件Specifications:CE certificate
More
Specifications
Cat. No.MFLB95820-11型号95820-11透明度TranslucentLength per pack1,5 m (5 ft.)最大压力27,6 bar (400 psi)最高温度480 °C (896 °F)最小温度–250 °C (–418 °F)Regulatory approvalsUSPSterilisation method autoclaveYesSterilisation method EtOYesSterilisation method gammaYes工作压力27,6 bar (400 psi)质保3 MonthsCE certificate - 13Transfer Tubing, Polyimide, 0,0575" ID x 0,0615" OD, 12"L; 5/PK1,461 mm (0,0575")0,051 mm1,562 mm (0,0615")Polymide0,3 m琥珀色Log in for availability1 * 5 件Specifications:CE certificate
More
Specifications
Cat. No.MFLB95820-12型号95820-12透明度TranslucentLength per pack1,5 m (5 ft.)最大压力24,1 bar (350 psi)最高温度480 °C (896 °F)最小温度–250 °C (–418 °F)Regulatory approvalsUSPSterilisation method autoclaveYesSterilisation method EtOYesSterilisation method gammaYes工作压力24,1 bar (350 psi)质保3 MonthsCE certificate - 14Transfer Tubing, Polyimide, 0,0710" ID x 0,0750" OD, 12"L; 5/PK1,803 mm (0,071")0,051 mm1,905 mm (0,075")Polymide0,3 m琥珀色Log in for availability1 * 5 件Specifications:CE certificate
More
Specifications
Cat. No.MFLB95820-13型号95820-13透明度TranslucentLength per pack1,5 m (5 ft.)最大压力19 bar (275 psi)最高温度480 °C (896 °F)最小温度–250 °C (–418 °F)Regulatory approvalsUSPSterilisation method autoclaveYesSterilisation method EtOYesSterilisation method gammaYes工作压力19 bar (275 psi)质保3 MonthsCE certificate