Masterflex® 聚乙烯燃料和润滑剂管,Avantor®
供应商: MASTERFLEX PREMIUM
关于此项目
No swelling or embrittlement.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimise downtime
- Regulatory approvals to match your unique application
材质: TYGON F-4040-A
应用范围: 燃料和工业用润滑油、汽油、煤油、加热油、切削化合物,以及乙二醇冷却液。耐受大多数碳氢化合物。
特点: 抗脆化和溶胀,耐臭氧和紫外线,萃取率低。半透明黄色。
认证: 管根据 GMP 要求生产。
温度范围: -35 到 165°F(-37 到 74°C)
灭菌: 不推荐。
10 Options Available Below
Product Details & Documents
No swelling or embrittlement.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimise downtime
- Regulatory approvals to match your unique application
材质: TYGON F-4040-A
应用范围: 燃料和工业用润滑油、汽油、煤油、加热油、切削化合物,以及乙二醇冷却液。耐受大多数碳氢化合物。
特点: 抗脆化和溶胀,耐臭氧和紫外线,萃取率低。半透明黄色。
认证: 管根据 GMP 要求生产。
温度范围: -35 到 165°F(-37 到 74°C)
灭菌: 不推荐。
Documents
Certificates
按批号搜索Specifications for Product Family
规格
- Catalog No:
- MFLB95633-08
- MFLB95633-03
- MFLB95633-01
- MFLB95633-10
- MFLB95633-05
- MFLB95633-02
- MFLB95633-00
- MFLB95633-09
- MFLB95633-04
- MFLB95633-06
- 型号:
- 95633-08
- 95633-03
- 95633-01
- 95633-10
- 95633-05
- 95633-02
- 95633-00
- 95633-09
- 95633-04
- 95633-06
- 透明度:
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- GMP compliant:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Length per pack:
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 最大压力:
- 2,1 bar (30 psi)
- 2,1 bar (30 psi)
- 3,6 bar (52 psi)
- 1,4 bar (21 psi)
- 1,4 bar (21 psi)
- 2,6 bar (37 psi)
- 3,6 bar (52 psi)
- 1,7 bar (25 psi)
- 1,7 bar (25 psi)
- 1,3 bar (19 psi)
- 最高温度:
- 73 °C (165 °F)
- 73 °C (165 °F)
- 73 °C (165 °F)
- 73 °C (165 °F)
- 73 °C (165 °F)
- 73 °C (165 °F)
- 73 °C (165 °F)
- 73 °C (165 °F)
- 73 °C (165 °F)
- 73 °C (165 °F)
- 最小温度:
- –37 °C (–35 °F)
- –37 °C (–35 °F)
- –37 °C (–35 °F)
- –37 °C (–35 °F)
- –37 °C (–35 °F)
- –37 °C (–35 °F)
- –37 °C (–35 °F)
- –37 °C (–35 °F)
- –37 °C (–35 °F)
- –37 °C (–35 °F)
- Sterilisation method autoclave:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Sterilisation method EtO:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Sterilisation method gamma:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- 工作压力:
- 2,1 bar (30 psi)
- 2,1 bar (30 psi)
- 3,6 bar (52 psi)
- 1,4 bar (21 psi)
- 1,4 bar (21 psi)
- 2,6 bar (37 psi)
- 3,6 bar (52 psi)
- 1,7 bar (25 psi)
- 1,7 bar (25 psi)
- 1,3 bar (19 psi)
- 质保:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
规格
Product Family Options
产品信息
- 顺序说明Tubing int.Ø壁厚Tubing ext.ØTubing material长度颜色可用库存Price
- 01Transfer Tubing, Tygon® F-4040-A Fuel and Lubricant, 1/2" ID x 3/4" OD; 50 Ft12,8 mm (0,5")3,2 mm19,1 mm (0,75")Tygon® Fuel and Lubricant15,2 m黄色Log in 查看库存1 * 50 Feet规格:CE certificate
More
Specifications
Cat. No.MFLB95633-08型号95633-08透明度TranslucentGMP compliantYesLength per pack15,2 m (50 ft.)最大压力2,1 bar (30 psi)最高温度73 °C (165 °F)最小温度–37 °C (–35 °F)Sterilisation method autoclaveNoSterilisation method EtONoSterilisation method gammaNo工作压力2,1 bar (30 psi)质保1 yearCE certificate - 02Transfer Tubing, Tygon® F-4040-A Fuel and Lubricant, 1/4" ID x 3/8" OD; 50 Ft6,4 mm (0,25")1,6 mm9,6 mm (0,375")Tygon® Fuel and Lubricant15,2 m黄色Log in 查看库存1 * 50 Feet规格:CE certificate
More
Specifications
Cat. No.MFLB95633-03型号95633-03透明度TranslucentGMP compliantYesLength per pack15,2 m (50 ft.)最大压力2,1 bar (30 psi)最高温度73 °C (165 °F)最小温度–37 °C (–35 °F)Sterilisation method autoclaveNoSterilisation method EtONoSterilisation method gammaNo工作压力2,1 bar (30 psi)质保1 yearCE certificate - 03Transfer Tubing, Tygon® F-4040-A Fuel and Lubricant, 1/8" ID x 1/4" OD; 50 Ft3,2 mm (0,125")1,6 mm6,4 mm (0,25")Tygon® Fuel and Lubricant15,2 m黄色Log in 查看库存1 * 50 Feet规格:CE certificate
More
Specifications
Cat. No.MFLB95633-01型号95633-01透明度TranslucentGMP compliantYesLength per pack15,2 m (50 ft.)最大压力3,6 bar (52 psi)最高温度73 °C (165 °F)最小温度–37 °C (–35 °F)Sterilisation method autoclaveNoSterilisation method EtONoSterilisation method gammaNo工作压力3,6 bar (52 psi)质保1 yearCE certificate - 04Transfer Tubing, Tygon® F-4040-A Fuel and Lubricant, 3/4" ID x 1" OD; 50 Ft19,1 mm (0,75")3,2 mm25,4 mm (1")Tygon® Fuel and Lubricant15,2 m黄色Log in 查看库存1 * 50 Feet规格:CE certificate
More
Specifications
Cat. No.MFLB95633-10型号95633-10透明度TranslucentGMP compliantYesLength per pack15,2 m (50 ft.)最大压力1,4 bar (21 psi)最高温度73 °C (165 °F)最小温度–37 °C (–35 °F)Sterilisation method autoclaveNoSterilisation method EtONoSterilisation method gammaNo工作压力1,4 bar (21 psi)质保1 yearCE certificate - 05Transfer Tubing, Tygon® F-4040-A Fuel and Lubricant, 3/8" ID x 1/2" OD; 50 Ft9,5 mm (0,375")1,6 mm12,7 mm (0,5")Tygon® Fuel and Lubricant15,2 m黄色Log in 查看库存1 * 50 Feet规格:CE certificate
More
Specifications
Cat. No.MFLB95633-05型号95633-05透明度TranslucentGMP compliantYesLength per pack15,2 m (50 ft.)最大压力1,4 bar (21 psi)最高温度73 °C (165 °F)最小温度–37 °C (–35 °F)Sterilisation method autoclaveNoSterilisation method EtONoSterilisation method gammaNo工作压力1,4 bar (21 psi)质保1 yearCE certificate - 06Transfer Tubing, Tygon® F-4040-A Fuel and Lubricant, 3/16" ID x 5/16" OD; 50 Ft4,8 mm (0,1875")1,6 mm8 mm (0,3125")Tygon® Fuel and Lubricant15,2 m黄色Log in 查看库存1 * 50 Feet规格:CE certificate
More
Specifications
Cat. No.MFLB95633-02型号95633-02透明度TranslucentGMP compliantYesLength per pack15,2 m (50 ft.)最大压力2,6 bar (37 psi)最高温度73 °C (165 °F)最小温度–37 °C (–35 °F)Sterilisation method autoclaveNoSterilisation method EtONoSterilisation method gammaNo工作压力2,6 bar (37 psi)质保1 yearCE certificate - 07Transfer Tubing, Tygon® F-4040-A Fuel and Lubricant, 3/32" ID x 3/16" OD; 50 Ft2,4 mm (0,09375")1,2 mm4,8 mm (0,1875")Tygon® Fuel and Lubricant15,2 m黄色Log in 查看库存1 * 50 Feet规格:CE certificate
More
Specifications
Cat. No.MFLB95633-00型号95633-00透明度TranslucentGMP compliantYesLength per pack15,2 m (50 ft.)最大压力3,6 bar (52 psi)最高温度73 °C (165 °F)最小温度–37 °C (–35 °F)Sterilisation method autoclaveNoSterilisation method EtONoSterilisation method gammaNo工作压力3,6 bar (52 psi)质保1 yearCE certificate - 08Transfer Tubing, Tygon® F-4040-A Fuel and Lubricant, 5/8" ID x 7/8" OD; 50 Ft16 mm (0,625")3,2 mm22,2 mm (0,875")Tygon® Fuel and Lubricant15,2 m黄色Log in 查看库存1 * 50 Feet规格:CE certificate
More
Specifications
Cat. No.MFLB95633-09型号95633-09透明度TranslucentGMP compliantYesLength per pack15,2 m (50 ft.)最大压力1,7 bar (25 psi)最高温度73 °C (165 °F)最小温度–37 °C (–35 °F)Sterilisation method autoclaveNoSterilisation method EtONoSterilisation method gammaNo工作压力1,7 bar (25 psi)质保1 yearCE certificate - 09Transfer Tubing, Tygon® F-4040-A Fuel and Lubricant, 5/16" ID x 7/16" OD; 50 Ft8 mm (0,3125")1,6 mm11,1 mm (0,4375")Tygon® Fuel and Lubricant15,2 m黄色Log in 查看库存1 * 50 Feet规格:CE certificate
More
Specifications
Cat. No.MFLB95633-04型号95633-04透明度TranslucentGMP compliantYesLength per pack15,2 m (50 ft.)最大压力1,7 bar (25 psi)最高温度73 °C (165 °F)最小温度–37 °C (–35 °F)Sterilisation method autoclaveNoSterilisation method EtONoSterilisation method gammaNo工作压力1,7 bar (25 psi)质保1 yearCE certificate - 10Transfer Tubing, Tygon® F-4040-A Fuel and Lubricant, 7/16" ID x 9/16" OD; 50 Ft11,1 mm (0,4375")1,6 mm14,3 mm (0,5625")Tygon® Fuel and Lubricant15,2 m黄色Log in 查看库存1 * 50 Feet规格:CE certificate
More
Specifications
Cat. No.MFLB95633-06型号95633-06透明度TranslucentGMP compliantYesLength per pack15,2 m (50 ft.)最大压力1,3 bar (19 psi)最高温度73 °C (165 °F)最小温度–37 °C (–35 °F)Sterilisation method autoclaveNoSterilisation method EtONoSterilisation method gammaNo工作压力1,3 bar (19 psi)质保1 yearCE certificate