Orders LinkContactUsLinkComponent
Masterflex® 单通道注射泵,Avantor®
Masterflex® 单通道注射泵,Avantor®
Certificates
Masterflex® 单通道注射泵,Avantor®
2 Options Available Below
Product Details & Documents
非常适合高精确,低流速,低压应用。
  • 背光照明字母数字LCD显示屏,协助指导您完成设定和运行步骤
  • 非易失性存储器,即使泵关闭时也可存储记忆当前设定参数
  • 精度达 +/-0.5%
单通道注射泵,使用微机马达控制,精确传动。微机自动识别注射泵所使用的注射器,并据其选择正确的显示单位。精确控制流速,可低至0.2uL/h。动态量程(6000:1)非常宽,使用更灵活。通过前面板键盘能够编程所有的命令。即使泵正在运行,也可以随时更改参数设定。
 : 使用说明书,电源线
Specifications for Product Family
Specifications
  • 型号:
  • 74900-00
  • 74900-05
  • 准确度:
  • ±1%
  • ±0.5%
  • Linear force:
  • 20 lbs.
  • 20 lbs.
  • 可重现性:
  • ±0,1%
  • ±0,2%
  • Syringe size accepted:
  • 10 µl - 60 ml
  • 10 µl - 60 ml
  • Syringe travel rate:
  • <126,7"
  • <126,7"
  • 质保:
  • 1 year
  • 1 year
Specifications
Product Family Options
Product Information
  • 顺序
    说明
    高度
    长度
    宽度
    Min. flow rate per syringe
    Max. flow rate per syringe
    Power
    Availability
    Price
  • MFLA74900-00
    01
    Single-syringe infusion pump, 115 VAC
    15,2 cm
    22,86 cm
    102 mm
    0,0033 µl/min (1 pl/min)
    8,33 ml/min
    115 VAC
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
    Specifications:
    More
    Specifications
    Cat. No.
    MFLA74900-00
    型号
    74900-00
    准确度
    ±1%
    Linear force
    20 lbs.
    可重现性
    ±0,1%
    Syringe size accepted
    10 µl - 60 ml
    Syringe travel rate
    <126,7"
    质保
    1 year
  • MFLA74900-05
    02
    Single-syringe infusion pump, 230 VAC
    15,2 cm
    22,86 cm
    102 mm
    0,0033 µl/min
    8,33 ml/min
    230 VAC
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
    Specifications:
    CE Compliant
    More
    Specifications
    Cat. No.
    MFLA74900-05
    型号
    74900-05
    准确度
    ±0.5%
    Linear force
    20 lbs.
    可重现性
    ±0,2%
    Syringe size accepted
    10 µl - 60 ml
    Syringe travel rate
    <126,7"
    质保