Masterflex® L/S® Easy-Load II泵头,用于高性能精密泵管,Avantor®
Supplier: MASTERFLEX PROPRIETARY
Certificates
About this item
自动固位装置可将泵管牢固固定到位,无需人工调节,—只需装载泵管,然后启动泵即可。
- 四辊轴马达提高了泵头的压力性能和机械稳定性,同时降低了流动脉动。
- 进改良的咬合床符合几何学原理,延长了泵管使用寿命。
- 聚苯硫醚(PPS)外壳,不锈钢(SS)和冷轧钢(CRS)两种转子型号可选—CRS转子的泵头价格实惠,而SS抗化学性更佳且更耐用。
- 77201-62型泵头咬合可调节,您可放松咬合以延长泵管使用寿命,或调紧咬合以获得更高的压力。
- 每一种兼容高效精密泵管的泵头都可兼容L/S® 15, L/S® 24, L/S® 36, 和L/S® 35泵管。
- 流速范围可达1.7~2900 mL/min (1 ~ 600 rpm)。
这些泵头可容纳 4 种尺寸的高性能精密管路。高性能精密管路的管壁比精密管路的管壁厚,可生成更高的压力、更大的吸引升力,并可更好地泵送粘性流体。
随机配备:单通道安装配件和一根 15'' (38-cm) 长硅胶泵管。如需叠连多个泵头(最多可达驱动器的规定极限数额),请单独订购 2、3 及 4 通道配件。
Delivery: Includes single-channel mounting hardware. For stacking multiple heads (up to the specified limits of your drive) order two- three- or four-channel hardware separately.
- Environmentally Preferable:
Specifications
- 型号:
- 77200-52
- 77200-32
- 77200-62
- 77201-62
- Gear ratio:
- 1:1
- 1:1
- 1:1
- 1:1
- 通道数量:
- 1
- 1
- 1
- 1
- Occlusion:
- Fixed
- Fixed
- Fixed
- Adjustable
- Open-head sensor:
- No
- Yes
- No
- No
- Optimum RPM range:
- 0,02 - 600 rpm
- 0,02 - 600 rpm
- 0,02 - 600 rpm
- 0,02 - 600 rpm
- Pump series:
- L/S
- L/S
- L/S
- L/S
- Suction lift:
- 8,8 m (29 ft.)
- 8,8 m (29 ft.)
- 8,8 m (29 ft.)
- 8,8 m (29 ft.)
- Tubing retention:
- Automatic
- Automatic
- Automatic
- Automatic
- Tubing sizes accepted:
- L/S® 15, L/S 24, L/S 35, L/S 36
- L/S® 15, L/S 24, L/S 35, L/S 36
- L/S® 15, L/S 24, L/S 35, L/S 36
- L/S® 15, L/S 24, L/S 35, L/S 36
- Volume per revolution with L/S 15 tubing:
- 1,7 ml
- 1,7 ml
- 1,7 ml
- 1,7 ml
- Volume per revolution with L/S 24 tubing:
- 2,8 ml
- 2,8 ml
- 2,8 ml
- 2,8 ml
- Volume per revolution with L/S 35 tubing:
- 3,8 ml
- 3,8 ml
- 3,8 ml
- 3,8 ml
- Volume per revolution with L/S 36 tubing:
- 4,8 ml
- 4,8 ml
- 4,8 ml
- 4,8 ml
- 质保:
- 2 years
- 2 years
- 2 years
- 2 years