Masterflex® Normally Open Low-Flow Compact Switches, Avantor®
证书
关于此项目
Provides reliable operation in clean air and water applications.
- Constructed of Noryl®, 316 stainless steel, and epoxy
These normally open SPST switches are specifically designed for reliable operation in clean air and water applications. Select from gas or liquid flow switches. Use a 50-µm filter to protect your switch. Switches include 18" (45.7 cm) L leads.
Delivery: Includes 18" (45.7 cm) L leads.
- Environmentally Preferable:
规格
- 型号:
- 32778-34
- 32778-33
- 32778-36
- 32778-32
- 32778-31
- 32778-35
- 32778-30
- 最小流速:
- 2,5 ft.³/min
- 5 ft.³/min
- 1 ft.³/min
- 25 ft.³/min
- 最高运行温度:
- 100 °C (212 °F)
- 100 °C (212 °F)
- 100 °C (212 °F)
- 100 °C (212 °F)
- 100 °C (212 °F)
- 100 °C (212 °F)
- 100 °C (212 °F)
- 浸湿材料:
- Noryl®, 316 stainless steel, and epoxy
- Noryl®, 316 stainless steel, and epoxy
- Noryl®, 316 stainless steel, and epoxy
- Noryl®, 316 stainless steel, and epoxy
- Noryl®, 316 stainless steel, and epoxy
- Noryl®, 316 stainless steel, and epoxy
- Noryl®, 316 stainless steel, and epoxy
- 高度:
- 1869"
- 1869"
- 1869"
- 1869"
- 1869"
- 1869"
- 1869"
- 质保:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year