Masterflex® 常开液体 SPST 开关,Avantor®
Supplier: MASTERFLEX PROPRIETARY
Certificates
About this item
Minimizes flow constriction.
- Provides low pressure drop
These SPST switches feature a unique, dual-diameter, internal bore and piston configuration to minimize flow constriction and provide low pressure drop. Use a 150 um filter to protect your switch.
Delivery information: Includes 1/4" male quick connect terminals.
- Environmentally Preferable:
Specifications
- 最大压力:
- 200 psi
- 200 psi
- 200 psi
- 200 psi
- 最高运行温度:
- 100 °C (212 °F)
- 100 °C (212 °F)
- 100 °C (212 °F)
- 100 °C (212 °F)
- 浸湿材料:
- PP, Ryton® PPS, Viton®, and 316 SS
- PP, Ryton® PPS, Viton®, and 316 SS
- PP, Ryton® PPS, Viton®, and 316 SS
- PP, Ryton® PPS, Viton®, and 316 SS
- 连接:
- 1/4" male quick connect terminals
- 1/4" male quick connect terminals
- 1/4" male quick connect terminals
- 1/4" male quick connect terminals
- 功率:
- 20 VA VAC
- 20 VA VAC
- 20 VA VAC
- 20 VA VAC
- 质保:
- 1 year
- 1 year
- 1 year
- 1 year