Orders LinkContactUsLinkComponent
Masterflex® 阀筒,Avantor®
Masterflex® 阀筒,Avantor®
Masterflex® 阀筒,Avantor®
Certificates
Masterflex® 阀筒,Avantor®
16 Options Available Below
Product Details & Documents
Control your flow with the addition of a valve.
  • Aluminum is economical and good for general use with noncorrosive gases and liquids
  • 316 Stainless steel (SS) withstands higher temperatures and pressures, and features excellent chemical compatibility
选择适合您的框架的阀筒。将这三个针阀插入流量计框架来控制流速。选择16转,高分辨率的阀筒,保证更精确的流速。如果您的流量泵管不是采用mL/min单位,将它转换成mL/min,从下表中选择该流量范围内的阀。转换因子见1255页。
DELRIN® TM E.I. DuPont de Nemours & Co.
KEL-F® TM 3M Co.
VITON® TM E. I. DuPont Elastomers L.L.C.
警告:本产品未获准、不适用于且不得用于医疗、临床、手术或其他面向患者的应用领域。
Specifications for Product Family
规格
  • 型号:
  • 03217-82
  • 03217-84
  • 03217-86
  • 03217-88
  • 03217-92
  • 03217-96
  • 03218-61
  • 03218-62
  • 03218-63
  • 03218-64
  • 03218-67
  • 03218-68
  • 03218-69
  • 03218-70
  • 03218-72
  • 03218-74
  • Min. air flow rate:
  • 5000 ml/min
  • 20000 ml/min
  • 60001 ml/min
  • 20000 ml/min
  • 5000 ml/min
  • 60001 ml/min
  • 201 ml/min
  • 1001 ml/min
  • 2501 ml/min
  • 6201 ml/min
  • 201 ml/min
  • 1001 ml/min
  • 2501 ml/min
  • 6201 ml/min
  • 0 ml/min
  • 0 ml/min
  • Max. air flow rate:
  • 20000 ml/min
  • 60000 ml/min
  • 60001 ml/min
  • 60000 ml/min
  • 20000 ml/min
  • 600001 ml/min
  • 1000 ml/min
  • 2500 ml/min
  • 6200 ml/min
  • 6201 ml/min
  • 1000 ml/min
  • 2500 ml/min
  • 6200 ml/min
  • 6201 ml/min
  • 200 ml/min
  • 200 ml/min
  • Min. water flow rate:
  • 0 ml/min
  • 0 ml/min
  • 0 ml/min
  • 0 ml/min
  • 0 ml/min
  • 0 ml/min
  • 3,1 ml/min
  • 30 ml/min
  • 71 ml/min
  • 650 ml/min
  • 3,1 ml/min
  • 30 ml/min
  • 71 ml/min
  • 650 ml/min
  • 0 ml/min
  • 0 ml/min
  • Max. water flow rate:
  • 350 ml/min
  • 1200 ml/min
  • 3500 ml/min
  • 1200 ml/min
  • 350 ml/min
  • 3500 ml/min
  • 30 ml/min
  • 70 ml/min
  • 200 ml/min
  • 650 ml/min
  • 30 ml/min
  • 70 ml/min
  • 200 ml/min
  • 650 ml/min
  • 6 ml/min
  • 6 ml/min
  • O-Ring material:
  • Viton®
  • Viton®
  • Viton®
  • Buna-N
  • Buna-N
  • Buna-N
  • Buna-N
  • Buna-N
  • Buna-N
  • Buna-N
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Buna-N
  • Viton®
  • 宽度:
  • 1754"
  • 1754"
  • 1754"
  • 1754"
  • 1754"
  • 1754"
  • 1754"
  • 1754"
  • 1754"
  • 1754"
  • 1754"
  • 1754"
  • 1754"
  • 1754"
  • 1754"
  • 1754"
  • 质保:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
规格
Product Family Options
产品信息
  • 顺序
    说明
    Maximum Flow Rate
    Housing material
    可用库存
    Price
  • MFLA03217-82
    01
    Variable-Area Flowmeter Valve Cartridge, 10-Turn, for Stainless Steel Frame Systems, Low Flow
    20000 ml/min
    316 stainless steel
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
    规格:
    More
    Specifications
    Cat. No.
    MFLA03217-82
    型号
    03217-82
    Min. air flow rate
    5000 ml/min
    Max. air flow rate
    20000 ml/min
    Min. water flow rate
    0 ml/min
    Max. water flow rate
    350 ml/min
    O-Ring material
    Viton®
    宽度
    1754"
    质保
    1 year
  • MFLA03217-84
    02
    Variable-Area Flowmeter Valve Cartridge, 10-Turn, for Stainless Steel Frame Systems, Medium Flow
    60000 ml/min
    316 stainless steel
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
    规格:
    More
    Specifications
    Cat. No.
    MFLA03217-84
    型号
    03217-84
    Min. air flow rate
    20000 ml/min
    Max. air flow rate
    60000 ml/min
    Min. water flow rate
    0 ml/min
    Max. water flow rate
    1200 ml/min
    O-Ring material
    Viton®
    宽度
    1754"
    质保
    1 year
  • MFLA03217-86
    03
    Variable-Area Flowmeter Valve Cartridge, 10-Turn, for Stainless Steel Frame Systems, High Flow
    60001 ml/min
    316 stainless steel
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
    规格:
    More
    Specifications
    Cat. No.
    MFLA03217-86
    型号
    03217-86
    Min. air flow rate
    60001 ml/min
    Max. air flow rate
    60001 ml/min
    Min. water flow rate
    0 ml/min
    Max. water flow rate
    3500 ml/min
    O-Ring material
    Viton®
    宽度
    1754"
    质保
    1 year
  • MFLA03217-88
    04
    Variable-Area Flowmeter Valve Cartridge, 10-Turn, for Aluminum Frame Systems, Medium Flow
    60000 ml/min
    Aluminium
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
    规格:
    More
    Specifications
    Cat. No.
    MFLA03217-88
    型号
    03217-88
    Min. air flow rate
    20000 ml/min
    Max. air flow rate
    60000 ml/min
    Min. water flow rate
    0 ml/min
    Max. water flow rate
    1200 ml/min
    O-Ring material
    Buna-N
    宽度
    1754"
    质保
    1 year
  • MFLA03217-92
    05
    Variable-Area Flowmeter Valve Cartridge, 10-Turn, for Aluminum Frame Systems, Low Flow
    20000 ml/min
    Aluminium
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
    规格:
    More
    Specifications
    Cat. No.
    MFLA03217-92
    型号
    03217-92
    Min. air flow rate
    5000 ml/min
    Max. air flow rate
    20000 ml/min
    Min. water flow rate
    0 ml/min
    Max. water flow rate
    350 ml/min
    O-Ring material
    Buna-N
    宽度
    1754"
    质保
    1 year
  • MFLA03217-96
    06
    Variable-Area Flowmeter Valve Cartridge, 10-Turn, for Aluminum Frame Systems, High Flow
    600001 ml/min
    Aluminium
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
    规格:
    More
    Specifications
    Cat. No.
    MFLA03217-96
    型号
    03217-96
    Min. air flow rate
    60001 ml/min
    Max. air flow rate
    600001 ml/min
    Min. water flow rate
    0 ml/min
    Max. water flow rate
    3500 ml/min
    O-Ring material
    Buna-N
    宽度
    1754"
    质保
    1 year
  • MFLA03218-61
    07
    Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Aluminum Frame Systems, Range 3
    1000 ml/min
    Aluminium
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
    规格:
    More
    Specifications
    Cat. No.
    MFLA03218-61
    型号
    03218-61
    Min. air flow rate
    201 ml/min
    Max. air flow rate
    1000 ml/min
    Min. water flow rate
    3,1 ml/min
    Max. water flow rate
    30 ml/min
    O-Ring material
    Buna-N
    宽度
    1754"
    质保
    1 year
  • MFLA03218-62
    08
    Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Aluminum Frame Systems, Range 4
    2500 ml/min
    Aluminium
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
    规格:
    More
    Specifications
    Cat. No.
    MFLA03218-62
    型号
    03218-62
    Min. air flow rate
    1001 ml/min
    Max. air flow rate
    2500 ml/min
    Min. water flow rate
    30 ml/min
    Max. water flow rate
    70 ml/min
    O-Ring material
    Buna-N
    宽度
    1754"
    质保
    1 year
  • MFLA03218-63
    09
    Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Aluminum Frame Systems, Range 5
    6200 ml/min
    Aluminium
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
    规格:
    More
    Specifications
    Cat. No.
    MFLA03218-63
    型号
    03218-63
    Min. air flow rate
    2501 ml/min
    Max. air flow rate
    6200 ml/min
    Min. water flow rate
    71 ml/min
    Max. water flow rate
    200 ml/min
    O-Ring material
    Buna-N
    宽度
    1754"
    质保
    1 year
  • MFLA03218-64
    10
    Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Aluminum Frame Systems, Range 6
    6201 ml/min
    Aluminium
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
    规格:
    More
    Specifications
    Cat. No.
    MFLA03218-64
    型号
    03218-64
    Min. air flow rate
    6201 ml/min
    Max. air flow rate
    6201 ml/min
    Min. water flow rate
    650 ml/min
    Max. water flow rate
    650 ml/min
    O-Ring material
    Buna-N
    宽度
    1754"
    质保
    1 year
  • MFLA03218-67
    11
    Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Stainless Steel Frame Systems, Range 3
    1000 ml/min
    316 stainless steel
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
    规格:
    More
    Specifications
    Cat. No.
    MFLA03218-67
    型号
    03218-67
    Min. air flow rate
    201 ml/min
    Max. air flow rate
    1000 ml/min
    Min. water flow rate
    3,1 ml/min
    Max. water flow rate
    30 ml/min
    O-Ring material
    Viton®
    宽度
    1754"
    质保
    1 year
  • MFLA03218-68
    12
    Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Stainless Steel Frame Systems, Range 4
    2500 ml/min
    316 stainless steel
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
    规格:
    More
    Specifications
    Cat. No.
    MFLA03218-68
    型号
    03218-68
    Min. air flow rate
    1001 ml/min
    Max. air flow rate
    2500 ml/min
    Min. water flow rate
    30 ml/min
    Max. water flow rate
    70 ml/min
    O-Ring material
    Viton®
    宽度
    1754"
    质保
    1 year
  • MFLA03218-69
    13
    Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Stainless Steel Frame Systems, Range 5
    6200 ml/min
    316 stainless steel
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
    规格:
    More
    Specifications
    Cat. No.
    MFLA03218-69
    型号
    03218-69
    Min. air flow rate
    2501 ml/min
    Max. air flow rate
    6200 ml/min
    Min. water flow rate
    71 ml/min
    Max. water flow rate
    200 ml/min
    O-Ring material
    Viton®
    宽度
    1754"
    质保
    1 year
  • MFLB03218-70
    14
    Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Stainless Steel Frame Systems, Range 6
    6201 ml/min
    316 stainless steel
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
    规格:
    More
    Specifications
    Cat. No.
    MFLB03218-70
    型号
    03218-70
    Min. air flow rate
    6201 ml/min
    Max. air flow rate
    6201 ml/min
    Min. water flow rate
    650 ml/min
    Max. water flow rate
    650 ml/min
    O-Ring material
    Viton®
    宽度
    1754"
    质保
    1 year
  • MFLA03218-72
    15
    Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Aluminum Frame Systems, Range 1
    200 ml/min
    Aluminium
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
    规格:
    More
    Specifications
    Cat. No.
    MFLA03218-72
    型号
    03218-72
    Min. air flow rate
    0 ml/min
    Max. air flow rate
    200 ml/min
    Min. water flow rate
    0 ml/min
    Max. water flow rate
    6 ml/min
    O-Ring material
    Buna-N
    宽度
    1754"
    质保
    1 year
  • MFLA03218-74
    16
    Variable-Area Flowmeter Valve Cartridge, 16-Turn, for Stainless Steel Frame Systems, Range 1
    200 ml/min
    316 stainless steel
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
    规格:
    More
    Specifications
    Cat. No.
    MFLA03218-74
    型号
    03218-74
    Min. air flow rate
    0 ml/min
    Max. air flow rate
    200 ml/min
    Min. water flow rate
    0 ml/min
    Max. water flow rate
    6 ml/min
    O-Ring material
    Viton®
    宽度
    1754"
    质保
    1 year