Orders LinkContactUsLinkComponent
Masterflex® 有阀门大流量校正流量计,Avantor®
Masterflex® 有阀门大流量校正流量计,Avantor®
Masterflex® 有阀门大流量校正流量计,Avantor®
Certificates
Masterflex® 有阀门大流量校正流量计,Avantor®
Masterflex® 有阀门大流量校正流量计,Avantor®
11 Options Available Below
Product Details & Documents
Don’t sacrifice suitability for precision.
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. Valved meters have valves mounted at the bottom/inlet, typically used for positive pressure with liquids and gases. The valve can be reconfigured to mount at the top/outlet, often for vacuum applications. All correlated flowmeters come with data sheets for water and air at standard temperature and pressure (STP).
Specifications for Product Family
Specifications
  • 型号:
  • 03217-32
  • 03217-34
  • 03217-36
  • 03217-87
  • 03217-90
  • 03219-33
  • 03219-35
  • 03219-37
  • 03294-32
  • 03294-34
  • 03294-36
  • 准确度:
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • Float material:
  • Glass
  • 316 SS
  • Carboloy
  • Sapphire
  • Glass
  • 316 SS
  • Carboloy
  • Glass
  • 316 SS
  • Carboloy
  • Flow Tube material:
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • Borosilicate glass
  • 最高运行温度:
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 65 °C (150 °F)
  • 65 °C (150 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • O-Ring material:
  • Buna-N
  • Buna-N
  • Buna-N
  • PTFE
  • PTFE
  • Viton®
  • Viton®
  • Viton®
  • Buna-N
  • Buna-N
  • Buna-N
  • 可重复性:
  • ±0,25%
  • ±0,25%
  • ±0,25%
  • ±0,25% full-scale
  • ±0,25%
  • ±0,25%
  • ±0,25%
  • ±0,25%
  • ±0,25%
  • ±0,25%
  • ±0,25%
  • 质保:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
Specifications
Product Family Options
Product Information
  • 顺序
    说明
    Valve material
    Fitting material
    Housing material
    Process connection
    Availability
    Price
  • MFLA03217-32
    01
    150-mm Correlated Flowmeter, w/ valve, aluminum, 23,742 L/min air
    Aluminum
    Aluminum
    Aluminium
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
    Specifications:
    More
    Specifications
    Cat. No.
    MFLA03217-32
    型号
    03217-32
    准确度
    ±2% full-scale
    Float material
    Glass
    Flow Tube material
    Borosilicate glass
    最高运行温度
    121 °C (250 °F)
    O-Ring material
    Buna-N
    可重复性
    ±0,25%
    质保
    1 year
  • MFLA03217-34
    02
    150-mm Correlated Flowmeter, w/ valve, aluminum, 45,227 L/min air
    Aluminum
    Aluminum
    Aluminium
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
    Specifications:
    More
    Specifications
    Cat. No.
    MFLA03217-34
    型号
    03217-34
    准确度
    ±2% full-scale
    Float material
    316 SS
    Flow Tube material
    Borosilicate glass
    最高运行温度
    121 °C (250 °F)
    O-Ring material
    Buna-N
    可重复性
    ±0,25%
    质保
    1 year
  • MFLA03217-36
    03
    150-mm Correlated Flowmeter, w/ valve, aluminum, 66,346 L/min air
    Aluminum
    Aluminum
    Aluminium
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
    Specifications:
    More
    Specifications
    Cat. No.
    MFLA03217-36
    型号
    03217-36
    准确度
    ±2% full-scale
    Float material
    Carboloy
    Flow Tube material
    Borosilicate glass
    最高运行温度
    121 °C (250 °F)
    O-Ring material
    Buna-N
    可重复性
    ±0,25%
    质保
    1 year
  • MFLB03217-87
    04
    150-mm Correlated Flowmeter, w/ valve, PTFE, 22,536 L/min air
    PTFE and Kel-F®
    PTFE and Kel-F®
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
    Specifications:
    More
    Specifications
    Cat. No.
    MFLB03217-87
    型号
    03217-87
    准确度
    ±2% full-scale
    Float material
    Flow Tube material
    Borosilicate glass
    最高运行温度
    65 °C (150 °F)
    O-Ring material
    PTFE
    可重复性
    ±0,25% full-scale
    质保
    1 year
  • MFLA03217-90
    05
    150-mm Correlated Flowmeter, w/ valve, PTFE, 29,56 L/min air
    PTFE and Kel-F®
    PTFE and Kel-F®
    PTFE
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
    Specifications:
    More
    Specifications
    Cat. No.
    MFLA03217-90
    型号
    03217-90
    准确度
    ±2% full-scale
    Float material
    Sapphire
    Flow Tube material
    Borosilicate glass
    最高运行温度
    65 °C (150 °F)
    O-Ring material
    PTFE
    可重复性
    ±0,25%
    质保
    1 year
  • MFLA03219-33
    06
    150-mm Correlated Flowmeter, w/ valve, 316SS, 23,742 L/min air
    316 SS
    316 stainless steel
    316 stainless steel
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
    Specifications:
    More
    Specifications
    Cat. No.
    MFLA03219-33
    型号
    03219-33
    准确度
    ±2% full-scale
    Float material
    Glass
    Flow Tube material
    Borosilicate glass
    最高运行温度
    121 °C (250 °F)
    O-Ring material
    Viton®
    可重复性
    ±0,25%
    质保
    1 year
  • MFLA03219-35
    07
    150-mm Correlated Flowmeter, w/ valve, 316SS, 45,227 L/min air
    316 SS
    316 stainless steel
    316 stainless steel
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
    Specifications:
    More
    Specifications
    Cat. No.
    MFLA03219-35
    型号
    03219-35
    准确度
    ±2% full-scale
    Float material
    316 SS
    Flow Tube material
    Borosilicate glass
    最高运行温度
    121 °C (250 °F)
    O-Ring material
    Viton®
    可重复性
    ±0,25%
    质保
    1 year
  • MFLA03219-37
    08
    150-mm Correlated Flowmeter, w/ valve, 316SS, 66,346 L/min air
    316 SS
    316 stainless steel
    316 stainless steel
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
    Specifications:
    More
    Specifications
    Cat. No.
    MFLA03219-37
    型号
    03219-37
    准确度
    ±2% full-scale
    Float material
    Carboloy
    Flow Tube material
    Borosilicate glass
    最高运行温度
    121 °C (250 °F)
    O-Ring material
    Viton®
    可重复性
    ±0,25%
    质保
    1 year
  • MFLB03294-32
    09
    150-mm Correlated Flowmeter, w/ valve, brass, 23,742 L/min air
    Chrome-plated brass
    Chrome-plated brass
    Brass
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
    Specifications:
    More
    Specifications
    Cat. No.
    MFLB03294-32
    型号
    03294-32
    准确度
    ±2% full-scale
    Float material
    Glass
    Flow Tube material
    Borosilicate glass
    最高运行温度
    121 °C (250 °F)
    O-Ring material
    Buna-N
    可重复性
    ±0,25%
    质保
    1 year
  • MFLA03294-34
    10
    150-mm Correlated Flowmeter, w/ valve, brass, 45,227 L/min air
    Chrome-plated brass
    Chrome-plated brass
    Brass
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
    Specifications:
    More
    Specifications
    Cat. No.
    MFLA03294-34
    型号
    03294-34
    准确度
    ±2% full-scale
    Float material
    316 SS
    Flow Tube material
    Borosilicate glass
    最高运行温度
    121 °C (250 °F)
    O-Ring material
    Buna-N
    可重复性
    ±0,25%
    质保
    1 year
  • MFLA03294-36
    11
    150-mm Correlated Flowmeter, w/ valve, brass, 66,346 L/min air
    Chrome-plated brass
    Chrome-plated brass
    Brass
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
    Specifications:
    More
    Specifications
    Cat. No.
    MFLA03294-36
    型号
    03294-36
    准确度
    ±2% full-scale
    Float material
    Carboloy
    Flow Tube material
    Borosilicate glass
    最高运行温度
    121 °C (250 °F)
    O-Ring material
    Buna-N
    可重复性
    ±0,25%
    质保
    1 year