Masterflex® 装有高分辨率阀门的校正流量计,Avantor®
Don’t sacrifice suitability for precision.
- Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. Valved meters have valves mounted at the bottom/inlet, typically used for positive pressure with liquids and gases. The valve can be reconfigured to mount at the top/outlet, often for vacuum applications. All correlated flowmeters come with data sheets for water and air at standard temperature and pressure (STP).
Don’t sacrifice suitability for precision.
- Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. Valved meters have valves mounted at the bottom/inlet, typically used for positive pressure with liquids and gases. The valve can be reconfigured to mount at the top/outlet, often for vacuum applications. All correlated flowmeters come with data sheets for water and air at standard temperature and pressure (STP).
- Catalog No:
- MFLA32044-00
- MFLA32044-02
- MFLB32044-04
- MFLA32044-06
- MFLA32044-08
- MFLB32044-10
- MFLA32044-12
- MFLA32044-14
- MFLB32044-15
- MFLB32044-16
- MFLB32044-18
- MFLB32044-20
- MFLB32044-22
- MFLB32044-24
- MFLB32044-26
- MFLA32044-28
- MFLA32044-30
- MFLB32044-32
- MFLA32044-34
- MFLB32045-00
- MFLB32045-02
- MFLB32045-04
- MFLB32045-06
- MFLB32045-08
- MFLB32045-10
- MFLB32045-12
- MFLB32045-14
- MFLA32045-15
- MFLB32045-17
- MFLB32045-18
- MFLB32045-20
- MFLA32045-26
- MFLB32045-32
- MFLA32046-01
- MFLA32046-03
- MFLB32046-05
- MFLB32046-07
- MFLB32046-09
- MFLB32046-11
- MFLA32046-13
- MFLA32046-15
- MFLA32046-16
- MFLB32046-18
- MFLB32046-19
- MFLB32046-21
- MFLA32046-23
- MFLB32046-31
- MFLB32046-33
- MFLA32046-35
- 型号:
- 32044-00
- 32044-02
- 32044-04
- 32044-06
- 32044-08
- 32044-10
- 32044-12
- 32044-14
- 32044-15
- 32044-16
- 32044-18
- 32044-20
- 32044-22
- 32044-24
- 32044-26
- 32044-28
- 32044-30
- 32044-32
- 32044-34
- 32045-00
- 32045-02
- 32045-04
- 32045-06
- 32045-08
- 32045-10
- 32045-12
- 32045-14
- 32045-15
- 32045-17
- 32045-18
- 32045-20
- 32045-26
- 32045-32
- 32046-01
- 32046-03
- 32046-05
- 32046-07
- 32046-09
- 32046-11
- 32046-13
- 32046-15
- 32046-16
- 32046-18
- 32046-19
- 32046-21
- 32046-23
- 32046-31
- 32046-33
- 32046-35
- 准确度:
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- ±2% full-scale
- Float material:
- Glass
- 316 SS
- Glass
- 316 SS
- Glass
- 316 SS
- Glass
- 316 SS
- Glass
- Glass
- 316 SS
- Glass
- 316 SS
- Glass
- 316 SS
- Glass
- 316 SS
- Glass
- 316 SS
- Glass
- 316 SS
- Glass
- 316 SS
- Glass
- 316 SS
- Glass
- 316 SS
- Glass
- 316 SS
- 316 SS
- Glass
- 316 SS
- Glass
- Glass
- 316 SS
- Glass
- 316 SS
- Glass
- 316 SS
- Glass
- 316 SS
- Glass
- 316 SS
- 316 SS
- Glass
- 316 SS
- 316 SS
- Glass
- 316 SS
- Flow Tube material:
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- Borosilicate glass
- 最高运行温度:
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- 121 °C (250 °F)
- O-Ring material:
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Buna-N
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- Viton®
- 可重复性:
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- ±0,25%
- 质保:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
Product Family Options
- 顺序说明Maximum Flow RateFluid typeValve materialFitting materialHousing materialProcess connection
- 0165-mm Correlated Flowmeter, high-res valve, aluminum, 5,8 ml/min air0,065 ml/minwaterAluminumAluminumAluminium0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLA32044-00型号32044-00准确度±2% full-scaleFloat materialGlassFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialBuna-N可重复性±0,25%质保1 year - 0265-mm Correlated Flowmeter, high-res valve, aluminum, 16 ml/min air0,283 ml/minwaterAluminumAluminumAluminium0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLA32044-02型号32044-02准确度±2% full-scaleFloat material316 SSFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialBuna-N可重复性±0,25%质保1 year - 0365-mm Correlated Flowmeter, high-res valve, aluminum, 49 ml/min air0,55 ml/minwaterAluminumAluminumAluminium0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLB32044-04型号32044-04准确度±2% full-scaleFloat materialGlassFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialBuna-N可重复性±0,25%质保1 year - 0465-mm Correlated Flowmeter, high-res valve, aluminum, 145 ml/min air2,38 ml/minwaterAluminumAluminumAluminium0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLA32044-06型号32044-06准确度±2% full-scaleFloat material316 SSFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialBuna-N可重复性±0,25%质保1 year - 0565-mm Correlated Flowmeter, high-res valve, aluminum, 104 ml/min air175 ml/minwaterAluminumAluminumAluminium0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLA32044-08型号32044-08准确度±2% full-scaleFloat materialGlassFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialBuna-N可重复性±0,25%质保1 year - 0665-mm Correlated Flowmeter, high-res valve, aluminum, 299 ml/min air7,74 ml/minwaterAluminumAluminumAluminium0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLB32044-10型号32044-10准确度±2% full-scaleFloat material316 SSFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialBuna-N可重复性±0,25%质保1 year - 0765-mm Correlated Flowmeter, high-res valve, aluminum, 202 ml/min air2,6 ml/minwaterAluminumAluminumAluminium0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLA32044-12型号32044-12准确度±2% full-scaleFloat materialGlassFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialBuna-N可重复性±0,25%质保1 year - 0865-mm Correlated Flowmeter, high-res valve, aluminum, 522 ml/min air12 ml/minwaterAluminumAluminumAluminium0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLA32044-14型号32044-14准确度±2% full-scaleFloat material316 SSFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialBuna-N可重复性±0,25%质保1 year - 0965-mm Correlated Flowmeter, high-res valve, aluminum, 992 ml/min air20,5 ml/minwaterAluminumAluminumAluminium0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLB32044-15型号32044-15准确度±2% full-scaleFloat materialGlassFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialBuna-N可重复性±0,25%质保1 year - 1065-mm Correlated Flowmeter, high-res valve, aluminum, 1,25 L/min air27 ml/minwaterAluminumAluminumAluminium0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLB32044-16型号32044-16准确度±2% full-scaleFloat materialGlassFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialBuna-N可重复性±0,25%质保1 year - 1165-mm Correlated Flowmeter, high-res valve, aluminum, 2,52 L/min air70,7 ml/minwaterAluminumAluminumAluminium0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLB32044-18型号32044-18准确度±2% full-scaleFloat material316 SSFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialBuna-N可重复性±0,25%质保1 year - 1265-mm Correlated Flowmeter, high-res valve, aluminum, 2,04 L/min air39,7 ml/minwaterAluminumAluminumAluminium0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLB32044-20型号32044-20准确度±2% full-scaleFloat materialGlassFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialBuna-N可重复性±0,25%质保1 year - 1365-mm Correlated Flowmeter, high-res valve, aluminum, 3,99 L/min air108,3 ml/minwaterAluminumAluminumAluminium0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLB32044-22型号32044-22准确度±2% full-scaleFloat material316 SSFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialBuna-N可重复性±0,25%质保1 year - 1465-mm Correlated Flowmeter, high-res valve, aluminum, 2,678 L/min air52 ml/minwaterAluminumAluminumAluminium0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLB32044-24型号32044-24准确度±2% full-scaleFloat materialGlassFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialBuna-N可重复性±0,25%质保1 year - 1565-mm Correlated Flowmeter, high-res valve, aluminum, 4,922 L/min air150 ml/minwaterAluminumAluminumAluminium0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLB32044-26型号32044-26准确度±2% full-scaleFloat material316 SSFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialBuna-N可重复性±0,25%质保1 year - 1665-mm Correlated Flowmeter, high-res valve, aluminum, 6,318 L/min air147 ml/minwaterAluminumAluminumAluminium0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLA32044-28型号32044-28准确度±2% full-scaleFloat materialGlassFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialBuna-N可重复性±0,25%质保1 year - 1765-mm Correlated Flowmeter, high-res valve, aluminum, 12,058 L/min air364 ml/minwaterAluminumAluminumAluminium0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLA32044-30型号32044-30准确度±2% full-scaleFloat material316 SSFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialBuna-N可重复性±0,25%质保1 year - 1865-mm Correlated Flowmeter, high-res valve, aluminum, 13,153 L/min air309 ml/minwaterAluminumAluminumAluminium0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLB32044-32型号32044-32准确度±2% full-scaleFloat materialGlassFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialBuna-N可重复性±0,25%质保1 year - 1965-mm Correlated Flowmeter, high-res valve, aluminum, 24,680 L/min air745 ml/minwaterAluminumAluminumAluminium0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLA32044-34型号32044-34准确度±2% full-scaleFloat material316 SSFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialBuna-N可重复性±0,25%质保1 year - 2065-mm Correlated Flowmeter, high-res valve, brass, 5,8 ml/min air0,065 ml/minwaterChrome-plated brassChrome-plated brassBrass0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLB32045-00型号32045-00准确度±2% full-scaleFloat materialGlassFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialBuna-N可重复性±0,25%质保1 year - 2165-mm Correlated Flowmeter, high-res valve, brass, 16,7 ml/min air0,283 ml/minwaterChrome-plated brassChrome-plated brassBrass0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLB32045-02型号32045-02准确度±2% full-scaleFloat material316 SSFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialBuna-N可重复性±0,25%质保1 year - 2265-mm Correlated Flowmeter, high-res valve, brass, 48,7 ml/min air0,55 ml/minwaterChrome-plated brassChrome-plated brassBrass0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLB32045-04型号32045-04准确度±2% full-scaleFloat materialGlassFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialBuna-N可重复性±0,25%质保1 year - 2365-mm Correlated Flowmeter, high-res valve, brass, 145 ml/min air2,38 ml/minwaterChrome-plated brassChrome-plated brassBrass0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLB32045-06型号32045-06准确度±2% full-scaleFloat material316 SSFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialBuna-N可重复性±0,25%质保1 year - 2465-mm Correlated Flowmeter, high-res valve, brass, 104 ml/min air1,75 ml/minwaterChrome-plated brassChrome-plated brassBrass0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLB32045-08型号32045-08准确度±2% full-scaleFloat materialGlassFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialBuna-N可重复性±0,25%质保1 year - 2565-mm Correlated Flowmeter, high-res valve, brass, 299 ml/min air7,74 ml/minwaterChrome-plated brassChrome-plated brassBrass0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLB32045-10型号32045-10准确度±2% full-scaleFloat material316 SSFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialBuna-N可重复性±0,25%质保1 year - 2665-mm Correlated Flowmeter, high-res valve, brass, 202 ml/min air2,6 ml/minwaterChrome-plated brassChrome-plated brassBrass0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLB32045-12型号32045-12准确度±2% full-scaleFloat materialGlassFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialBuna-N可重复性±0,25%质保1 year - 2765-mm Correlated Flowmeter, high-res valve, brass, 522 ml/min air12 ml/minwaterChrome-plated brassChrome-plated brassBrass0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLB32045-14型号32045-14准确度±2% full-scaleFloat material316 SSFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialBuna-N可重复性±0,25%质保1 year - 2865-mm Correlated Flowmeter, high-res valve, brass, 992 ml/min air20,5 ml/minwaterChrome-plated brassChrome-plated brassBrass0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLA32045-15型号32045-15准确度±2% full-scaleFloat materialGlassFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialBuna-N可重复性±0,25%质保1 year - 2965-mm Correlated Flowmeter, high-res valve, brass, 1946 ml/min air55,5 ml/minwaterChrome-plated brassChrome-plated brassBrass0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLB32045-17型号32045-17准确度±2% full-scaleFloat material316 SSFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialBuna-N可重复性±0,25%质保1 year - 3065-mm Correlated Flowmeter, high-res valve, brass, 2520 ml/min air70,7 ml/minwaterChrome-plated brassChrome-plated brassBrass0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLB32045-18型号32045-18准确度±2% full-scaleFloat material316 SSFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialBuna-N可重复性±0,25%质保1 year - 3165-mm Correlated Flowmeter, high-res valve, brass, 2040 ml/min air39,7 ml/minwaterChrome-plated brassChrome-plated brassBrass0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLB32045-20型号32045-20准确度±2% full-scaleFloat materialGlassFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialBuna-N可重复性±0,25%质保1 year - 3265-mm Correlated Flowmeter, high-res valve, brass, 4922 ml/min air150 ml/minwaterChrome-plated brassChrome-plated brassBrass0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLA32045-26型号32045-26准确度±2% full-scaleFloat material316 SSFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialBuna-N可重复性±0,25%质保1 year - 3365-mm Correlated Flowmeter, high-res valve, brass, 13,153 L/min air309 ml/minwaterChrome-plated brassChrome-plated brassBrass0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLB32045-32型号32045-32准确度±2% full-scaleFloat materialGlassFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialBuna-N可重复性±0,25%质保1 year - 3465-mm Correlated Flowmeter, hi-res valve, 316 SS, 5,8 ml/min air0,065 ml/minwater316 SS316 stainless steel316 stainless steel0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLA32046-01型号32046-01准确度±2% full-scaleFloat materialGlassFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialViton®可重复性±0,25%质保1 year - 3565-mm Correlated Flowmeter, hi-res valve, 316 SS, 16,7 ml/min air0,283 ml/minwater316 SS316 stainless steel316 stainless steel0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLA32046-03型号32046-03准确度±2% full-scaleFloat material316 SSFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialViton®可重复性±0,25%质保1 year - 3665-mm Correlated Flowmeter, hi-res valve, 316 SS, 48,7 ml/min air0,55 ml/minwater316 SS316 stainless steel316 stainless steel0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLB32046-05型号32046-05准确度±2% full-scaleFloat materialGlassFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialViton®可重复性±0,25%质保1 year - 3765-mm Correlated Flowmeter, hi-res valve, 316 SS, 145 ml/min air2,38 ml/minwater316 SS316 stainless steel316 stainless steel0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLB32046-07型号32046-07准确度±2% full-scaleFloat material316 SSFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialViton®可重复性±0,25%质保1 year - 3865-mm Correlated Flowmeter, hi-res valve, 316 SS, 104 ml/min air1,75 ml/minwater316 SS316 stainless steel316 stainless steel0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLB32046-09型号32046-09准确度±2% full-scaleFloat materialGlassFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialViton®可重复性±0,25%质保1 year - 3965-mm Correlated Flowmeter, hi-res valve, 316 SS, 299 ml/min air7,74 ml/minwater316 SS316 stainless steel316 stainless steel0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLB32046-11型号32046-11准确度±2% full-scaleFloat material316 SSFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialViton®可重复性±0,25%质保1 year - 4065-mm Correlated Flowmeter, hi-res valve, 316 SS, 202,1 ml/min air2,6 ml/minwater316 SS316 stainless steel316 stainless steel0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLA32046-13型号32046-13准确度±2% full-scaleFloat materialGlassFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialViton®可重复性±0,25%质保1 year - 4165-mm Correlated Flowmeter, hi-res valve, 316 SS, 522 ml/min air12 ml/minwater316 SS316 stainless steel316 stainless steel0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLA32046-15型号32046-15准确度±2% full-scaleFloat material316 SSFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialViton®可重复性±0,25%质保1 year - 4265-mm Correlated Flowmeter, hi-res valve, 316 SS, 992 ml/min air20,5 ml/minwater316 SS316 stainless steel316 stainless steel0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLA32046-16型号32046-16准确度±2% full-scaleFloat materialGlassFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialViton®可重复性±0,25%质保1 year - 4365-mm Correlated Flowmeter, hi-res valve, 316 SS, 1946 ml/min air55,5 ml/minwater316 SS316 stainless steel316 stainless steel0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLB32046-18型号32046-18准确度±2% full-scaleFloat material316 SSFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialViton®可重复性±0,25%质保1 year - 4465-mm Correlated Flowmeter, hi-res valve, 316 SS, 2520 ml/min air70,7 ml/minwater316 SS316 stainless steel316 stainless steel0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLB32046-19型号32046-19准确度±2% full-scaleFloat material316 SSFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialViton®可重复性±0,25%质保1 year - 4565-mm Correlated Flowmeter, hi-res valve, 316 SS, 2040 ml/min air39,7 ml/minwater316 SS316 stainless steel316 stainless steel0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLB32046-21型号32046-21准确度±2% full-scaleFloat materialGlassFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialViton®可重复性±0,25%质保1 year - 4665-mm Correlated Flowmeter, hi-res valve, 316 SS, 3990 ml/min air108,3 ml/minwater316 SS316 stainless steel316 stainless steel0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLA32046-23型号32046-23准确度±2% full-scaleFloat material316 SSFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialViton®可重复性±0,25%质保1 year - 4765-mm Correlated Flowmeter, hi-res valve, 316 SS, 12,058 L/min air364 ml/minwater316 SS316 stainless steel316 stainless steel0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLB32046-31型号32046-31准确度±2% full-scaleFloat material316 SSFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialViton®可重复性±0,25%质保1 year - 4865-mm Correlated Flowmeter, hi-res valve, 316 SS, 13,153 L/min air309 ml/minwater316 SS316 stainless steel316 stainless steel0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLB32046-33型号32046-33准确度±2% full-scaleFloat materialGlassFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialViton®可重复性±0,25%质保1 year - 4965-mm Correlated Flowmeter, hi-res valve, 316 SS, 24,680 L/min air745 ml/minwater316 SS316 stainless steel316 stainless steel0,125" NPT(F)1 * 1 件
Specifications
Cat. No.MFLA32046-35型号32046-35准确度±2% full-scaleFloat material316 SSFlow Tube materialBorosilicate glass最高运行温度121 °C (250 °F)O-Ring materialViton®可重复性±0,25%质保1 year