Orders LinkContactUsLinkComponent
Masterflex® 空气用铝制流量计,Avantor®
Masterflex® 空气用铝制流量计,Avantor®
Masterflex® 空气用铝制流量计,Avantor®
Certificates
Masterflex® 空气用铝制流量计,Avantor®
Masterflex® 空气用铝制流量计,Avantor®
9 Options Available Below
Product Details & Documents
Easily read graph-style measurements directly.
  • Direct-reading flowmeters feature a calibrated scale for a specific medium - no need to convert readings to get flow rate│Use with air for laboratory and industrial applications
Use for high pressure industrial applications—install in any configuration, even next to a bend in your pipeline. A calibrated retention spring eliminates the need to mount flowmeters vertically or run straight lengths of pipe prior to or after the meter. These rugged flowmeters are relatively insensitive to shock and vibrations—the spring, piston assembly, and internal magnet compensate for rough process environments.
Specifications for Product Family
规格
  • 准确度:
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • ±2% full-scale
  • 最高运行温度:
  • 115,5 °C (240 °F)
  • 115,5 °C (240 °F)
  • 115,5 °C (240 °F)
  • 115,5 °C (240 °F)
  • 115,5 °C (240 °F)
  • 115,5 °C (240 °F)
  • 115,5 °C (240 °F)
  • 115,5 °C (240 °F)
  • 115,5 °C (240 °F)
  • 最大压力:
  • 1000 psi
  • 1000 psi
  • 1000 psi
  • 1000 psi
  • 1000 psi
  • 1000 psi
  • 1000 psi
  • 1000 psi
  • 1000 psi
  • O-Ring material:
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • Viton®
  • 可重复性:
  • ±1%
  • ±1%
  • ±1%
  • ±1%
  • ±1%
  • ±1%
  • ±1%
  • ±1%
  • ±1%
  • 质保:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
规格
Product Family Options
产品信息
  • 顺序
    说明
    Min. flow rate
    Maximum Flow Rate
    Media type
    Valve
    Fitting material
    Housing material
    Process connection
    可用库存
    Price
  • MFLB32206-02
    01
    Variable-Area Flowmeter, Direct Reading, Graph-Style, Aluminum Housing, 5 ft.³/min Air
    0,5 ft.³/min
    5 ft.³/min
    Air
    No
    Anodized aluminum
    Aluminium
    0,25" NPT(F)
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
    规格:
    CSA Certified
    ISO9001
    UL Listed
    More
    Specifications
    Cat. No.
    MFLB32206-02
    准确度
    ±2% full-scale
    最高运行温度
    115,5 °C (240 °F)
    最大压力
    1000 psi
    O-Ring material
    Viton®
    可重复性
    ±1%
    质保
    1 year
    CSA Certified
    ISO9001
    UL Listed
  • MFLB32206-04
    02
    Variable-Area Flowmeter, Direct Reading, Graph-Style, Aluminum Housing, 10 ft.³/min Air
    1 ft.³/min
    10 ft.³/min
    Air
    No
    Anodized aluminum
    Aluminium
    0,25" NPT(F)
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
    规格:
    CSA Certified
    ISO9001
    UL Listed
    More
    Specifications
    Cat. No.
    MFLB32206-04
    准确度
    ±2% full-scale
    最高运行温度
    115,5 °C (240 °F)
    最大压力
    1000 psi
    O-Ring material
    Viton®
    可重复性
    ±1%
    质保
    1 year
    CSA Certified
    ISO9001
    UL Listed
  • MFLB32206-06
    03
    Variable-Area Flowmeter, Direct Reading, Graph-Style, Aluminum Housing, 20 ft.³/min Air
    2 ft.³/min
    20 ft.³/min
    Air
    No
    Anodized aluminum
    Aluminium
    0,25" NPT(F)
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
    规格:
    CSA Certified
    ISO9001
    UL Listed
    More
    Specifications
    Cat. No.
    MFLB32206-06
    准确度
    ±2% full-scale
    最高运行温度
    115,5 °C (240 °F)
    最大压力
    1000 psi
    O-Ring material
    Viton®
    可重复性
    ±1%
    质保
    1 year
    CSA Certified
    ISO9001
    UL Listed
  • MFLB32206-08
    04
    Variable-Area Flowmeter, Direct Reading, Graph-Style, Aluminum Housing, 30 ft.³/min Air
    3 ft.³/min
    30 ft.³/min
    Air
    No
    Anodized aluminum
    Aluminium
    0,25" NPT(F)
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
    规格:
    CSA Certified
    ISO9001
    UL Listed
    More
    Specifications
    Cat. No.
    MFLB32206-08
    准确度
    ±2% full-scale
    最高运行温度
    115,5 °C (240 °F)
    最大压力
    1000 psi
    O-Ring material
    Viton®
    可重复性
    ±1%
    质保
    1 year
    CSA Certified
    ISO9001
    UL Listed
  • MFLA32206-12
    05
    Variable-Area Flowmeter, Direct Reading, Graph-Style, Aluminum Housing, 25 ft.³/min Air
    3 ft.³/min
    25 ft.³/min
    Air
    No
    Anodized aluminum
    Aluminium
    0,5" NPT(F)
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
    规格:
    CSA Certified
    ISO9001
    UL Listed
    More
    Specifications
    Cat. No.
    MFLA32206-12
    准确度
    ±2% full-scale
    最高运行温度
    115,5 °C (240 °F)
    最大压力
    1000 psi
    O-Ring material
    Viton®
    可重复性
    ±1%
    质保
    1 year
    CSA Certified
    ISO9001
    UL Listed
  • MFLB32206-56
    06
    Variable-Area Flowmeter, Direct Reading, Graph-Style, Aluminum Housing, with Built-In Pressure Gauge, 20 ft.³/min Air
    2 ft.³/min
    20 ft.³/min
    Air
    No
    Anodized aluminum
    Aluminium
    0,25" NPT(F)
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
    规格:
    CSA Certified
    ISO9001
    UL Listed
    More
    Specifications
    Cat. No.
    MFLB32206-56
    准确度
    ±2% full-scale
    最高运行温度
    115,5 °C (240 °F)
    最大压力
    1000 psi
    O-Ring material
    Viton®
    可重复性
    ±1%
    质保
    1 year
    CSA Certified
    ISO9001
    UL Listed
  • MFLB32206-62
    07
    Variable-Area Flowmeter, Direct Reading, Graph-Style, Aluminum Housing, with Built-In Pressure Gauge, 25 ft.³/min Air
    3 ft.³/min
    25 ft.³/min
    Air
    No
    Anodized aluminum
    Aluminium
    0,5" NPT(F)
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
    规格:
    CSA Certified
    ISO9001
    UL Listed
    More
    Specifications
    Cat. No.
    MFLB32206-62
    准确度
    ±2% full-scale
    最高运行温度
    115,5 °C (240 °F)
    最大压力
    1000 psi
    O-Ring material
    Viton®
    可重复性
    ±1%
    质保
    1 year
    CSA Certified
    ISO9001
    UL Listed
  • MFLB32206-66
    08
    Variable-Area Flowmeter, Direct Reading, Graph-Style, Aluminum Housing, with Built-In Pressure Gauge, 100 ft.³/min Air
    10 ft.³/min
    100 ft.³/min
    Air
    No
    Anodized aluminum
    Aluminium
    0,5" NPT(F)
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
    规格:
    CSA Certified
    ISO9001
    UL Listed
    More
    Specifications
    Cat. No.
    MFLB32206-66
    准确度
    ±2% full-scale
    最高运行温度
    115,5 °C (240 °F)
    最大压力
    1000 psi
    O-Ring material
    Viton®
    可重复性
    ±1%
    质保
    1 year
    CSA Certified
    ISO9001
    UL Listed
  • MFLB32206-72
    09
    Variable-Area Flowmeter, Direct Reading, Graph-Style, Aluminum Housing, with Built-In Pressure Gauge, 150 ft.³/min Air
    15 ft.³/min
    150 ft.³/min
    Air
    No
    Anodized aluminum
    Aluminium
    0,75" NPT(F)
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
    规格:
    CSA Certified
    ISO9001
    UL Listed
    More
    Specifications
    Cat. No.
    MFLB32206-72
    准确度
    ±2% full-scale
    最高运行温度
    115,5 °C (240 °F)
    最大压力
    1000 psi
    O-Ring material
    Viton®
    可重复性
    ±1%
    质保
    1 year
    CSA Certified
    ISO9001
    UL Listed