ContactUsLinkComponent
CPC® Caps for MPC Quick-Disconnect Fittings
CPC® Caps for MPC Quick-Disconnect Fittings
供应商:  MASTERFLEX PREMIUM
证书
undefined
CPC® Caps for MPC Quick-Disconnect Fittings
供应商:  MASTERFLEX PREMIUM
证书
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
规格
  • 最大运行压力:
  • 60 psi
  • 60 psi
  • 最高温度:
  • 148 °C (300 °F)
  • 148 °C (300 °F)
  • 最大真空度:
  • 634 mm Hg (25" Hg)
  • 634 mm Hg (25" Hg)
  • 最小温度:
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • 灭菌:
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • 质保:
  • None
规格
相关信息