Orders LinkContactUsLinkComponent
Masterflex® 带倒钩的 NPT 母螺纹直型转接头,Avantor®
Masterflex® 带倒钩的 NPT 母螺纹直型转接头,Avantor®
Certificates
Masterflex® 带倒钩的 NPT 母螺纹直型转接头,Avantor®
Masterflex® 带倒钩的 NPT 母螺纹直型转接头,Avantor®
7 Options Available Below
Product Details & Documents
No tools needed for assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
Specifications for Product Family
规格
  • 型号:
  • 31500-40
  • 31500-41
  • 31501-40
  • 31501-41
  • 31701-85
  • 31702-20
  • 31702-22
  • Orifice dimensions:
  • 1,588 mm (0,0625")
  • 1,588 mm (0,0625")
  • 1,588 mm (0,0625")
  • 1,588 mm (0,0625")
  • 灭菌:
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • 质保:
  • 1 year
  • 1 year
  • 1 year
规格
Product Family Options
产品信息
  • 顺序
    说明
    Barb size/tubing int.Ø (port 2)
    Connection Type
    Fitting material
    Flow pattern
    Thread size NPT(F) (port 1)
    Thread size NPT(M) (port 1)
    可用库存
    Price
  • MFLA31500-40
    01
    Fitting, Nylon, Straight, Hosebarb to Thread Adapter, 1/8" ID x 1/8" NPT(F); 25/PK
    1/8"
    Thread to Barb
    Nylon
    Straight
    1/8"
    1/8"
    Log in 查看库存
    Log in 查看库存
    1 * 25 件
    1 * 25 件
    规格:
    More
    Specifications
    Cat. No.
    MFLA31500-40
    型号
    31500-40
    Orifice dimensions
    1,588 mm (0,0625")
    灭菌
    Ethylene oxide (ETO) only
    质保
  • MFLA31500-41
    02
    Fitting, Nylon, Straight, Hosebarb to Thread Adapter, 1/8" ID x 1/4" NPT(F); 25/PK
    1/8"
    Thread to Barb
    Nylon
    Straight
    1/4"
    1/4"
    Log in 查看库存
    Log in 查看库存
    1 * 25 件
    1 * 25 件
    规格:
    More
    Specifications
    Cat. No.
    MFLA31500-41
    型号
    31500-41
    Orifice dimensions
    1,588 mm (0,0625")
    灭菌
    Ethylene oxide (ETO) only
    质保
  • MFLA31501-40
    03
    Fitting, Clear Polypropylene, Straight, Hose Barb to Threaded Adapter, 1/8" ID x 1/8" NPT(F); 25/PK
    1/8"
    Thread to Barb
    Polypropylene
    Straight
    1/8"
    1/8"
    Log in 查看库存
    Log in 查看库存
    1 * 25 件
    1 * 25 件
    规格:
    More
    Specifications
    Cat. No.
    MFLA31501-40
    型号
    31501-40
    Orifice dimensions
    1,588 mm (0,0625")
    灭菌
    Autoclave or ethylene oxide (ETO)
    质保
  • MFLA31501-41
    04
    Fitting, Clear Polypropylene, Straight, Hose Barb to Threaded Adapter, 1/8" ID x 1/4" NPT(F); 25/PK
    1/8"
    Thread to Barb
    Polypropylene
    Straight
    1/4"
    1/4"
    Log in 查看库存
    Log in 查看库存
    1 * 25 件
    1 * 25 件
    规格:
    More
    Specifications
    Cat. No.
    MFLA31501-41
    型号
    31501-41
    Orifice dimensions
    1,588 mm (0,0625")
    灭菌
    Autoclave or ethylene oxide (ETO)
    质保
  • MFLB31701-85
    05
    Fitting, Nylon, Straight, Hosebarb to Thread Adapter, 1/4" ID x 1/4" NPT(F); 100/PK
    1/4"
    Thread to Barb
    Nylon
    Straight
    1/4"
    Log in 查看库存
    Log in 查看库存
    1 * 100 件
    1 * 100 件
    规格:
    More
    Specifications
    Cat. No.
    MFLB31701-85
    型号
    31701-85
    Orifice dimensions
    灭菌
    Ethylene oxide (ETO) only
    质保
    1 year
  • MFLB31702-20
    06
    Fitting, Nylon, Straight, Hosebarb to Thread Adapter, 1/4" ID x 1/4" NPT(F); 10/PK
    1/4"
    Thread to Barb
    Nylon
    Straight
    1/4"
    Log in 查看库存
    Log in 查看库存
    1 * 10 件
    1 * 10 件
    规格:
    More
    Specifications
    Cat. No.
    MFLB31702-20
    型号
    31702-20
    Orifice dimensions
    灭菌
    Ethylene oxide (ETO) only
    质保
    1 year
  • MFLB31702-22
    07
    Fitting, Nylon, Straight, Hosebarb to Thread Adapter, 3/8" ID x 1/4" NPT(F); 10/PK
    3/8"
    Thread to Barb
    Nylon
    Straight
    1/4"
    Log in 查看库存
    Log in 查看库存
    1 * 10 件
    1 * 10 件
    规格:
    More
    Specifications
    Cat. No.
    MFLB31702-22
    型号
    31702-22
    Orifice dimensions
    灭菌
    Ethylene oxide (ETO) only
    质保
    1 year