Masterflex® 倒刺T型接头,Avantor®
关于此项目
No tools needed for assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed tee fittings are designed for quick and easy assembly. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
13 Options Available Below
Product Details & Documents
No tools needed for assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed tee fittings are designed for quick and easy assembly. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
Documents
Certificates
按批号搜索Specifications for Product Family
规格
- Catalog No:
- MFLA30620-25
- MFLA30620-26
- MFLA30620-27
- MFLA30620-28
- MFLA30620-29
- MFLA30620-30
- MFLA30620-31
- MFLA30620-32
- MFLA31208-31
- MFLA31208-32
- MFLA31208-34
- MFLA31208-35
- MFLA31208-37
- 型号:
- 30620-25
- 30620-26
- 30620-27
- 30620-28
- 30620-29
- 30620-30
- 30620-31
- 30620-32
- 31208-31
- 31208-32
- 31208-34
- 31208-35
- 31208-37
- Orifice dimensions:
- 0,125"
- 0,125"
- 0,1875"
- 0,1875"
- 0,3125"
- 0,375"
- 0,5"
- 0,6875"
- 0,0625"
- 0,03125"
- 0,125"
- 0,1875"
- 0,28125"
- 灭菌:
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- Autoclave, ethylene oxide (ETO), or gamma
- 质保:
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
规格
Product Family Options
产品信息
- 顺序说明Barb size/tubing int.Ø (port 1)Barb size/tubing int.Ø (port 2)Barb size/tubing int.Ø (port 3)Connection TypeFitting materialFlow pattern可用库存Price
- 01Fitting, Stainless Steel, Tee, Hose Barb Union, 1/8" ID1/8"1/8"1/8"Barbed UnionStainless Steel 316TLog in 查看库存1 * 1 件规格:
More
Specifications
Cat. No.MFLA30620-25型号30620-25Orifice dimensions0,125"灭菌Autoclave, ethylene oxide (ETO), or gamma质保1 Month - 02Fitting, Stainless Steel, Tee, Hose Barb Union, 3/16" ID3/16"3/16"3/16"Barbed UnionStainless Steel 316TLog in 查看库存1 * 1 件规格:
More
Specifications
Cat. No.MFLA30620-26型号30620-26Orifice dimensions0,125"灭菌Autoclave, ethylene oxide (ETO), or gamma质保1 Month - 03Fitting, Stainless Steel, Tee, Hose Barb Union, 1/4" ID1/4"1/4"1/4"Barbed UnionStainless Steel 316TLog in 查看库存1 * 1 件规格:
More
Specifications
Cat. No.MFLA30620-27型号30620-27Orifice dimensions0,1875"灭菌Autoclave, ethylene oxide (ETO), or gamma质保1 Month - 04Fitting, Stainless Steel, Tee, Hose Barb Union, 5/16" ID5/16"5/16"5/16"Barbed UnionStainless Steel 316TLog in 查看库存1 * 1 件规格:
More
Specifications
Cat. No.MFLA30620-28型号30620-28Orifice dimensions0,1875"灭菌Autoclave, ethylene oxide (ETO), or gamma质保1 Month - 05Fitting, Stainless Steel, Tee, Hose Barb Union, 3/8" ID3/8"3/8"3/8"Barbed UnionStainless Steel 316TLog in 查看库存1 * 1 件规格:
More
Specifications
Cat. No.MFLA30620-29型号30620-29Orifice dimensions0,3125"灭菌Autoclave, ethylene oxide (ETO), or gamma质保1 Month - 06Fitting, Stainless Steel, Tee, Hose Barb Union, 1/2" ID1/2"1/2"1/2"Barbed UnionStainless Steel 316TLog in 查看库存1 * 1 件规格:
More
Specifications
Cat. No.MFLA30620-30型号30620-30Orifice dimensions0,375"灭菌Autoclave, ethylene oxide (ETO), or gamma质保1 Month - 07Fitting, Stainless Steel, Tee, Hose Barb Union, 5/8" ID5/8"5/8"5/8"Barbed UnionStainless Steel 316TLog in 查看库存1 * 1 件规格:
More
Specifications
Cat. No.MFLA30620-31型号30620-31Orifice dimensions0,5"灭菌Autoclave, ethylene oxide (ETO), or gamma质保1 Month - 08Fitting, Stainless Steel, Tee, Hose Barb Union, 3/4" ID3/4"3/4"3/4"Barbed UnionStainless Steel 316TLog in 查看库存1 * 1 件规格:
More
Specifications
Cat. No.MFLA30620-32型号30620-32Orifice dimensions0,6875"灭菌Autoclave, ethylene oxide (ETO), or gamma质保1 Month - 10Fitting, 303 Stainless Steel, Tee, Hose Barb Union, 1/16" ID; 1EA1/16"1/16"1/16"Barbed UnionStainless steel 303TLog in 查看库存1 * 1 件规格:
More
Specifications
Cat. No.MFLA31208-31型号31208-31Orifice dimensions0,0625"灭菌质保1 year - 11Fitting, 303 Stainless Steel, Tee, Hose Barb Union, 1/8" ID; 1EA1/8"1/8"1/8"Barbed UnionStainless steel 303TLog in 查看库存1 * 1 件规格:
More
Specifications
Cat. No.MFLA31208-32型号31208-32Orifice dimensions0,03125"灭菌质保1 year - 12Fitting, 303 Stainless Steel, Tee, Hose Barb Union, 3/16" ID; 1EA3/16"3/16"3/16"Barbed UnionStainless steel 303TLog in 查看库存1 * 1 件规格:
More
Specifications
Cat. No.MFLA31208-34型号31208-34Orifice dimensions0,125"灭菌质保1 year - 13Fitting, 303 Stainless Steel, Tee, Hose Barb Union, 1/4" ID; 1EA1/4"1/4"1/4"Barbed UnionStainless steel 303TLog in 查看库存1 * 1 件规格:
More
Specifications
Cat. No.MFLA31208-35型号31208-35Orifice dimensions0,1875"灭菌质保1 year - 14Fitting, 303 Stainless Steel, Tee, Hose Barb Union, 3/8" ID; 1EA3/8"3/8"3/8"Barbed UnionStainless steel 303TLog in 查看库存1 * 1 件规格:
More
Specifications
Cat. No.MFLA31208-37型号31208-37Orifice dimensions0,28125"灭菌质保1 year