Orders LinkContactUsLinkComponent
Masterflex® 直通倒刺接头,Avantor®
Masterflex® 直通倒刺接头,Avantor®
Certificates
Masterflex® 直通倒刺接头,Avantor®
26 Options Available Below
Product Details & Documents
Quick and easy assembly without tools.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Chemical resistant
These economical straight barbed fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
Specifications for Product Family
规格
  • 型号:
  • 06365-11
  • 06365-22
  • 06365-33
  • 30612-51
  • 31407-00
  • 31407-02
  • 40610-00
  • 40610-01
  • 40610-02
  • 40610-03
  • 40610-04
  • 40610-05
  • 40610-06
  • 40622-52
  • 40622-57
  • 40625-91
  • 40640-00
  • 40640-01
  • 40640-03
  • 40640-05
  • 40640-06
  • 50622-46
  • 50622-49
  • 50626-46
  • 50626-49
  • 51518-05
  • Orifice dimensions:
  • 0,7925 mm (0,03125")
  • 1,588 mm (0,0625")
  • 1,588 mm (0,0625")
  • 27,78 mm (1,09375")
  • 45,242 mm (1,78125")
  • 3,175 mm (0,125")
  • 4,763 mm (0,1875")
  • 6,35 mm (0,25")
  • 8,73 mm (0,34375")
  • 12,7 mm (0,5")
  • 14,29 mm (0,5625")
  • 19,84 mm (0,78125")
  • 0,125"
  • 0,1875"
  • 0,34375"
  • 0,5625"
  • 0,78125"
  • 灭菌:
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene Oxide (ETO) or Gamma
  • Ethylene Oxide (ETO) or Gamma
  • Ethylene Oxide (ETO) or Autoclave
  • Ethylene Oxide (ETO) or Gamma
  • Ethylene Oxide (ETO)
  • 质保:
  • 1 year
  • 1 year
  • Out of box
  • Out of box
  • Out of box
  • Out of box
  • Out of box
  • Out of box
  • Out of box
  • Out of box
  • Out of box
  • Out of box
  • Out of box
  • Out of box
规格
Product Family Options
产品信息
  • 顺序
    说明
    Barb size/tubing int.Ø (port 1)
    Barb size/tubing int.Ø (port 2)
    Connection Type
    Fitting material
    Flow pattern
    可用库存
    Price
  • MFLA06365-11
    07
    Fitting, Polypropylene, Straight, Hose Barb Union, ¹⁄₁₆" ID; 25/PK
    ¹⁄₁₆"
    ¹⁄₁₆"
    Barbed Union
    Polypropylene
    Straight
    Log in 查看库存
    Log in 查看库存
    1 * 25 件
    1 * 25 件
    规格:
    More
    Specifications
    Cat. No.
    MFLA06365-11
    型号
    06365-11
    Orifice dimensions
    0,7925 mm (0,03125")
    灭菌
    Autoclave or ethylene oxide (ETO)
    质保
  • MFLA06365-22
    08
    Fitting, Polypropylene, Straight, Hose Barb Union, ⅛" ID; 25/PK
    ⅛"
    ⅛"
    Barbed Union
    Polypropylene
    Straight
    Log in 查看库存
    Log in 查看库存
    1 * 25 件
    1 * 25 件
    规格:
    More
    Specifications
    Cat. No.
    MFLA06365-22
    型号
    06365-22
    Orifice dimensions
    1,588 mm (0,0625")
    灭菌
    Autoclave or ethylene oxide (ETO)
    质保
  • MFLA06365-33
    09
    Fitting, Polypropylene, Straight, Hose Barb Union, ³⁄₃₂" ID; 25/PK
    ³⁄₃₂"
    ³⁄₃₂"
    Barbed Union
    Polypropylene
    Straight
    Log in 查看库存
    Log in 查看库存
    1 * 25 件
    1 * 25 件
    规格:
    More
    Specifications
    Cat. No.
    MFLA06365-33
    型号
    06365-33
    Orifice dimensions
    1,588 mm (0,0625")
    灭菌
    Autoclave or ethylene oxide (ETO)
    质保
  • 25
    Fitting, Polypropylene, Straight, Hose Barb Union, ³⁄₁₆" ID; 10/PK
    ³⁄₁₆"
    ³⁄₁₆"
    Barbed Union
    Polypropylene
    Straight
    Log in 查看库存
    Log in 查看库存
    1 * 10 件
    1 * 10 件
    规格:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLA30612-51
    型号
    30612-51
    Orifice dimensions
    灭菌
    Autoclave or ethylene oxide (ETO)
    质保
    CE certificate
  • MFLA31407-00
    32
    Fitting, Black Polypropylene, Straight, Hose Barb Union, 1 ¼" ID; 5/PK
    1¼"
    1¼"
    Barbed Union
    Polypropylene
    Straight
    Log in 查看库存
    Log in 查看库存
    1 * 5 件
    1 * 5 件
    规格:
    More
    Specifications
    Cat. No.
    MFLA31407-00
    型号
    31407-00
    Orifice dimensions
    27,78 mm (1,09375")
    灭菌
    Autoclave or ethylene oxide (ETO)
    质保
    1 year
  • MFLA31407-02
    33
    Fitting, Black Polypropylene, Straight, Hose Barb Union, 2" ID; 5/PK
    2"
    2"
    Barbed Union
    Polypropylene
    Straight
    Log in 查看库存
    Log in 查看库存
    1 * 5 件
    1 * 5 件
    规格:
    More
    Specifications
    Cat. No.
    MFLA31407-02
    型号
    31407-02
    Orifice dimensions
    45,242 mm (1,78125")
    灭菌
    Autoclave or ethylene oxide (ETO)
    质保
    1 year
  • 34
    Fitting, Polypropylene, Straight, Hose Barb Union, ¼" ID; 10/PK
    ¼"
    ¼"
    Barbed Union
    Polypropylene
    Straight
    Log in 查看库存
    Log in 查看库存
    1 * 10 件
    1 * 10 件
    规格:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLA40610-00
    型号
    40610-00
    Orifice dimensions
    3,175 mm (0,125")
    灭菌
    Ethylene oxide (ETO) or gamma
    质保
    Out of box
    CE certificate
  • 35
    Fitting, Polypropylene, Straight, Hose Barb Union, ⁵⁄₁₆" ID; 10/PK
    ⁵⁄₁₆"
    ⁵⁄₁₆"
    Barbed Union
    Polypropylene
    Straight
    Log in 查看库存
    Log in 查看库存
    1 * 10 件
    1 * 10 件
    规格:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLA40610-01
    型号
    40610-01
    Orifice dimensions
    4,763 mm (0,1875")
    灭菌
    Ethylene oxide (ETO) or gamma
    质保
    Out of box
    CE certificate
  • 36
    Fitting, Polypropylene, Straight, Hose Barb Union, ⅜" ID; 10/PK
    ⅜"
    ⅜"
    Barbed Union
    Polypropylene
    Straight
    Log in 查看库存
    Log in 查看库存
    1 * 10 件
    1 * 10 件
    规格:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLA40610-02
    型号
    40610-02
    Orifice dimensions
    6,35 mm (0,25")
    灭菌
    Ethylene oxide (ETO) or gamma
    质保
    Out of box
    CE certificate
  • 37
    Fitting, Polypropylene, Straight, Hose Barb Union, ½" ID; 10/PK
    ½"
    ½"
    Barbed Union
    Polypropylene
    Straight
    Log in 查看库存
    Log in 查看库存
    1 * 10 件
    1 * 10 件
    规格:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLA40610-03
    型号
    40610-03
    Orifice dimensions
    8,73 mm (0,34375")
    灭菌
    Ethylene oxide (ETO) or gamma
    质保
    Out of box
    CE certificate
  • 38
    Fitting, Polypropylene, Straight, Hose Barb Union, ⅝" ID; 10/PK
    ⅝"
    ⅝"
    Barbed Union
    Polypropylene
    Straight
    Log in 查看库存
    Log in 查看库存
    1 * 10 件
    1 * 10 件
    规格:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLA40610-04
    型号
    40610-04
    Orifice dimensions
    12,7 mm (0,5")
    灭菌
    Ethylene oxide (ETO) or gamma
    质保
    Out of box
    CE certificate
  • 39
    Fitting, Polypropylene, Straight, Hose Barb Union, ¾" ID; 5/PK
    ¾"
    ¾"
    Barbed Union
    Polypropylene
    Straight
    Log in 查看库存
    Log in 查看库存
    1 * 5 件
    1 * 5 件
    规格:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLA40610-05
    型号
    40610-05
    Orifice dimensions
    14,29 mm (0,5625")
    灭菌
    Ethylene oxide (ETO) or gamma
    质保
    Out of box
    CE certificate
  • 40
    Fitting, Polypropylene, Straight, Hose Barb Union, 1" ID; 5/PK
    1"
    1"
    Barbed Union
    Polypropylene
    Straight
    Log in 查看库存
    Log in 查看库存
    1 * 5 件
    1 * 5 件
    规格:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLA40610-06
    型号
    40610-06
    Orifice dimensions
    19,84 mm (0,78125")
    灭菌
    Ethylene oxide (ETO) or gamma
    质保
    Out of box
    CE certificate
  • 43
    Fitting, Polypropylene, Straight, Hose Barb Union, ⁵⁄₃₂"; 10/PK
    ⁵⁄₃₂"
    ⁵⁄₃₂"
    Barbed Union
    Polypropylene
    Straight
    Log in 查看库存
    Log in 查看库存
    1 * 10 件
    1 * 10 件
    规格:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLA40622-52
    型号
    40622-52
    Orifice dimensions
    灭菌
    Ethylene oxide (ETO) or gamma
    质保
    CE certificate
  • 44
    Fitting, Polypropylene, Straight, Hose Barb Union, ³⁄₁₆" ID; 10/PK
    ³⁄₁₆"
    ³⁄₁₆"
    Barbed Union
    Polypropylene
    Straight
    Log in 查看库存
    Log in 查看库存
    1 * 10 件
    1 * 10 件
    规格:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLA40622-57
    型号
    40622-57
    Orifice dimensions
    灭菌
    Ethylene oxide (ETO) or gamma
    质保
    CE certificate
  • 45
    Fitting, Polypropylene, Straight, Hose Barb Union, ⅜" ID; 100/PK
    ⅜"
    ⅜"
    Barbed Union
    Polypropylene
    Straight
    Log in 查看库存
    Log in 查看库存
    1 * 100 件
    1 * 100 件
    规格:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLA40625-91
    型号
    40625-91
    Orifice dimensions
    灭菌
    Ethylene oxide (ETO) or gamma
    质保
    CE certificate
  • 46
    Fitting, Polypropylene, Straight, Hose Barb Union, ¼" ID; 100/PK
    ¼"
    ¼"
    Barbed Union
    Polypropylene
    Straight
    Log in 查看库存
    Log in 查看库存
    1 * 100 件
    1 * 100 件
    规格:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLA40640-00
    型号
    40640-00
    Orifice dimensions
    0,125"
    灭菌
    Ethylene oxide (ETO) or gamma
    质保
    Out of box
    CE certificate
  • 47
    Fitting, Polypropylene, Straight, Hose Barb Union, ⁵⁄₁₆" ID; 100/PK
    ⁵⁄₁₆"
    ⁵⁄₁₆"
    Barbed Union
    Polypropylene
    Straight
    Log in 查看库存
    Log in 查看库存
    1 * 100 件
    1 * 100 件
    规格:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLA40640-01
    型号
    40640-01
    Orifice dimensions
    0,1875"
    灭菌
    Ethylene oxide (ETO) or gamma
    质保
    Out of box
    CE certificate
  • 49
    Fitting, Polypropylene, Straight, Hose Barb Union, ½" ID, 100/PK
    ½"
    ½"
    Barbed Union
    Polypropylene
    Straight
    Log in 查看库存
    Log in 查看库存
    1 * 100 件
    1 * 100 件
    规格:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLA40640-03
    型号
    40640-03
    Orifice dimensions
    0,34375"
    灭菌
    Ethylene oxide (ETO) or gamma
    质保
    Out of box
    CE certificate
  • 50
    Fitting, Polypropylene, Straight, Hose Barb Union, ¾" ID; 50/PK
    ¾"
    ¾"
    Barbed Union
    Polypropylene
    Straight
    Log in 查看库存
    Log in 查看库存
    1 * 50 件
    1 * 50 件
    规格:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLA40640-05
    型号
    40640-05
    Orifice dimensions
    0,5625"
    灭菌
    Ethylene oxide (ETO) or gamma
    质保
    Out of box
    CE certificate
  • 51
    Fitting, Polypropylene, Straight, Hose Barb Union, 1" ID; 50/PK
    1"
    1"
    Barbed Union
    Polypropylene
    Straight
    Log in 查看库存
    Log in 查看库存
    1 * 50 件
    1 * 50 件
    规格:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLA40640-06
    型号
    40640-06
    Orifice dimensions
    0,78125"
    灭菌
    Ethylene oxide (ETO) or gamma
    质保
    Out of box
    CE certificate
  • 52
    Fitting, Polypropylene, Straight, Hose Barb Union, ³⁄₃₂" ID; 10/PK
    ³⁄₃₂"
    ³⁄₃₂"
    Barbed Union
    Polypropylene
    Straight
    Log in 查看库存
    Log in 查看库存
    1 * 10 件
    1 * 10 件
    规格:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLA50622-46
    型号
    50622-46
    Orifice dimensions
    灭菌
    Ethylene Oxide (ETO) or Gamma
    质保
    CE certificate
  • 53
    Fitting, Polypropylene, Straight, Hose Barb Union, ¹⁄₁₆" ID; 10/PK
    ¹⁄₁₆"
    ¹⁄₁₆"
    Barbed Union
    Polypropylene
    Straight
    Log in 查看库存
    Log in 查看库存
    1 * 10 件
    1 * 10 件
    规格:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLA50622-49
    型号
    50622-49
    Orifice dimensions
    灭菌
    Ethylene Oxide (ETO) or Gamma
    质保
    CE certificate
  • 54
    Fitting, Polypropylene, Straight, Hose Barb Union, ³⁄₃₂" ID; 100/PK
    ³⁄₃₂"
    ³⁄₃₂"
    Barbed Union
    Polypropylene
    Straight
    Log in 查看库存
    Log in 查看库存
    1 * 100 件
    1 * 100 件
    规格:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLA50626-46
    型号
    50626-46
    Orifice dimensions
    灭菌
    Ethylene Oxide (ETO) or Autoclave
    质保
    CE certificate
  • 55
    Fitting, Polypropylene, Straight, Hose Barb Union, ¹⁄₁₆" ID; 100/PK
    ¹⁄₁₆"
    ¹⁄₁₆"
    Barbed Union
    Polypropylene
    Straight
    Log in 查看库存
    Log in 查看库存
    1 * 100 件
    1 * 100 件
    规格:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLA50626-49
    型号
    50626-49
    Orifice dimensions
    灭菌
    Ethylene Oxide (ETO) or Gamma
    质保
    CE certificate
  • 56
    Fitting, Polypropylene, Straight, Hose Barb Union, ⅛" ID; 10/PK
    ⅛"
    ⅛"
    Barbed Union
    Polypropylene
    Straight
    Log in 查看库存
    Log in 查看库存
    1 * 10 件
    1 * 10 件
    规格:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLA51518-05
    型号
    51518-05
    Orifice dimensions
    灭菌
    Ethylene Oxide (ETO)
    质保
    CE certificate