VWR®, Centrifuge Tubes, Conical Bottom, PP
Supplier: VWR Collection
Certificates
About this item
Conical bottom centrifuge tubes are made from high grade, gamma resistant polypropylene.
- Easy to read engraved graduations
- Autoclavable at 121 °C and freezable to −196 °C
- Leakproof and latex-free
- 3 year shelf life
- Manufactured in a Class 100 000 cleanroom environment
The tubes feature a flat seal cap made from polyethylene. Sterilised by gamma irradiation to SAL 10⁻⁶ (ISO 11137).
Certifications: Meets the requirements of BSE/TSE. Certified DNase-/RNase-free and non pyrogenic.
Caution: Height of 250 ml tubes has been changed from November 6th 2022, lot number: 221106334A.
Specifications
- Catalog No:
- VWRI525-1597
- VWRI525-1598
- VWRI525-1300
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- 环境认证:
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M