Syringe filters, non-sterile, Whatman™ Anotop
Supplier: CYTIVA WHATMAN
Certificates
About this item
Whatman Anotop non sterile syringe filters from Cytiva feature an inorganic Anopore membrane in a polypropylene housing.
- Aluminum-based inorganic Anopore membrane available in three pore sizes
- Effective with most organic solvents and aqueous samples
- Low hold-up volume (< 20 μl for Anotop 10) for maximum sample recovery
- Low protein binding filter minimizes protein loss
- Capillary pore structure with narrow pore-size distribution
- Recyclable after use. Learn how you can recycle it
The Anotop aluminum-based Anopore membrane is compatible with most organic solvents and aqueous samples. Aluminum-based membranes are inorganic, designed for chemical inertness and functionality in a wide range of temperatures. This syringe filter is commonly used in sample preparation for analytical instrumentation.
Unlike most organic membranes, the aluminum Anopore membrane has highly defined capillary pore structures and narrow pore-size distribution. This structure enables a high level of consistency in particle retention.
Specifications
- Catalog No:
- WHAT6809-1012
- WHAT6809-1022
- WHAT6809-2002
- WHAT6809-2012
- WHAT6809-1002
- WHAT6809-2022
- WHAT6809-2024
- 类型:
- Anotop™ 10
- Anotop™ 10
- Anotop™ 25
- Anotop™ 25
- Anotop™ 10
- Anotop™ 25
- Anotop™ 25
- 直径:
- 10 mm
- 10 mm
- 25 mm
- 25 mm
- 10 mm
- 25 mm
- 25 mm
- 尺寸:
- 15,4×18,5 mm
- 15,4×18,5 mm
- 36,8×26,3 mm
- 36,8×26,3 mm
- 15,4×18,5 mm
- 36,8×26,3 mm
- 36,8×26,3 mm
- 最大运行压力:
- 6,9 bar
- 6,9 bar
- 6,9 bar
- 6,9 bar
- 6,9 bar
- 6,9 bar
- 6,9 bar
- 前置过滤器:
- -
- -
- -
- -
- -
- -
- -