ContactUsLinkComponent
Autoclaves, horizontal and vertical, D-Line series
Autoclaves, horizontal and vertical, D-Line series
Catalog # TUTT2840EL
Supplier:  TUTTNAUER
undefined
Autoclaves, horizontal and vertical, D-Line series
Catalog # TUTT2840EL
Supplier:  TUTTNAUER
Specifications
  • 容量:
    28 L
  • 温度:
    134 °C
  • 说明:
    Autoclave, front loading, 2840 EL-D, with advanced microprocessor control panel
  • 顺序:
    02
  • 型号:
    2840 EL-D
  • 内部直径×高:
    280×400 mm
  • 重量:
    55 kg
  • 外部宽度×深度×高度:
    530×630×440 mm
  • 包装规格:
    1
Specifications
About this item
Choosing the right steam steriliser depends on several considerations; load diversity, frequency of use, available services and load volumes. The Tuttnauer line of vertical and benchtop sterilisers for the life sciences successfully meets the challenges posed today in laboratories and research institutes. The chamber is constructed of long lasting 316 L/Ti grade stainless steel with superior corrosion resistance, the generator is made from stainless steel.
  • The autoclaves are designed for easy servicing allowing maintenance access to all components
  • The microprocessor control system offers enhanced monitoring and control
  • Programs are designed to treat all load types including liquids, instruments and waste
  • All programs are fully customisable to suit the specific needs and requirements of the end user
  • Tuttnauer's D-Line models use multicolour display technology
  • The User Interface (UI) has been redesigned with the following considerations; multicolour display for easier reading, colour is used to indicate the stage of the cycle, easy operation, built-in view of historical cycle data, graphical display of temperature and pressure trend graphs
Safety features: A safety device prevents the operator from opening the door when chamber is pressurised; steam is not allowed to enter the chamber when the door is open; a cycle cannot start if the door is open or not properly locked; the door will not open until the temperature is below a specified safe level.
Double independent monitoring: The combined electronic and mechanical monitoring guarantees accurate results. The operator has two independent means to monitor temperature and pressure.
Reduced cycle times with optional fast cooling:
Tuttnauer’s accelerated cooling technology reduces cycle time and protects the load by minimising its exposure to high temperatures. Rapid cooling typically reduces cooling time by as much as 75%.
Fan assisted cooling:
An additional feature that further enhances the cooling stage is the optional radial ventilator. The fan circulates the hot air, inside the chamber, transferring the heat to the cooled chamber walls or cooling coils. The overall chamber and load temperature rapidly drops.
Efficient air removal by optional vacuum:
The vacuum pump is optional and if not selected the gravity displacement air removal method is used. Fractioned pre-vacuum air removal eliminates air pockets from all load types and maximises steam penetration. A pre-vacuum pump removes the air from the chamber. This allows for complete air removal with more efficient steam penetration throughout the entire load.
Optional bio-hazardous waste sterilisation:
The Tuttnauer line of advanced laboratory autoclaves supplies a full sterilisation solution for the treatment of bio-hazardous waste. The autoclaves can be supplied with an optional exhaust filtration system. During the air removal stage, prior to sterilisation, all effluent is passed through a biological filter that provides an extra level of protection. During the sterilisation phase, condensate does not leave the autoclave chamber. It is removed only after the sterilisation phase is complete. The laboratory staff and environment are therefore protected from contamination.
Caution: Please contact your local Avantor office to clarify the the correct model / features that suit your application.