Orders LinkContactUsLinkComponent
Level indicators, b.safe
Level indicators, b.safe
供应商:   BOHLENDER
Level indicators, b.safe
供应商:  BOHLENDER
Certificates
Level indicators, b.safe
Level indicators, b.safe
8 Options Available Below
Product Details & Documents
Insert with screw-in thread for barrels or screw cap for canisters. With integrated level indicator to monitor the liquid level during filling as well as a neck with GL 45 thread for the assembly of a b.safe funnel with GL 45 thread.
  • Prevent the waste canisters from overflowing
  • Available for canisters and barrels
  • Suitable for flammable liquid
The red visual pin goes up with increasing filling level. As soon as the visual pin fills the inspection glass completely, the canister is full and has to be exchanged.
Use of a b.safe funnel for filling of liquids and monitoring of the filling level at the same time to avoid run over.
Product Family Options
产品信息
  • 顺序
    说明
    材质
    长度
    用于
    直径
    可用库存
    Price
  • 01
    Level indicator with GL 45 funnel connection, static dissipative
    聚乙烯
    Female G2" / Tri-Sure 2"
    — mm
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 01
    Level indicator for barrels
    聚四氟乙烯/全氟烷氧基树脂
    86 mm
    Female G³/₄"
    23 mm
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 02
    Level indicator with GL 45 funnel connection, static dissipative
    聚乙烯
    Female Mauser 2"
    — mm
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 02
    Level indicator for barrels
    聚乙烯
    86 mm
    Female G³/₄"
    23 mm
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 03
    Level indicator with GL 45 funnel connection, static dissipative
    聚乙烯
    Male S 60
    — mm
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 06
    Level indicator for barrels
    聚乙烯
    238 mm
    Opening Ø 50 - 100 mm
    50 mm
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 07
    Level indicator for barrels
    聚乙烯
    250 mm
    Dairy pipe thread
    31 mm
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 08
    Level indicator for barrels, static dissipative
    聚乙烯
    86 mm
    Female G³/₄"
    23 mm