Orders LinkContactUsLinkComponent
Microscope slides, Corning®
Microscope slides, Corning®
01, Plain slides, 无色, 玻璃, 75×25 mm
目录编号 CORN2947-75X25
Microscope slides, Corning®
目录编号 CORN2947-75X25
Microscope slides, Corning®
Microscope slides, Corning®
01, Plain slides, 无色, 玻璃, 75×25 mm
Jump to:
Price
询价
Log in 查看库存
1 * 1.440 件
Product Details & Documents
These slides are either frosted on one side, on both sides, or plain.
  • Clarity reinforced water-white glass
  • Frosted slides are sandblasted, easy to write on and highly legible
  • Each slide is inspected for chips and rough edge before being shipped
Documents
Certificates
按批号搜索
规格
  • 颜色:
    无色
  • 说明:
    Plain slides
  • 长度×宽度:
    75×25 mm
  • 材质:
    玻璃
  • 顺序:
    01