磁力加热板搅拌器,Advanced系列
关于此项目
VWR® Advanced magnetic hotplate stirrers deliver accurate, repeatable results. Units are microprocessor controlled and have an easy to read LED display for set point temperature. The control panel features easy to use controls which allow users to dial in adjustments for temperature, the stirrer control dial has rpm adjustment markings. Rear housing features an integral support rod holder with locking knob to accept an optional support rod and clamp kit.
- Excellent temperature uniformity with consistent stirring at all speeds
- Microprocessor controlled with built-in memory allowing recall of last set temperature (even if unit has been turned off)
- Cool touch, chemically resistant housing
- LED display for temperature
- Choice of models with ceramic or aluminium top plates
The low profile design takes up less space and fits into fume hoods. Spill-resistant housing channels fluids away from internal components. Safety features include a 'hot' symbol warning light which is illuminated when heat is turned on and remains on until top plate cools down. Stir protection if stirrer motor stops or fails, unit will automatically shut down the heater. Enhanced electronics regulate both heating and stirring and bring samples to temperature quickly and efficiently. Temperature measurement is controlled more accurately by using a resistance thermometer (RTD). Ramping feature slowly increases speed for improved safety and enhanced coupling, avoids splashing, improves spin bar control, and provides excellent low end speed control. Ceramic tops feature a chemically resistant, reflective white top plate surface that is easy to clean. Durable aluminium tops will not crack or chip, and provide a more even heating surface.
Delivery information: Each unit is supplied complete with a PTFE coated magnetic stirring bar. Optional support rod kit is available which includes a 457 mm stainless steel support rod, thermometer/temperature probe extension clamp, three-prong medium swivel clamp and hook connector.
Caution: * Below 100 °C ±2 °C environmental and sample conditions permitting.
1 Options Available Below
Product Details & Documents
VWR® Advanced magnetic hotplate stirrers deliver accurate, repeatable results. Units are microprocessor controlled and have an easy to read LED display for set point temperature. The control panel features easy to use controls which allow users to dial in adjustments for temperature, the stirrer control dial has rpm adjustment markings. Rear housing features an integral support rod holder with locking knob to accept an optional support rod and clamp kit.
- Excellent temperature uniformity with consistent stirring at all speeds
- Microprocessor controlled with built-in memory allowing recall of last set temperature (even if unit has been turned off)
- Cool touch, chemically resistant housing
- LED display for temperature
- Choice of models with ceramic or aluminium top plates
The low profile design takes up less space and fits into fume hoods. Spill-resistant housing channels fluids away from internal components. Safety features include a 'hot' symbol warning light which is illuminated when heat is turned on and remains on until top plate cools down. Stir protection if stirrer motor stops or fails, unit will automatically shut down the heater. Enhanced electronics regulate both heating and stirring and bring samples to temperature quickly and efficiently. Temperature measurement is controlled more accurately by using a resistance thermometer (RTD). Ramping feature slowly increases speed for improved safety and enhanced coupling, avoids splashing, improves spin bar control, and provides excellent low end speed control. Ceramic tops feature a chemically resistant, reflective white top plate surface that is easy to clean. Durable aluminium tops will not crack or chip, and provide a more even heating surface.
Delivery information: Each unit is supplied complete with a PTFE coated magnetic stirring bar. Optional support rod kit is available which includes a 457 mm stainless steel support rod, thermometer/temperature probe extension clamp, three-prong medium swivel clamp and hook connector.
Caution: * Below 100 °C ±2 °C environmental and sample conditions permitting.
Specifications for Product Family
规格
- Catalog No:
- VWRI444-0596
- VWRI444-0598
- VWRI444-0608
- VWRI444-0593
- VWRI444-0595
- VWRI444-0599
- 型号:
- 铝制平台
- 铝制平台
- 铝制平台
- 陶瓷平台
- 陶瓷平台
- 陶瓷平台
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing
- Waste Reducing
- 板材料:
- 铝
- 铝
- 铝
- 陶瓷
- 陶瓷
- 陶瓷
- 板尺寸:
- 100×100 mm
- 100×100 mm
- 250×250 mm
- 100×100 mm
- 100×100 mm
- 180×180 mm
- 最大搅拌容量 水:
- 600 ml
- 600 ml
- 6 L
- 600 ml
- 600 ml
- 2,5 L
- 速度范围:
- 60 - 1600 min⁻¹
- 60 - 1600 min⁻¹
- 60 - 1600 min⁻¹
- 60 - 1600 min⁻¹
- 60 - 1600 min⁻¹
- 60 - 1600 min⁻¹
- 速度稳定性:
- ±2%
- ±2%
- ±2%
- ±2%
- ±2%
- ±2%
- 宽度x深度x高度:
- 167×274×108 mm
- 167×274×108 mm
- 330×455×108 mm
- 167×274×108 mm
- 167×274×108 mm
- 250×375×108 mm
- 温度稳定性:
- ±2 %*
- ±2 %*
- ±2 %*
- ±3 %*
- ±3 %*
- ±3 %*
- 温度范围:
- 环境温度+5....400 °C
- 环境温度+5....400 °C
- 环境温度+5....400 °C
- 环境温度+5....500 °C
- 环境温度+5....500 °C
- 环境温度+5....500 °C
- 热输出 (W):
- 400 W
- 400 W
- 1600 W
- 400 W
- 400 W
- 1050 W
- 重量:
- 2,0 kg
- 2,0 kg
- 6 kg
- 2,0 kg
- 2,0 kg
- 4,2 kg
规格
Product Family Options
带铝顶板的型号
产品信息
- 顺序说明可用库存Price
- 001磁力加热板搅拌器,铝制平台,100×100 mm,欧标插头Discontinued-规格:
Waste Reducing More
Specifications
Cat. No.VWRI444-0596型号铝制平台Environmentally PreferableWaste Reducing板材料铝板尺寸100×100 mm最大搅拌容量 水600 ml速度范围60 - 1600 min⁻¹速度稳定性±2%宽度x深度x高度167×274×108 mm温度稳定性±2 %*温度范围环境温度+5....400 °C热输出 (W)400 W重量2,0 kgWaste Reducing - 003磁力加热板搅拌器,铝制平台,100×100 mm,瑞士插头Discontinued-规格:
Waste Reducing More
Specifications
Cat. No.VWRI444-0598型号铝制平台Environmentally PreferableWaste Reducing板材料铝板尺寸100×100 mm最大搅拌容量 水600 ml速度范围60 - 1600 min⁻¹速度稳定性±2%宽度x深度x高度167×274×108 mm温度稳定性±2 %*温度范围环境温度+5....400 °C热输出 (W)400 W重量2,0 kgWaste Reducing - 规格:
Waste Reducing More
Specifications
Cat. No.VWRI444-0608型号铝制平台Environmentally PreferableWaste Reducing板材料铝板尺寸250×250 mm最大搅拌容量 水6 L速度范围60 - 1600 min⁻¹速度稳定性±2%宽度x深度x高度330×455×108 mm温度稳定性±2 %*温度范围环境温度+5....400 °C热输出 (W)1600 W重量6 kgWaste Reducing
Accessories
产品信息
- 目录编号说明用于可用库存Price
- VWRU11301-110搅拌器支撑杆夹具套件(支撑杆,2个夹具和夹钩连接器)444-0566, 444-0567, 444-0568,444-0569, 444-0570, 444-0571, 444-0572, 444-0573, 444-0574, 444-0575, 444-0576, 444-0577, 444-0578, 444-0579, 444-0580, 444-0581, 444-0582, 444-0583, 444-0584, 444-0585, 444-0586, 444-0587, 444-0588, 444-0589, 444-0590, 444-0591, 444-0592, 444-0593, 444-0594, 444-0595, 444-0596, 444-0597, 444-0598, 444-0599, 444-0600, 444-0601, 444-0602, 444-0603, 444-0604, 444-0605, 444-0606, 444-0607, 444-0608, 444-0609, 444-0610Log in 查看库存1 * 1 件 - ¥2,435.00
产品信息
- 目录编号说明用于可用库存Price
- VWRI444-2851100 mm搅拌器和热板搅拌器用支撑板444-0566, 444-0567, 444-0568,444-0575, 444-0576, 444-0577, 444-0578, 444-0579, 444-0580, 444-0593, 444-0594, 444-0595, 444-0596, 444-0597, 444-0598Log in 查看库存1 * 1 件 - ¥5,983.00
产品信息
- 目录编号说明用于可用库存Price
- VWRI444-2844搅拌器支撑杆夹具套件(支撑杆,2个夹具和夹钩连接器)444-0566, 444-0567, 444-0568,444-0569, 444-0570, 444-0571, 444-0572, 444-0573, 444-0574, 444-0575, 444-0576, 444-0577, 444-0578, 444-0579, 444-0580, 444-0581, 444-0582, 444-0583, 444-0584, 444-0585, 444-0586, 444-0587, 444-0588, 444-0589, 444-0590, 444-0591, 444-0592, 444-0593, 444-0594, 444-0595, 444-0596, 444-0597, 444-0598, 444-0599, 444-0600, 444-0601, 444-0602, 444-0603, 444-0604, 444-0605, 444-0606, 444-0607, 444-0608, 444-0609, 444-0610Log in 查看库存1 * 1 件 - ¥2,941.00