跳转到 页眉
跳转到 主要内容
跳转到 页脚
高级搜索
SDS
Certificates
制造商数据
0
¥0.00
订单输入
登录
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
您的账户
Cart
China
Order Entry
Order Status
您的账户
Cart
Orders Link
ContactUsLinkComponent
Home
缓冲(液)
组织培养缓冲液
Phosphate Buffered Saline
Phosphate Buffered Saline
Print
Phosphate Buffered Saline
Phosphate buffered saline tablets, 100 ml, 100
目录编号 OXOIBR0014G
Certificates
供应商:
THERMO FISHER DIAGNOSTICS
Phosphate Buffered Saline
目录编号 OXOIBR0014G
供应商:
THERMO FISHER DIAGNOSTICS
CAS号:
Certificates
展开图片
Return to Top
Phosphate Buffered Saline
Phosphate buffered saline tablets, 100 ml, 100
规格
Add to cart
Jump to:
规格
CAS号:
Price
询价
Log in
查看库存
规格
1 * 100 件
-
+
询价
Return Policy
Product Details & Documents
Documents
Certificates
按批号搜索
规格
规格
包装规格:
100
说明:
Phosphate buffered saline tablets
填充:
100 ml
规格