Avantor® Hichrom HI-PAH 2, GC Columns
Supplier: Avantor
Certificates
About this item
Application-specific column for polycyclic aromatics hydrocarbons.
Max. temperature limits may change depending on the film thickness.
Specifications
- Catalog No:
- HIGCHI49-15-010-15
- HIGCHI49-18-010-40
- HIGCHI49-18-010-20
- HIGCHI49-25-010-60
- HIGCHI49-25-010-30
- Phase name:
- HI-PAH 2
- HI-PAH 2
- HI-PAH 2
- HI-PAH 2
- HI-PAH 2
- Phase composition:
- Proprietary Phase developed for EU PAHs analysis
- Proprietary Phase developed for EU PAHs analysis
- Proprietary Phase developed for EU PAHs analysis
- Proprietary Phase developed for EU PAHs analysis
- Proprietary Phase developed for EU PAHs analysis
- Crossbond:
- Yes
- Yes
- Yes
- Yes
- Yes
- 温度限值:
- 350 °C
- 350 °C
- 350 °C
- 350 °C
- 350 °C